Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232531232-232531658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRND
Entrez Gene ID
1144
Gene Name
cholinergic receptor nicotinic delta subunit
Gene Aliases
ACHRD, CMS2A, CMS3A, CMS3B, CMS3C, FCCMS, SCCMS
UniGene
Hs.156289
Gene Chromosome Location
Chr.2: 232526160-232537907 on Build GRCh38
Transcripts
NM_000751.2 NM_001256657.1 NM_001311195.1 NM_001311196.1 XM_011510524.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187695 c.941T>A p.L314H Chr.2: 232531550-232531550 on Build GRCh38
3579107 c.831G>A p.K277K Chr.2: 232531362-232531362 on Build GRCh38
720239 c.896T>A p.L299Q Chr.2: 232531427-232531427 on Build GRCh38
3579109 c.934T>C p.F312L Chr.2: 232531543-232531543 on Build GRCh38
4092407 c.966G>A p.M322I Chr.2: 232531575-232531575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920100609 Chr.2: 232531536-232531536 on Build GRCh38
rs748649589 Chr.2: 232531549-232531549 on Build GRCh38
rs774786826 Chr.2: 232531292-232531292 on Build GRCh38
rs745432322 Chr.2: 232531572-232531572 on Build GRCh38
rs766506210 Chr.2: 232531532-232531532 on Build GRCh38
rs748559696 Chr.2: 232531465-232531465 on Build GRCh38
rs199592458 Chr.2: 232531533-232531533 on Build GRCh38
rs901666285 Chr.2: 232531382-232531382 on Build GRCh38
rs374817005 Chr.2: 232531458-232531458 on Build GRCh38
rs1048192152 Chr.2: 232531538-232531538 on Build GRCh38
rs769282027 Chr.2: 232531574-232531574 on Build GRCh38
rs576315583 Chr.2: 232531609-232531609 on Build GRCh38
rs746002657 Chr.2: 232531353-232531353 on Build GRCh38
rs750664390 Chr.2: 232531526-232531526 on Build GRCh38
rs758862277 Chr.2: 232531550-232531550 on Build GRCh38
rs370309294 Chr.2: 232531473-232531473 on Build GRCh38
rs2853452 Chr.2: 232531470-232531470 on Build GRCh38
rs750598323 Chr.2: 232531448-232531448 on Build GRCh38
rs531329199 Chr.2: 232531557-232531557 on Build GRCh38
rs797045474 Chr.2: 232531468-232531468 on Build GRCh38
rs779641797 Chr.2: 232531323-232531334 on Build GRCh38
rs113196165 Chr.2: 232531299-232531299 on Build GRCh38
rs771670415 Chr.2: 232531459-232531459 on Build GRCh38
rs139722128 Chr.2: 232531424-232531424 on Build GRCh38
rs894195871 Chr.2: 232531516-232531516 on Build GRCh38
rs530117757 Chr.2: 232531265-232531299 on Build GRCh38
rs749757968 Chr.2: 232531380-232531380 on Build GRCh38
rs772540464 Chr.2: 232531466-232531466 on Build GRCh38
rs774802325 Chr.2: 232531388-232531388 on Build GRCh38
rs201702120 Chr.2: 232531325-232531325 on Build GRCh38
rs780672505 Chr.2: 232531379-232531379 on Build GRCh38
rs948279065 Chr.2: 232531255-232531255 on Build GRCh38
rs139190636 Chr.2: 232531555-232531555 on Build GRCh38
rs553886556 Chr.2: 232531317-232531317 on Build GRCh38
rs761495130 Chr.2: 232531308-232531308 on Build GRCh38
rs145311396 Chr.2: 232531606-232531606 on Build GRCh38
rs951291427 Chr.2: 232531309-232531309 on Build GRCh38
rs373187583 Chr.2: 232531354-232531354 on Build GRCh38
rs775473919 Chr.2: 232531576-232531576 on Build GRCh38
rs367837942 Chr.2: 232531472-232531472 on Build GRCh38
rs867211733 Chr.2: 232531489-232531489 on Build GRCh38
rs377455260 Chr.2: 232531423-232531423 on Build GRCh38
rs755073623 Chr.2: 232531545-232531545 on Build GRCh38
rs886038365 Chr.2: 232531300-232531334 on Build GRCh38
rs1026784533 Chr.2: 232531303-232531303 on Build GRCh38
rs187055404 Chr.2: 232531421-232531421 on Build GRCh38
rs763796108 Chr.2: 232531504-232531504 on Build GRCh38
rs771806326 Chr.2: 232531399-232531399 on Build GRCh38
rs749895309 Chr.2: 232531312-232531312 on Build GRCh38
rs756237105 Chr.2: 232531529-232531529 on Build GRCh38
rs763965619 Chr.2: 232531593-232531593 on Build GRCh38
rs41265127 Chr.2: 232531393-232531393 on Build GRCh38
rs1057518958 Chr.2: 232531353-232531353 on Build GRCh38
rs757411934 Chr.2: 232531347-232531347 on Build GRCh38
rs995293066 Chr.2: 232531289-232531289 on Build GRCh38
rs894269607 Chr.2: 232531263-232531263 on Build GRCh38
rs1044534360 Chr.2: 232531256-232531256 on Build GRCh38
rs775383951 Chr.2: 232531484-232531484 on Build GRCh38
rs779609809 Chr.2: 232531547-232531547 on Build GRCh38
rs762875734 Chr.2: 232531350-232531350 on Build GRCh38
rs121909502 Chr.2: 232531397-232531397 on Build GRCh38
rs778673027 Chr.2: 232531326-232531326 on Build GRCh38
rs201370782 Chr.2: 232531446-232531446 on Build GRCh38
rs760991691 Chr.2: 232531602-232531602 on Build GRCh38
rs372983474 Chr.2: 232531314-232531314 on Build GRCh38
rs114022008 Chr.2: 232531483-232531483 on Build GRCh38
rs375623674 Chr.2: 232531616-232531616 on Build GRCh38
rs374563037 Chr.2: 232531480-232531480 on Build GRCh38
rs561839978 Chr.2: 232531435-232531435 on Build GRCh38
rs765029351 Chr.2: 232531428-232531428 on Build GRCh38
rs142063328 Chr.2: 232531450-232531450 on Build GRCh38
rs765400853 Chr.2: 232531629-232531629 on Build GRCh38
rs200290614 Chr.2: 232531415-232531415 on Build GRCh38
rs759490772 Chr.2: 232531482-232531482 on Build GRCh38
rs767153407 Chr.2: 232531310-232531310 on Build GRCh38
rs374944586 Chr.2: 232531301-232531301 on Build GRCh38
rs777931139 Chr.2: 232531554-232531554 on Build GRCh38
rs762813704 Chr.2: 232531591-232531591 on Build GRCh38
rs747361396 Chr.2: 232531492-232531492 on Build GRCh38
rs145981548 Chr.2: 232531430-232531430 on Build GRCh38
rs754087173 Chr.2: 232531615-232531615 on Build GRCh38
rs371376389 Chr.2: 232531451-232531451 on Build GRCh38
rs759403975 Chr.2: 232531479-232531479 on Build GRCh38
rs769150854 Chr.2: 232531381-232531381 on Build GRCh38
rs548284269 Chr.2: 232531264-232531264 on Build GRCh38
rs773972915 Chr.2: 232531597-232531597 on Build GRCh38
rs751784704 Chr.2: 232531344-232531344 on Build GRCh38
rs776316455 Chr.2: 232531571-232531571 on Build GRCh38

View Full Product Details