Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112737818-112738338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP2L
Entrez Gene ID
150468
Gene Name
cytoskeleton associated protein 2 like
Gene Aliases
-
UniGene
Hs.434250
Gene Chromosome Location
Chr.2: 112736349-112764677 on Build GRCh38
Transcripts
NM_001304361.1 NM_152515.4 NR_130712.1 XM_011510666.2 XM_011512262.2 XM_011512265.2 XM_017005474.1 XM_017005475.1 XM_017005476.1 XM_017005477.1 XM_017005478.1

Gene Symbol
NT5DC4
Entrez Gene ID
284958
Gene Name
5'-nucleotidase domain containing 4
Gene Aliases
-
UniGene
Hs.589714
Gene Chromosome Location
Chr.2: 112717245-112742879 on Build GRCh38
Transcripts
NM_001304361.1 NM_152515.4 NR_130712.1 XM_011510666.2 XM_011512262.2 XM_011512265.2 XM_017005474.1 XM_017005475.1 XM_017005476.1 XM_017005477.1 XM_017005478.1

SNP Detail

SNP ID Chromosome Location
rs549229413 Chr.2: 112738095-112738095 on Build GRCh38
rs974977477 Chr.2: 112738100-112738100 on Build GRCh38
rs184155476 Chr.2: 112738136-112738136 on Build GRCh38
rs2716671 Chr.2: 112738056-112738056 on Build GRCh38
rs149559633 Chr.2: 112738250-112738250 on Build GRCh38
rs554704754 Chr.2: 112738138-112738139 on Build GRCh38
rs1053323982 Chr.2: 112737990-112737990 on Build GRCh38
rs942235099 Chr.2: 112737888-112737891 on Build GRCh38
rs909387534 Chr.2: 112737877-112737877 on Build GRCh38
rs769334060 Chr.2: 112738231-112738231 on Build GRCh38
rs934503393 Chr.2: 112737964-112737964 on Build GRCh38
rs932332877 Chr.2: 112737915-112737915 on Build GRCh38
rs1016147802 Chr.2: 112737987-112737987 on Build GRCh38
rs1011880362 Chr.2: 112737993-112737993 on Build GRCh38
rs878911957 Chr.2: 112738120-112738120 on Build GRCh38
rs10206303 Chr.2: 112737868-112737868 on Build GRCh38
rs946184352 Chr.2: 112737847-112737847 on Build GRCh38
rs775849165 Chr.2: 112738255-112738257 on Build GRCh38
rs950830795 Chr.2: 112738054-112738054 on Build GRCh38
rs950912786 Chr.2: 112737860-112737863 on Build GRCh38
rs901910234 Chr.2: 112737890-112737890 on Build GRCh38
rs189855678 Chr.2: 112738137-112738137 on Build GRCh38
rs1006232280 Chr.2: 112737982-112737982 on Build GRCh38
rs1001995945 Chr.2: 112738313-112738313 on Build GRCh38
rs963590465 Chr.2: 112738098-112738098 on Build GRCh38
rs905820140 Chr.2: 112738288-112738288 on Build GRCh38
rs138177762 Chr.2: 112738126-112738126 on Build GRCh38
rs993469353 Chr.2: 112738223-112738223 on Build GRCh38
rs1050464946 Chr.2: 112738202-112738202 on Build GRCh38
rs535409974 Chr.2: 112738034-112738034 on Build GRCh38
rs957662264 Chr.2: 112738104-112738104 on Build GRCh38
rs2676122 Chr.2: 112738057-112738057 on Build GRCh38
rs889154257 Chr.2: 112737936-112737936 on Build GRCh38
rs980524982 Chr.2: 112738084-112738084 on Build GRCh38
rs112680000 Chr.2: 112737960-112737960 on Build GRCh38
rs897720582 Chr.2: 112738219-112738219 on Build GRCh38
rs542967543 Chr.2: 112738241-112738241 on Build GRCh38
rs181492903 Chr.2: 112738138-112738138 on Build GRCh38
rs989125978 Chr.2: 112738111-112738111 on Build GRCh38
rs997623407 Chr.2: 112737902-112737902 on Build GRCh38
rs181016968 Chr.2: 112738002-112738002 on Build GRCh38
rs1002870172 Chr.2: 112738023-112738023 on Build GRCh38
rs375913114 Chr.2: 112738024-112738024 on Build GRCh38
rs962003802 Chr.2: 112737998-112737998 on Build GRCh38
rs573553559 Chr.2: 112738209-112738209 on Build GRCh38
rs993706148 Chr.2: 112738000-112738000 on Build GRCh38
rs576564143 Chr.2: 112738291-112738291 on Build GRCh38
rs10209160 Chr.2: 112738208-112738208 on Build GRCh38
rs975008578 Chr.2: 112737890-112737894 on Build GRCh38
rs889204919 Chr.2: 112738203-112738203 on Build GRCh38
rs568970679 Chr.2: 112738123-112738123 on Build GRCh38
rs375252836 Chr.2: 112737991-112737991 on Build GRCh38
rs150416463 Chr.2: 112738029-112738029 on Build GRCh38
rs1028995051 Chr.2: 112737934-112737934 on Build GRCh38

View Full Product Details