Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43598389-43598925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT1B
Entrez Gene ID
1159
Gene Name
creatine kinase, mitochondrial 1B
Gene Aliases
CKMT, CKMT1, UMTCK
UniGene
Hs.741420
Gene Chromosome Location
Chr.15: 43592857-43599406 on Build GRCh38
Transcripts
NM_020990.4 NR_135748.1 NR_135749.1 NR_135750.1 NR_135751.1 NR_135752.1 XM_005254150.3 XM_011521194.1 XM_011521195.2 XM_011521196.1 XM_011521197.2 XM_011521198.1 XM_011521199.2 XM_017021902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1727316 c.1058G>A p.R353H Chr.15: 43598873-43598873 on Build GRCh38
5614224 c.1093G>T p.G365C Chr.15: 43598908-43598908 on Build GRCh38
1373057 c.1070G>A p.G357E Chr.15: 43598885-43598885 on Build GRCh38
1241018 c.1056A>G p.K352K Chr.15: 43598871-43598871 on Build GRCh38
6015754 c.1074G>A p.V358V Chr.15: 43598889-43598889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368122068 Chr.15: 43598830-43598830 on Build GRCh38
rs376023151 Chr.15: 43598889-43598889 on Build GRCh38
rs1031978788 Chr.15: 43598637-43598637 on Build GRCh38
rs371585335 Chr.15: 43598868-43598868 on Build GRCh38
rs374254621 Chr.15: 43598814-43598814 on Build GRCh38
rs764800222 Chr.15: 43598810-43598810 on Build GRCh38
rs932722903 Chr.15: 43598419-43598419 on Build GRCh38
rs186688987 Chr.15: 43598729-43598729 on Build GRCh38
rs974742255 Chr.15: 43598744-43598744 on Build GRCh38
rs768487664 Chr.15: 43598882-43598882 on Build GRCh38
rs752104222 Chr.15: 43598828-43598828 on Build GRCh38
rs868169473 Chr.15: 43598656-43598656 on Build GRCh38
rs551083664 Chr.15: 43598758-43598758 on Build GRCh38
rs571311617 Chr.15: 43598793-43598793 on Build GRCh38
rs912613433 Chr.15: 43598839-43598839 on Build GRCh38
rs577910518 Chr.15: 43598529-43598529 on Build GRCh38
rs542681063 Chr.15: 43598651-43598651 on Build GRCh38
rs13234 Chr.15: 43598901-43598901 on Build GRCh38
rs563498942 Chr.15: 43598626-43598626 on Build GRCh38
rs1000758407 Chr.15: 43598631-43598631 on Build GRCh38
rs764016501 Chr.15: 43598785-43598785 on Build GRCh38
rs971545337 Chr.15: 43598698-43598704 on Build GRCh38
rs919925045 Chr.15: 43598775-43598775 on Build GRCh38
rs565117058 Chr.15: 43598508-43598508 on Build GRCh38
rs772902861 Chr.15: 43598899-43598899 on Build GRCh38
rs9571 Chr.15: 43598871-43598871 on Build GRCh38
rs756293987 Chr.15: 43598838-43598838 on Build GRCh38
rs555601064 Chr.15: 43598421-43598421 on Build GRCh38
rs749354164 Chr.15: 43598857-43598857 on Build GRCh38
rs370436452 Chr.15: 43598823-43598823 on Build GRCh38
rs56368539 Chr.15: 43598704-43598704 on Build GRCh38
rs908712019 Chr.15: 43598724-43598724 on Build GRCh38
rs769390403 Chr.15: 43598897-43598897 on Build GRCh38
rs776417653 Chr.15: 43598888-43598888 on Build GRCh38
rs761467622 Chr.15: 43598800-43598800 on Build GRCh38
rs530779588 Chr.15: 43598834-43598834 on Build GRCh38
rs144997771 Chr.15: 43598873-43598873 on Build GRCh38
rs558172008 Chr.15: 43598519-43598519 on Build GRCh38
rs543706626 Chr.15: 43598555-43598555 on Build GRCh38
rs776131004 Chr.15: 43598794-43598794 on Build GRCh38
rs143381903 Chr.15: 43598833-43598833 on Build GRCh38
rs879209746 Chr.15: 43598506-43598506 on Build GRCh38
rs959329589 Chr.15: 43598640-43598640 on Build GRCh38
rs1020752910 Chr.15: 43598533-43598533 on Build GRCh38
rs572528695 Chr.15: 43598447-43598447 on Build GRCh38
rs529459726 Chr.15: 43598641-43598641 on Build GRCh38
rs183676883 Chr.15: 43598692-43598692 on Build GRCh38
rs778161700 Chr.15: 43598853-43598853 on Build GRCh38
rs747021769 Chr.15: 43598872-43598872 on Build GRCh38
rs981572667 Chr.15: 43598710-43598710 on Build GRCh38
rs771051027 Chr.15: 43598471-43598471 on Build GRCh38
rs367738721 Chr.15: 43598719-43598720 on Build GRCh38
rs1008049404 Chr.15: 43598524-43598524 on Build GRCh38
rs890960174 Chr.15: 43598516-43598516 on Build GRCh38
rs541120135 Chr.15: 43598715-43598715 on Build GRCh38

View Full Product Details