Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178605155-178605671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK4
Entrez Gene ID
57396
Gene Name
CDC like kinase 4
Gene Aliases
-
UniGene
Hs.406557
Gene Chromosome Location
Chr.5: 178602664-178627053 on Build GRCh38
Transcripts
NM_020666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409536 c.1192C>T p.Q398* Chr.5: 178605325-178605325 on Build GRCh38
1620123 c.1214+3A>G p.? Chr.5: 178605300-178605300 on Build GRCh38
3342107 c.1214+1G>A p.? Chr.5: 178605302-178605302 on Build GRCh38
5737446 c.1172G>A p.R391Q Chr.5: 178605345-178605345 on Build GRCh38
6630522 c.1171C>T p.R391* Chr.5: 178605346-178605346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181456729 Chr.5: 178605440-178605440 on Build GRCh38
rs780495111 Chr.5: 178605391-178605391 on Build GRCh38
rs370188010 Chr.5: 178605407-178605407 on Build GRCh38
rs932988793 Chr.5: 178605629-178605629 on Build GRCh38
rs531615836 Chr.5: 178605216-178605216 on Build GRCh38
rs142832595 Chr.5: 178605232-178605232 on Build GRCh38
rs776764994 Chr.5: 178605276-178605276 on Build GRCh38
rs924972274 Chr.5: 178605595-178605595 on Build GRCh38
rs374064650 Chr.5: 178605279-178605279 on Build GRCh38
rs935000119 Chr.5: 178605635-178605635 on Build GRCh38
rs373404202 Chr.5: 178605431-178605431 on Build GRCh38
rs754990853 Chr.5: 178605402-178605402 on Build GRCh38
rs759607570 Chr.5: 178605432-178605432 on Build GRCh38
rs759961032 Chr.5: 178605278-178605279 on Build GRCh38
rs759050698 Chr.5: 178605293-178605293 on Build GRCh38
rs960240161 Chr.5: 178605508-178605508 on Build GRCh38
rs765607741 Chr.5: 178605326-178605326 on Build GRCh38
rs780357156 Chr.5: 178605626-178605626 on Build GRCh38
rs773038549 Chr.5: 178605413-178605413 on Build GRCh38
rs756796951 Chr.5: 178605201-178605201 on Build GRCh38
rs935259588 Chr.5: 178605272-178605272 on Build GRCh38
rs769924199 Chr.5: 178605205-178605205 on Build GRCh38
rs974644474 Chr.5: 178605577-178605577 on Build GRCh38
rs375483389 Chr.5: 178605291-178605291 on Build GRCh38
rs901545511 Chr.5: 178605271-178605271 on Build GRCh38
rs770077676 Chr.5: 178605278-178605278 on Build GRCh38
rs530218875 Chr.5: 178605282-178605282 on Build GRCh38
rs369629452 Chr.5: 178605266-178605266 on Build GRCh38
rs748328593 Chr.5: 178605278-178605278 on Build GRCh38
rs556387948 Chr.5: 178605582-178605582 on Build GRCh38
rs750805790 Chr.5: 178605341-178605341 on Build GRCh38
rs767149773 Chr.5: 178605298-178605298 on Build GRCh38
rs943328250 Chr.5: 178605231-178605231 on Build GRCh38
rs748996840 Chr.5: 178605419-178605419 on Build GRCh38
rs547918566 Chr.5: 178605362-178605362 on Build GRCh38
rs568060433 Chr.5: 178605530-178605532 on Build GRCh38
rs755792349 Chr.5: 178605313-178605313 on Build GRCh38
rs531419708 Chr.5: 178605346-178605346 on Build GRCh38
rs766468707 Chr.5: 178605287-178605287 on Build GRCh38
rs950864507 Chr.5: 178605483-178605483 on Build GRCh38
rs746419236 Chr.5: 178605480-178605480 on Build GRCh38
rs891502987 Chr.5: 178605319-178605319 on Build GRCh38
rs1010155900 Chr.5: 178605328-178605328 on Build GRCh38
rs548658690 Chr.5: 178605285-178605285 on Build GRCh38
rs769948857 Chr.5: 178605412-178605412 on Build GRCh38
rs984607011 Chr.5: 178605203-178605203 on Build GRCh38
rs747516169 Chr.5: 178605392-178605392 on Build GRCh38
rs1052749472 Chr.5: 178605640-178605640 on Build GRCh38
rs552520405 Chr.5: 178605524-178605524 on Build GRCh38
rs776079002 Chr.5: 178605293-178605294 on Build GRCh38
rs778390790 Chr.5: 178605361-178605361 on Build GRCh38
rs376727729 Chr.5: 178605265-178605265 on Build GRCh38
rs987784185 Chr.5: 178605643-178605643 on Build GRCh38
rs538140369 Chr.5: 178605555-178605555 on Build GRCh38
rs1035921260 Chr.5: 178605488-178605488 on Build GRCh38
rs763319198 Chr.5: 178605306-178605307 on Build GRCh38
rs1002487575 Chr.5: 178605410-178605410 on Build GRCh38
rs748396812 Chr.5: 178605409-178605409 on Build GRCh38
rs887097145 Chr.5: 178605467-178605470 on Build GRCh38
rs533092691 Chr.5: 178605492-178605492 on Build GRCh38
rs758789388 Chr.5: 178605370-178605370 on Build GRCh38
rs574645939 Chr.5: 178605587-178605587 on Build GRCh38
rs186040385 Chr.5: 178605525-178605525 on Build GRCh38

View Full Product Details