Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 122996572-122997075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLVS2
Entrez Gene ID
134829
Gene Name
clavesin 2
Gene Aliases
C6orf212, C6orf213, RLBP1L2, bA160A10.4
UniGene
Hs.486361
Gene Chromosome Location
Chr.6: 122993693-123081282 on Build GRCh38
Transcripts
NM_001010852.3 XM_017010266.1

SNP Detail

SNP ID Chromosome Location
rs1027273322 Chr.6: 122996996-122996996 on Build GRCh38
rs755343392 Chr.6: 122996672-122996672 on Build GRCh38
rs943836669 Chr.6: 122996877-122996877 on Build GRCh38
rs931341438 Chr.6: 122996834-122996834 on Build GRCh38
rs554613762 Chr.6: 122996674-122996674 on Build GRCh38
rs985537390 Chr.6: 122996836-122996836 on Build GRCh38
rs1055451218 Chr.6: 122996923-122996923 on Build GRCh38
rs987229240 Chr.6: 122997047-122997047 on Build GRCh38
rs998950064 Chr.6: 122996935-122996935 on Build GRCh38
rs150863155 Chr.6: 122996885-122996885 on Build GRCh38
rs911341446 Chr.6: 122996874-122996874 on Build GRCh38
rs531381765 Chr.6: 122997035-122997035 on Build GRCh38
rs13199828 Chr.6: 122996620-122996620 on Build GRCh38
rs928932746 Chr.6: 122996596-122996596 on Build GRCh38
rs574469555 Chr.6: 122996686-122996686 on Build GRCh38
rs760516146 Chr.6: 122996598-122996598 on Build GRCh38
rs972726307 Chr.6: 122996817-122996817 on Build GRCh38
rs13200040 Chr.6: 122996669-122996669 on Build GRCh38
rs960609387 Chr.6: 122996783-122996783 on Build GRCh38
rs1034096690 Chr.6: 122996743-122996743 on Build GRCh38
rs1035951149 Chr.6: 122996730-122996730 on Build GRCh38
rs1023169435 Chr.6: 122996797-122996797 on Build GRCh38
rs535238872 Chr.6: 122996659-122996659 on Build GRCh38
rs950561343 Chr.6: 122997025-122997025 on Build GRCh38
rs766387854 Chr.6: 122997018-122997018 on Build GRCh38
rs545647589 Chr.6: 122996879-122996879 on Build GRCh38
rs1026071525 Chr.6: 122997010-122997010 on Build GRCh38
rs1021644077 Chr.6: 122996677-122996677 on Build GRCh38
rs572115109 Chr.6: 122996812-122996812 on Build GRCh38
rs1040972355 Chr.6: 122996703-122996703 on Build GRCh38
rs973534170 Chr.6: 122996994-122996994 on Build GRCh38
rs916407841 Chr.6: 122996726-122996726 on Build GRCh38
rs940017527 Chr.6: 122996658-122996658 on Build GRCh38
rs904177376 Chr.6: 122996897-122996897 on Build GRCh38
rs991749691 Chr.6: 122996794-122996794 on Build GRCh38
rs796357986 Chr.6: 122996917-122996917 on Build GRCh38
rs1023200544 Chr.6: 122996965-122996965 on Build GRCh38
rs961612563 Chr.6: 122996986-122996986 on Build GRCh38
rs772933240 Chr.6: 122996878-122996878 on Build GRCh38
rs566228454 Chr.6: 122996594-122996594 on Build GRCh38
rs572669011 Chr.6: 122996975-122996975 on Build GRCh38
rs890336011 Chr.6: 122996649-122996649 on Build GRCh38
rs13199829 Chr.6: 122996622-122996622 on Build GRCh38
rs1001184942 Chr.6: 122996902-122996902 on Build GRCh38
rs1025200315 Chr.6: 122996810-122996810 on Build GRCh38
rs904230823 Chr.6: 122996732-122996732 on Build GRCh38
rs760688837 Chr.6: 122996892-122996892 on Build GRCh38
rs542582977 Chr.6: 122997049-122997049 on Build GRCh38
rs1043133461 Chr.6: 122996901-122996901 on Build GRCh38
rs889620983 Chr.6: 122996689-122996689 on Build GRCh38
rs985484986 Chr.6: 122997030-122997030 on Build GRCh38
rs973167873 Chr.6: 122996826-122996826 on Build GRCh38
rs1051699067 Chr.6: 122996613-122996613 on Build GRCh38
rs943888992 Chr.6: 122996700-122996700 on Build GRCh38
rs1039317066 Chr.6: 122996662-122996662 on Build GRCh38
rs895805058 Chr.6: 122996951-122996951 on Build GRCh38
rs1009882933 Chr.6: 122996713-122996713 on Build GRCh38
rs952680389 Chr.6: 122996997-122996997 on Build GRCh38
rs999230833 Chr.6: 122996702-122996702 on Build GRCh38
rs537027463 Chr.6: 122996734-122996734 on Build GRCh38
rs987503770 Chr.6: 122996872-122996872 on Build GRCh38
rs928995913 Chr.6: 122996870-122996870 on Build GRCh38
rs992362425 Chr.6: 122996591-122996591 on Build GRCh38
rs1013610844 Chr.6: 122996964-122996964 on Build GRCh38
rs568848042 Chr.6: 122996893-122996893 on Build GRCh38
rs918981273 Chr.6: 122996857-122996857 on Build GRCh38
rs918950248 Chr.6: 122996595-122996595 on Build GRCh38
rs1021715203 Chr.6: 122996725-122996725 on Build GRCh38
rs556716525 Chr.6: 122996760-122996760 on Build GRCh38
rs891732188 Chr.6: 122996708-122996708 on Build GRCh38
rs1036136788 Chr.6: 122996940-122996940 on Build GRCh38
rs911287667 Chr.6: 122997034-122997034 on Build GRCh38

View Full Product Details