Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 123010995-123011544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLVS2
Entrez Gene ID
134829
Gene Name
clavesin 2
Gene Aliases
C6orf212, C6orf213, RLBP1L2, bA160A10.4
UniGene
Hs.486361
Gene Chromosome Location
Chr.6: 122993693-123081282 on Build GRCh38
Transcripts
NM_001010852.3 XM_017010266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4403135 c.556G>A p.G186S Chr.6: 123011151-123011151 on Build GRCh38
54526 c.498C>T p.N166N Chr.6: 123011093-123011093 on Build GRCh38
739367 c.503C>A p.T168N Chr.6: 123011098-123011098 on Build GRCh38
7332682 c.487G>C p.D163H Chr.6: 123011082-123011082 on Build GRCh38
398601 c.415G>T p.A139S Chr.6: 123011010-123011010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778898775 Chr.6: 123011130-123011130 on Build GRCh38
rs771884601 Chr.6: 123011137-123011137 on Build GRCh38
rs1054118307 Chr.6: 123011261-123011262 on Build GRCh38
rs375354563 Chr.6: 123011165-123011165 on Build GRCh38
rs766902762 Chr.6: 123011103-123011103 on Build GRCh38
rs930384462 Chr.6: 123011329-123011329 on Build GRCh38
rs980239020 Chr.6: 123011135-123011135 on Build GRCh38
rs779775195 Chr.6: 123011163-123011163 on Build GRCh38
rs765323473 Chr.6: 123011051-123011051 on Build GRCh38
rs1026282277 Chr.6: 123011429-123011429 on Build GRCh38
rs770861038 Chr.6: 123011202-123011202 on Build GRCh38
rs755782858 Chr.6: 123011119-123011119 on Build GRCh38
rs763406760 Chr.6: 123011102-123011102 on Build GRCh38
rs571522113 Chr.6: 123011457-123011457 on Build GRCh38
rs112239248 Chr.6: 123011212-123011212 on Build GRCh38
rs371688821 Chr.6: 123011109-123011109 on Build GRCh38
rs751923428 Chr.6: 123011105-123011105 on Build GRCh38
rs568011387 Chr.6: 123011339-123011339 on Build GRCh38
rs139717598 Chr.6: 123011128-123011128 on Build GRCh38
rs773317056 Chr.6: 123011081-123011081 on Build GRCh38
rs944635657 Chr.6: 123011360-123011360 on Build GRCh38
rs556847956 Chr.6: 123011387-123011387 on Build GRCh38
rs909925415 Chr.6: 123011075-123011075 on Build GRCh38
rs1012656272 Chr.6: 123011342-123011342 on Build GRCh38
rs886082776 Chr.6: 123011359-123011359 on Build GRCh38
rs770023200 Chr.6: 123011181-123011181 on Build GRCh38
rs1003716015 Chr.6: 123011416-123011416 on Build GRCh38
rs143427910 Chr.6: 123011354-123011354 on Build GRCh38
rs745736656 Chr.6: 123011131-123011131 on Build GRCh38
rs775004850 Chr.6: 123011204-123011204 on Build GRCh38
rs763072200 Chr.6: 123011198-123011198 on Build GRCh38
rs759996346 Chr.6: 123011106-123011106 on Build GRCh38
rs554184284 Chr.6: 123011282-123011282 on Build GRCh38
rs900340618 Chr.6: 123011484-123011484 on Build GRCh38
rs773228597 Chr.6: 123011188-123011188 on Build GRCh38
rs1040257660 Chr.6: 123011461-123011461 on Build GRCh38
rs573359717 Chr.6: 123011347-123011347 on Build GRCh38
rs367929205 Chr.6: 123011201-123011201 on Build GRCh38
rs761982510 Chr.6: 123011031-123011031 on Build GRCh38
rs760084392 Chr.6: 123011207-123011207 on Build GRCh38
rs147496902 Chr.6: 123011057-123011057 on Build GRCh38
rs920381124 Chr.6: 123011156-123011156 on Build GRCh38
rs201320099 Chr.6: 123011123-123011123 on Build GRCh38
rs534311192 Chr.6: 123011211-123011211 on Build GRCh38
rs200795338 Chr.6: 123011108-123011108 on Build GRCh38
rs894169240 Chr.6: 123011268-123011268 on Build GRCh38
rs576932411 Chr.6: 123011393-123011393 on Build GRCh38
rs996044308 Chr.6: 123011492-123011492 on Build GRCh38
rs376708439 Chr.6: 123011189-123011189 on Build GRCh38
rs951660709 Chr.6: 123011470-123011470 on Build GRCh38

View Full Product Details