Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47334041-47334547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK1
Entrez Gene ID
51727
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
UniGene
Hs.731647
Gene Chromosome Location
Chr.1: 47333797-47378839 on Build GRCh38
Transcripts
NM_001136140.1 NM_016308.2 NR_046394.1 NR_046395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4673005 c.108delG p.V37fs*63 Chr.1: 47334053-47334053 on Build GRCh38
4627293 c.164T>C p.I55T Chr.1: 47334109-47334109 on Build GRCh38
7307486 c.105G>A p.P35P Chr.1: 47334050-47334050 on Build GRCh38
6259637 c.118G>T p.V40F Chr.1: 47334063-47334063 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs185656774 Chr.1: 47334524-47334524 on Build GRCh38
rs943376928 Chr.1: 47334307-47334307 on Build GRCh38
rs965515527 Chr.1: 47334204-47334204 on Build GRCh38
rs917760842 Chr.1: 47334292-47334292 on Build GRCh38
rs1000413272 Chr.1: 47334436-47334436 on Build GRCh38
rs752989147 Chr.1: 47334092-47334092 on Build GRCh38
rs959385242 Chr.1: 47334271-47334271 on Build GRCh38
rs889862267 Chr.1: 47334177-47334177 on Build GRCh38
rs1010506829 Chr.1: 47334209-47334209 on Build GRCh38
rs745366019 Chr.1: 47334420-47334420 on Build GRCh38
rs765506640 Chr.1: 47334083-47334083 on Build GRCh38
rs919460384 Chr.1: 47334500-47334500 on Build GRCh38
rs889100665 Chr.1: 47334303-47334303 on Build GRCh38
rs1036931429 Chr.1: 47334301-47334302 on Build GRCh38
rs924937371 Chr.1: 47334270-47334270 on Build GRCh38
rs949321372 Chr.1: 47334137-47334137 on Build GRCh38
rs754520236 Chr.1: 47334133-47334133 on Build GRCh38
rs868747583 Chr.1: 47334078-47334078 on Build GRCh38
rs893932438 Chr.1: 47334323-47334323 on Build GRCh38
rs972245711 Chr.1: 47334202-47334202 on Build GRCh38
rs1006947993 Chr.1: 47334197-47334197 on Build GRCh38
rs901372468 Chr.1: 47334397-47334397 on Build GRCh38
rs779490247 Chr.1: 47334123-47334123 on Build GRCh38
rs1020081430 Chr.1: 47334329-47334329 on Build GRCh38
rs181114398 Chr.1: 47334439-47334439 on Build GRCh38
rs990506231 Chr.1: 47334276-47334276 on Build GRCh38
rs979506013 Chr.1: 47334239-47334239 on Build GRCh38
rs574613731 Chr.1: 47334140-47334140 on Build GRCh38
rs764535721 Chr.1: 47334102-47334102 on Build GRCh38
rs963136947 Chr.1: 47334463-47334463 on Build GRCh38
rs865865460 Chr.1: 47334079-47334079 on Build GRCh38
rs78041605 Chr.1: 47334425-47334425 on Build GRCh38
rs1020537115 Chr.1: 47334211-47334211 on Build GRCh38
rs1054892352 Chr.1: 47334061-47334061 on Build GRCh38
rs1048509911 Chr.1: 47334159-47334159 on Build GRCh38
rs61782734 Chr.1: 47334470-47334470 on Build GRCh38
rs763338621 Chr.1: 47334096-47334096 on Build GRCh38
rs1005631585 Chr.1: 47334480-47334480 on Build GRCh38
rs949207944 Chr.1: 47334295-47334295 on Build GRCh38
rs919423515 Chr.1: 47334208-47334208 on Build GRCh38
rs897181348 Chr.1: 47334155-47334155 on Build GRCh38
rs933554965 Chr.1: 47334251-47334251 on Build GRCh38
rs535190327 Chr.1: 47334158-47334158 on Build GRCh38
rs1010390978 Chr.1: 47334408-47334408 on Build GRCh38
rs1053861696 Chr.1: 47334311-47334311 on Build GRCh38
rs553776365 Chr.1: 47334263-47334263 on Build GRCh38
rs903004093 Chr.1: 47334379-47334379 on Build GRCh38
rs1015969840 Chr.1: 47334452-47334452 on Build GRCh38
rs1007000099 Chr.1: 47334339-47334339 on Build GRCh38
rs1036879478 Chr.1: 47334144-47334144 on Build GRCh38
rs1041122743 Chr.1: 47334353-47334353 on Build GRCh38
rs755588985 Chr.1: 47334116-47334116 on Build GRCh38
rs1012502444 Chr.1: 47334444-47334444 on Build GRCh38
rs955023678 Chr.1: 47334505-47334505 on Build GRCh38
rs966426536 Chr.1: 47334215-47334215 on Build GRCh38
rs537657978 Chr.1: 47334110-47334110 on Build GRCh38
rs889755911 Chr.1: 47334334-47334334 on Build GRCh38
rs72553939 Chr.1: 47334131-47334131 on Build GRCh38
rs4600090 Chr.1: 47334327-47334327 on Build GRCh38

View Full Product Details