Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6240339-6240833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA4
Entrez Gene ID
1262
Gene Name
cyclic nucleotide gated channel alpha 4
Gene Aliases
CNCA2, CNG-4, CNG4, CNG5, CNGB2, OCNC2, OCNCBETA, OCNCb
UniGene
Hs.434618
Gene Chromosome Location
Chr.11: 6234774-6245924 on Build GRCh38
Transcripts
NM_001037329.3 XM_017017217.1 XM_017017218.1 XM_017017219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
179414 c.700G>A p.D234N Chr.11: 6240494-6240494 on Build GRCh38
3451115 c.546G>A p.W182* Chr.11: 6240340-6240340 on Build GRCh38
3451117 c.626C>T p.P209L Chr.11: 6240420-6240420 on Build GRCh38
6256014 c.831G>T p.A277A Chr.11: 6240625-6240625 on Build GRCh38
3451119 c.722A>T p.E241V Chr.11: 6240516-6240516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377739445 Chr.11: 6240382-6240382 on Build GRCh38
rs941716652 Chr.11: 6240520-6240520 on Build GRCh38
rs369162347 Chr.11: 6240561-6240561 on Build GRCh38
rs773283676 Chr.11: 6240413-6240413 on Build GRCh38
rs139774505 Chr.11: 6240705-6240705 on Build GRCh38
rs779510105 Chr.11: 6240608-6240608 on Build GRCh38
rs752921983 Chr.11: 6240571-6240571 on Build GRCh38
rs148534804 Chr.11: 6240723-6240723 on Build GRCh38
rs530313384 Chr.11: 6240805-6240805 on Build GRCh38
rs759681883 Chr.11: 6240557-6240557 on Build GRCh38
rs774116011 Chr.11: 6240556-6240556 on Build GRCh38
rs772047753 Chr.11: 6240745-6240745 on Build GRCh38
rs965745067 Chr.11: 6240656-6240656 on Build GRCh38
rs371882863 Chr.11: 6240440-6240440 on Build GRCh38
rs747646079 Chr.11: 6240649-6240649 on Build GRCh38
rs745664118 Chr.11: 6240739-6240739 on Build GRCh38
rs768057433 Chr.11: 6240586-6240587 on Build GRCh38
rs754533455 Chr.11: 6240644-6240644 on Build GRCh38
rs1007823218 Chr.11: 6240390-6240390 on Build GRCh38
rs774358048 Chr.11: 6240683-6240683 on Build GRCh38
rs938343695 Chr.11: 6240742-6240742 on Build GRCh38
rs780923896 Chr.11: 6240527-6240527 on Build GRCh38
rs145756754 Chr.11: 6240391-6240391 on Build GRCh38
rs749125215 Chr.11: 6240676-6240676 on Build GRCh38
rs373972187 Chr.11: 6240748-6240748 on Build GRCh38
rs773195199 Chr.11: 6240539-6240539 on Build GRCh38
rs371173370 Chr.11: 6240409-6240409 on Build GRCh38
rs116499169 Chr.11: 6240721-6240721 on Build GRCh38
rs1011356156 Chr.11: 6240554-6240554 on Build GRCh38
rs1042878391 Chr.11: 6240569-6240569 on Build GRCh38
rs563032962 Chr.11: 6240543-6240543 on Build GRCh38
rs778641073 Chr.11: 6240734-6240734 on Build GRCh38
rs117364457 Chr.11: 6240471-6240471 on Build GRCh38
rs775132527 Chr.11: 6240387-6240387 on Build GRCh38
rs776855312 Chr.11: 6240749-6240749 on Build GRCh38
rs769615194 Chr.11: 6240650-6240650 on Build GRCh38
rs757534251 Chr.11: 6240607-6240607 on Build GRCh38
rs762221594 Chr.11: 6240369-6240369 on Build GRCh38
rs929035060 Chr.11: 6240810-6240810 on Build GRCh38
rs532111437 Chr.11: 6240698-6240698 on Build GRCh38
rs201514722 Chr.11: 6240754-6240754 on Build GRCh38
rs754251339 Chr.11: 6240594-6240594 on Build GRCh38
rs774714834 Chr.11: 6240474-6240474 on Build GRCh38
rs200149232 Chr.11: 6240504-6240504 on Build GRCh38
rs376486440 Chr.11: 6240432-6240432 on Build GRCh38
rs34731665 Chr.11: 6240753-6240753 on Build GRCh38
rs201720317 Chr.11: 6240392-6240392 on Build GRCh38
rs374795265 Chr.11: 6240535-6240535 on Build GRCh38
rs559326056 Chr.11: 6240400-6240400 on Build GRCh38
rs375229086 Chr.11: 6240452-6240452 on Build GRCh38
rs150625100 Chr.11: 6240684-6240684 on Build GRCh38
rs757078257 Chr.11: 6240733-6240733 on Build GRCh38
rs750848756 Chr.11: 6240643-6240643 on Build GRCh38
rs202030466 Chr.11: 6240368-6240368 on Build GRCh38
rs377158336 Chr.11: 6240695-6240695 on Build GRCh38
rs765465651 Chr.11: 6240701-6240701 on Build GRCh38
rs200921101 Chr.11: 6240414-6240414 on Build GRCh38
rs751074189 Chr.11: 6240791-6240791 on Build GRCh38
rs773216470 Chr.11: 6240544-6240544 on Build GRCh38
rs748833045 Chr.11: 6240731-6240731 on Build GRCh38
rs778274294 Chr.11: 6240376-6240376 on Build GRCh38
rs764288291 Chr.11: 6240581-6240581 on Build GRCh38
rs765078156 Chr.11: 6240412-6240412 on Build GRCh38
rs757820973 Chr.11: 6240495-6240495 on Build GRCh38
rs764343588 Chr.11: 6240697-6240697 on Build GRCh38
rs766965766 Chr.11: 6240720-6240720 on Build GRCh38
rs759205314 Chr.11: 6240746-6240746 on Build GRCh38
rs545343438 Chr.11: 6240672-6240672 on Build GRCh38
rs113053170 Chr.11: 6240681-6240681 on Build GRCh38
rs775604668 Chr.11: 6240694-6240694 on Build GRCh38
rs541605683 Chr.11: 6240526-6240526 on Build GRCh38
rs938566467 Chr.11: 6240480-6240480 on Build GRCh38
rs147119016 Chr.11: 6240437-6240437 on Build GRCh38
rs762816129 Chr.11: 6240541-6240541 on Build GRCh38
rs752866465 Chr.11: 6240453-6240453 on Build GRCh38
rs918940433 Chr.11: 6240783-6240783 on Build GRCh38
rs762262523 Chr.11: 6240699-6240699 on Build GRCh38
rs991448649 Chr.11: 6240484-6240484 on Build GRCh38
rs372711514 Chr.11: 6240579-6240579 on Build GRCh38
rs1052822086 Chr.11: 6240760-6240760 on Build GRCh38
rs746573667 Chr.11: 6240514-6240514 on Build GRCh38
rs796674427 Chr.11: 6240696-6240696 on Build GRCh38
rs771577653 Chr.11: 6240383-6240383 on Build GRCh38
rs1052335892 Chr.11: 6240638-6240638 on Build GRCh38
rs776120647 Chr.11: 6240406-6240406 on Build GRCh38
rs150305809 Chr.11: 6240728-6240728 on Build GRCh38
rs758884971 Chr.11: 6240713-6240713 on Build GRCh38
rs984325476 Chr.11: 6240362-6240362 on Build GRCh38
rs138569309 Chr.11: 6240646-6240646 on Build GRCh38
rs201076058 Chr.11: 6240565-6240565 on Build GRCh38
rs767436851 Chr.11: 6240560-6240560 on Build GRCh38
rs772044710 Chr.11: 6240687-6240687 on Build GRCh38
rs748585063 Chr.11: 6240361-6240361 on Build GRCh38
rs756427921 Chr.11: 6240369-6240369 on Build GRCh38

View Full Product Details