Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41971827-41972319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
UniGene
Hs.273621
Gene Chromosome Location
Chr.17: 41966741-41977736 on Build GRCh38
Transcripts
NM_033133.4 XM_006721701.3 XM_011524340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464857 c.746T>C p.V249A Chr.17: 41971961-41971961 on Build GRCh38
1201629 c.736G>A p.E246K Chr.17: 41972011-41972011 on Build GRCh38
5909573 c.739G>A p.E247K Chr.17: 41972014-41972014 on Build GRCh38
4781627 c.677-5G>A p.? Chr.17: 41971887-41971887 on Build GRCh38
4579699 c.806C>T p.A269V Chr.17: 41972021-41972021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986796351 Chr.17: 41972184-41972184 on Build GRCh38
rs782232248 Chr.17: 41971992-41971992 on Build GRCh38
rs1048695349 Chr.17: 41972006-41972006 on Build GRCh38
rs782737517 Chr.17: 41972028-41972028 on Build GRCh38
rs201235400 Chr.17: 41972069-41972069 on Build GRCh38
rs1012080521 Chr.17: 41972111-41972111 on Build GRCh38
rs782532988 Chr.17: 41971894-41971894 on Build GRCh38
rs377440178 Chr.17: 41972019-41972019 on Build GRCh38
rs781900322 Chr.17: 41971893-41971893 on Build GRCh38
rs766499063 Chr.17: 41972021-41972021 on Build GRCh38
rs782129468 Chr.17: 41971876-41971876 on Build GRCh38
rs370068279 Chr.17: 41971959-41971959 on Build GRCh38
rs782761945 Chr.17: 41971877-41971877 on Build GRCh38
rs142413251 Chr.17: 41971886-41971886 on Build GRCh38
rs782717785 Chr.17: 41971887-41971887 on Build GRCh38
rs782016500 Chr.17: 41971874-41971874 on Build GRCh38
rs781846708 Chr.17: 41971900-41971900 on Build GRCh38
rs957446064 Chr.17: 41972088-41972088 on Build GRCh38
rs782436135 Chr.17: 41971860-41971860 on Build GRCh38
rs146616068 Chr.17: 41972084-41972084 on Build GRCh38
rs782186076 Chr.17: 41971854-41971854 on Build GRCh38
rs782031149 Chr.17: 41971853-41971853 on Build GRCh38
rs199706471 Chr.17: 41971920-41971920 on Build GRCh38
rs782433314 Chr.17: 41971953-41971953 on Build GRCh38
rs781821664 Chr.17: 41972046-41972046 on Build GRCh38
rs964287378 Chr.17: 41972212-41972212 on Build GRCh38
rs369664943 Chr.17: 41971851-41971851 on Build GRCh38
rs930375858 Chr.17: 41972271-41972271 on Build GRCh38
rs781876675 Chr.17: 41972005-41972005 on Build GRCh38
rs782697508 Chr.17: 41972048-41972048 on Build GRCh38
rs907155084 Chr.17: 41972064-41972064 on Build GRCh38
rs1020202920 Chr.17: 41972112-41972112 on Build GRCh38
rs975584590 Chr.17: 41972213-41972213 on Build GRCh38
rs782060416 Chr.17: 41972047-41972047 on Build GRCh38
rs377734705 Chr.17: 41971923-41971923 on Build GRCh38
rs907667134 Chr.17: 41972281-41972281 on Build GRCh38
rs1045677886 Chr.17: 41972056-41972056 on Build GRCh38
rs751611071 Chr.17: 41972180-41972181 on Build GRCh38
rs370341204 Chr.17: 41972022-41972022 on Build GRCh38
rs912800936 Chr.17: 41972205-41972205 on Build GRCh38
rs782161229 Chr.17: 41972004-41972004 on Build GRCh38
rs374449448 Chr.17: 41971964-41971964 on Build GRCh38
rs76779828 Chr.17: 41972141-41972141 on Build GRCh38
rs559707553 Chr.17: 41972272-41972272 on Build GRCh38
rs782764186 Chr.17: 41972071-41972072 on Build GRCh38
rs782346718 Chr.17: 41972000-41972000 on Build GRCh38
rs782357830 Chr.17: 41972081-41972081 on Build GRCh38
rs782496835 Chr.17: 41971906-41971906 on Build GRCh38
rs200309815 Chr.17: 41972002-41972002 on Build GRCh38
rs781979771 Chr.17: 41971881-41971881 on Build GRCh38
rs782538797 Chr.17: 41972073-41972073 on Build GRCh38
rs782787488 Chr.17: 41971896-41971896 on Build GRCh38

View Full Product Details