Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98985668-98986175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260210 c.399G>A p.R133R Chr.9: 98985863-98985863 on Build GRCh38
7192119 c.585T>G p.A195A Chr.9: 98986049-98986049 on Build GRCh38
6632503 c.230G>A p.R77H Chr.9: 98985694-98985694 on Build GRCh38
1103163 c.273C>T p.F91F Chr.9: 98985737-98985737 on Build GRCh38
3652248 c.504C>T p.V168V Chr.9: 98985968-98985968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775618011 Chr.9: 98986016-98986016 on Build GRCh38
rs773097539 Chr.9: 98985779-98985779 on Build GRCh38
rs138366672 Chr.9: 98986015-98986015 on Build GRCh38
rs570016118 Chr.9: 98985957-98985957 on Build GRCh38
rs778572972 Chr.9: 98985824-98985824 on Build GRCh38
rs887513597 Chr.9: 98985972-98985972 on Build GRCh38
rs373086087 Chr.9: 98985885-98985885 on Build GRCh38
rs974480059 Chr.9: 98985741-98985741 on Build GRCh38
rs767075199 Chr.9: 98985930-98985930 on Build GRCh38
rs144123375 Chr.9: 98986101-98986101 on Build GRCh38
rs765891236 Chr.9: 98986060-98986060 on Build GRCh38
rs2075663 Chr.9: 98986074-98986074 on Build GRCh38
rs374984385 Chr.9: 98985801-98985801 on Build GRCh38
rs750622764 Chr.9: 98986002-98986004 on Build GRCh38
rs746005741 Chr.9: 98986014-98986014 on Build GRCh38
rs747201542 Chr.9: 98986017-98986017 on Build GRCh38
rs756805553 Chr.9: 98985908-98985908 on Build GRCh38
rs766766042 Chr.9: 98985781-98985781 on Build GRCh38
rs755701903 Chr.9: 98985751-98985751 on Build GRCh38
rs75461182 Chr.9: 98986007-98986007 on Build GRCh38
rs751914677 Chr.9: 98985746-98985746 on Build GRCh38
rs902739986 Chr.9: 98985714-98985714 on Build GRCh38
rs372275873 Chr.9: 98986135-98986135 on Build GRCh38
rs558739936 Chr.9: 98986031-98986031 on Build GRCh38
rs754797361 Chr.9: 98985924-98985924 on Build GRCh38
rs141708640 Chr.9: 98985737-98985737 on Build GRCh38
rs762408701 Chr.9: 98986054-98986054 on Build GRCh38
rs768805861 Chr.9: 98986120-98986120 on Build GRCh38
rs199564502 Chr.9: 98985772-98985772 on Build GRCh38
rs774534609 Chr.9: 98985937-98985937 on Build GRCh38
rs753271019 Chr.9: 98985795-98985795 on Build GRCh38
rs753527788 Chr.9: 98986100-98986100 on Build GRCh38
rs187697004 Chr.9: 98986030-98986030 on Build GRCh38
rs772923849 Chr.9: 98986089-98986090 on Build GRCh38
rs775541891 Chr.9: 98985771-98985771 on Build GRCh38
rs756050932 Chr.9: 98986009-98986009 on Build GRCh38
rs757910595 Chr.9: 98985964-98985964 on Build GRCh38
rs746093934 Chr.9: 98985941-98985941 on Build GRCh38
rs756750402 Chr.9: 98985813-98985813 on Build GRCh38
rs150514154 Chr.9: 98985960-98985960 on Build GRCh38
rs61999303 Chr.9: 98986062-98986062 on Build GRCh38
rs767388264 Chr.9: 98985692-98985692 on Build GRCh38
rs748577355 Chr.9: 98986147-98986147 on Build GRCh38
rs1016086727 Chr.9: 98985716-98985716 on Build GRCh38
rs748128818 Chr.9: 98986008-98986008 on Build GRCh38
rs765895796 Chr.9: 98985699-98985699 on Build GRCh38
rs777101028 Chr.9: 98985755-98985755 on Build GRCh38
rs200391843 Chr.9: 98985861-98985861 on Build GRCh38
rs368284198 Chr.9: 98985766-98985766 on Build GRCh38
rs141642715 Chr.9: 98985934-98985934 on Build GRCh38
rs968124809 Chr.9: 98986084-98986084 on Build GRCh38
rs749004076 Chr.9: 98985763-98985763 on Build GRCh38
rs771819188 Chr.9: 98985769-98985769 on Build GRCh38
rs756150683 Chr.9: 98986076-98986076 on Build GRCh38
rs374938012 Chr.9: 98985922-98985922 on Build GRCh38
rs762264972 Chr.9: 98985956-98985956 on Build GRCh38
rs754341520 Chr.9: 98985906-98985906 on Build GRCh38
rs745493353 Chr.9: 98985825-98985825 on Build GRCh38
rs201906975 Chr.9: 98985780-98985780 on Build GRCh38
rs759553086 Chr.9: 98985831-98985831 on Build GRCh38
rs774763433 Chr.9: 98985862-98985862 on Build GRCh38
rs533629578 Chr.9: 98986061-98986061 on Build GRCh38
rs752484757 Chr.9: 98986067-98986067 on Build GRCh38
rs781031691 Chr.9: 98985925-98985925 on Build GRCh38
rs750104746 Chr.9: 98985920-98985920 on Build GRCh38
rs755963031 Chr.9: 98986070-98986070 on Build GRCh38
rs751826956 Chr.9: 98985788-98985788 on Build GRCh38
rs2075662 Chr.9: 98985952-98985952 on Build GRCh38
rs367577178 Chr.9: 98986121-98986121 on Build GRCh38
rs781377732 Chr.9: 98986132-98986132 on Build GRCh38
rs34485462 Chr.9: 98986102-98986103 on Build GRCh38
rs781781615 Chr.9: 98985749-98985749 on Build GRCh38
rs751192017 Chr.9: 98985992-98985992 on Build GRCh38
rs773702295 Chr.9: 98986052-98986052 on Build GRCh38
rs147085668 Chr.9: 98985800-98985800 on Build GRCh38
rs770341356 Chr.9: 98986046-98986046 on Build GRCh38
rs768418278 Chr.9: 98985842-98985842 on Build GRCh38
rs1050611150 Chr.9: 98985961-98985961 on Build GRCh38
rs370081942 Chr.9: 98985750-98985750 on Build GRCh38
rs776616947 Chr.9: 98985851-98985851 on Build GRCh38
rs373530299 Chr.9: 98985880-98985880 on Build GRCh38
rs777141387 Chr.9: 98985953-98985953 on Build GRCh38
rs201085523 Chr.9: 98985881-98985881 on Build GRCh38
rs779670663 Chr.9: 98985832-98985832 on Build GRCh38
rs754779649 Chr.9: 98985993-98985993 on Build GRCh38
rs755380230 Chr.9: 98985747-98985747 on Build GRCh38
rs758507654 Chr.9: 98985721-98985721 on Build GRCh38
rs544454009 Chr.9: 98985789-98985789 on Build GRCh38
rs770506843 Chr.9: 98985765-98985765 on Build GRCh38
rs7045978 Chr.9: 98985902-98985902 on Build GRCh38
rs113791685 Chr.9: 98986035-98986035 on Build GRCh38
rs71501885 Chr.9: 98985840-98985840 on Build GRCh38
rs956964338 Chr.9: 98986123-98986123 on Build GRCh38
rs758460554 Chr.9: 98986080-98986080 on Build GRCh38
rs775770612 Chr.9: 98985949-98985949 on Build GRCh38
rs772130405 Chr.9: 98985942-98985942 on Build GRCh38
rs948226567 Chr.9: 98986034-98986034 on Build GRCh38
rs766012986 Chr.9: 98985968-98985968 on Build GRCh38
rs775282439 Chr.9: 98985693-98985693 on Build GRCh38
rs761016441 Chr.9: 98985951-98985951 on Build GRCh38
rs139245903 Chr.9: 98985694-98985694 on Build GRCh38
rs781086307 Chr.9: 98986000-98986000 on Build GRCh38
rs764674198 Chr.9: 98985913-98985913 on Build GRCh38
rs750678351 Chr.9: 98985711-98985711 on Build GRCh38
rs567540031 Chr.9: 98985923-98985923 on Build GRCh38
rs769945542 Chr.9: 98985778-98985778 on Build GRCh38
rs753786778 Chr.9: 98986077-98986077 on Build GRCh38
rs753231557 Chr.9: 98985804-98985804 on Build GRCh38
rs527804413 Chr.9: 98985893-98985893 on Build GRCh38
rs779150801 Chr.9: 98986010-98986010 on Build GRCh38

View Full Product Details