Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18904659-18905163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPE
Entrez Gene ID
11316
Gene Name
coatomer protein complex subunit epsilon
Gene Aliases
epsilon-COP
UniGene
Hs.10326
Gene Chromosome Location
Chr.19: 18899514-18919464 on Build GRCh38
Transcripts
NM_007263.3 NM_199442.1 NM_199444.1 XM_011527655.1 XM_011527656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232083 c.539C>T p.T180I Chr.19: 18904811-18904811 on Build GRCh38
4131483 c.528C>A p.D176E Chr.19: 18904822-18904822 on Build GRCh38
4131484 c.527A>G p.D176G Chr.19: 18904823-18904823 on Build GRCh38
6250890 c.522C>A p.D174E Chr.19: 18904828-18904828 on Build GRCh38
4131482 c.529G>C p.E177Q Chr.19: 18904821-18904821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545324646 Chr.19: 18905046-18905046 on Build GRCh38
rs757577358 Chr.19: 18905027-18905027 on Build GRCh38
rs78590319 Chr.19: 18904682-18904682 on Build GRCh38
rs773951287 Chr.19: 18904887-18904887 on Build GRCh38
rs1039069805 Chr.19: 18904815-18904815 on Build GRCh38
rs964925415 Chr.19: 18904694-18904694 on Build GRCh38
rs983636188 Chr.19: 18904737-18904737 on Build GRCh38
rs61418743 Chr.19: 18904798-18904798 on Build GRCh38
rs59049077 Chr.19: 18905093-18905093 on Build GRCh38
rs976158497 Chr.19: 18904917-18904961 on Build GRCh38
rs748210508 Chr.19: 18904797-18904797 on Build GRCh38
rs778330772 Chr.19: 18904943-18904943 on Build GRCh38
rs1016438586 Chr.19: 18905030-18905030 on Build GRCh38
rs924060417 Chr.19: 18905127-18905127 on Build GRCh38
rs1056307600 Chr.19: 18904924-18904924 on Build GRCh38
rs753371459 Chr.19: 18904816-18904816 on Build GRCh38
rs762785167 Chr.19: 18904867-18904867 on Build GRCh38
rs115146594 Chr.19: 18904736-18904736 on Build GRCh38
rs779330722 Chr.19: 18904740-18904740 on Build GRCh38
rs1023514013 Chr.19: 18904959-18904959 on Build GRCh38
rs745736917 Chr.19: 18904826-18904826 on Build GRCh38
rs113921790 Chr.19: 18904854-18904854 on Build GRCh38
rs962711954 Chr.19: 18904979-18904979 on Build GRCh38
rs1053596846 Chr.19: 18905133-18905133 on Build GRCh38
rs935164007 Chr.19: 18904905-18904905 on Build GRCh38
rs935179863 Chr.19: 18905115-18905115 on Build GRCh38
rs990419787 Chr.19: 18904783-18904783 on Build GRCh38
rs946713207 Chr.19: 18904841-18904841 on Build GRCh38
rs912054930 Chr.19: 18905050-18905050 on Build GRCh38
rs923863310 Chr.19: 18905100-18905100 on Build GRCh38
rs963395101 Chr.19: 18904772-18904772 on Build GRCh38
rs900500953 Chr.19: 18904952-18904952 on Build GRCh38
rs746824366 Chr.19: 18905079-18905081 on Build GRCh38
rs537331771 Chr.19: 18904896-18904900 on Build GRCh38
rs763703433 Chr.19: 18904870-18904870 on Build GRCh38
rs373188845 Chr.19: 18904885-18904885 on Build GRCh38
rs1017887850 Chr.19: 18904691-18904691 on Build GRCh38
rs146007560 Chr.19: 18904822-18904822 on Build GRCh38
rs1039013913 Chr.19: 18904951-18904951 on Build GRCh38
rs1053636250 Chr.19: 18904937-18904937 on Build GRCh38
rs781281885 Chr.19: 18905035-18905035 on Build GRCh38
rs1030956570 Chr.19: 18904726-18904726 on Build GRCh38
rs997801217 Chr.19: 18904990-18904990 on Build GRCh38
rs775143304 Chr.19: 18904859-18904859 on Build GRCh38
rs562760416 Chr.19: 18905017-18905017 on Build GRCh38
rs551330785 Chr.19: 18904939-18904939 on Build GRCh38
rs200769434 Chr.19: 18904860-18904860 on Build GRCh38
rs976251229 Chr.19: 18904710-18904710 on Build GRCh38
rs553840810 Chr.19: 18904890-18904890 on Build GRCh38
rs908945623 Chr.19: 18904747-18904747 on Build GRCh38
rs916288964 Chr.19: 18905051-18905051 on Build GRCh38
rs370664978 Chr.19: 18904838-18904838 on Build GRCh38
rs188669506 Chr.19: 18904938-18904938 on Build GRCh38
rs74936637 Chr.19: 18905026-18905026 on Build GRCh38
rs970360816 Chr.19: 18905065-18905065 on Build GRCh38
rs376379507 Chr.19: 18904745-18904745 on Build GRCh38
rs951118189 Chr.19: 18905031-18905031 on Build GRCh38
rs142579871 Chr.19: 18904780-18904780 on Build GRCh38

View Full Product Details