Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 965155-965662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX19
Entrez Gene ID
90639
Gene Name
COX19, cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.121593
Gene Chromosome Location
Chr.7: 964850-975599 on Build GRCh38
Transcripts
NM_001031617.2

SNP Detail

SNP ID Chromosome Location
rs947155993 Chr.7: 965370-965370 on Build GRCh38
rs966945155 Chr.7: 965527-965527 on Build GRCh38
rs533605409 Chr.7: 965217-965217 on Build GRCh38
rs111645884 Chr.7: 965506-965506 on Build GRCh38
rs575607637 Chr.7: 965537-965537 on Build GRCh38
rs974946093 Chr.7: 965346-965346 on Build GRCh38
rs867091594 Chr.7: 965581-965581 on Build GRCh38
rs996411869 Chr.7: 965628-965628 on Build GRCh38
rs1013132956 Chr.7: 965459-965459 on Build GRCh38
rs199837078 Chr.7: 965238-965238 on Build GRCh38
rs899027525 Chr.7: 965570-965570 on Build GRCh38
rs963465907 Chr.7: 965233-965233 on Build GRCh38
rs1026306197 Chr.7: 965480-965480 on Build GRCh38
rs539690949 Chr.7: 965372-965372 on Build GRCh38
rs897383500 Chr.7: 965247-965247 on Build GRCh38
rs190027195 Chr.7: 965239-965239 on Build GRCh38
rs117333129 Chr.7: 965457-965457 on Build GRCh38
rs975914680 Chr.7: 965345-965345 on Build GRCh38
rs924990896 Chr.7: 965386-965387 on Build GRCh38
rs1048690038 Chr.7: 965630-965630 on Build GRCh38
rs1046584182 Chr.7: 965454-965454 on Build GRCh38
rs1056809163 Chr.7: 965536-965536 on Build GRCh38
rs895660452 Chr.7: 965193-965193 on Build GRCh38
rs756197789 Chr.7: 965472-965472 on Build GRCh38
rs994468416 Chr.7: 965467-965467 on Build GRCh38
rs530682947 Chr.7: 965299-965299 on Build GRCh38
rs915421854 Chr.7: 965504-965504 on Build GRCh38
rs934357798 Chr.7: 965530-965530 on Build GRCh38
rs879844859 Chr.7: 965493-965493 on Build GRCh38
rs370027029 Chr.7: 965600-965600 on Build GRCh38
rs186130186 Chr.7: 965331-965331 on Build GRCh38
rs1018963215 Chr.7: 965620-965620 on Build GRCh38
rs138590416 Chr.7: 965290-965290 on Build GRCh38
rs141587624 Chr.7: 965347-965347 on Build GRCh38
rs924275637 Chr.7: 965508-965508 on Build GRCh38
rs774540092 Chr.7: 965179-965179 on Build GRCh38
rs772511141 Chr.7: 965237-965237 on Build GRCh38
rs934575293 Chr.7: 965443-965443 on Build GRCh38
rs964991502 Chr.7: 965498-965498 on Build GRCh38
rs577774271 Chr.7: 965199-965199 on Build GRCh38
rs994936400 Chr.7: 965479-965479 on Build GRCh38
rs181240328 Chr.7: 965283-965283 on Build GRCh38
rs1006982079 Chr.7: 965330-965330 on Build GRCh38
rs925170604 Chr.7: 965550-965550 on Build GRCh38
rs778023451 Chr.7: 965507-965507 on Build GRCh38
rs952687043 Chr.7: 965328-965328 on Build GRCh38
rs764595078 Chr.7: 965375-965375 on Build GRCh38
rs918764691 Chr.7: 965622-965622 on Build GRCh38
rs936304837 Chr.7: 965423-965423 on Build GRCh38
rs1044223727 Chr.7: 965564-965564 on Build GRCh38
rs1019579978 Chr.7: 965497-965497 on Build GRCh38
rs200476102 Chr.7: 965246-965248 on Build GRCh38
rs899226555 Chr.7: 965478-965478 on Build GRCh38
rs1054809912 Chr.7: 965426-965426 on Build GRCh38
rs763399750 Chr.7: 965341-965341 on Build GRCh38
rs1009273538 Chr.7: 965611-965611 on Build GRCh38
rs10282027 Chr.7: 965382-965382 on Build GRCh38
rs367607557 Chr.7: 965259-965259 on Build GRCh38
rs957713998 Chr.7: 965553-965553 on Build GRCh38
rs10235055 Chr.7: 965635-965635 on Build GRCh38
rs544047100 Chr.7: 965445-965445 on Build GRCh38
rs576714116 Chr.7: 965403-965403 on Build GRCh38
rs1025661851 Chr.7: 965246-965246 on Build GRCh38
rs535954171 Chr.7: 965481-965481 on Build GRCh38
rs2178110 Chr.7: 965486-965486 on Build GRCh38
rs60977172 Chr.7: 965192-965192 on Build GRCh38
rs948684262 Chr.7: 965563-965563 on Build GRCh38
rs779266392 Chr.7: 965584-965584 on Build GRCh38
rs139271203 Chr.7: 965586-965586 on Build GRCh38
rs916303523 Chr.7: 965431-965431 on Build GRCh38
rs764125344 Chr.7: 965456-965456 on Build GRCh38
rs772631544 Chr.7: 965627-965627 on Build GRCh38
rs528183911 Chr.7: 965273-965273 on Build GRCh38
rs1057429776 Chr.7: 965453-965453 on Build GRCh38
rs559064179 Chr.7: 965261-965261 on Build GRCh38
rs975291271 Chr.7: 965503-965503 on Build GRCh38

View Full Product Details