Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82571564-82572060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB1
Entrez Gene ID
64506
Gene Name
cytoplasmic polyadenylation element binding protein 1
Gene Aliases
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1
UniGene
Hs.459132
Gene Chromosome Location
Chr.15: 82543201-82648016 on Build GRCh38
Transcripts
NM_001079533.1 NM_001079534.1 NM_001079535.1 NM_001288819.1 NM_001288820.1 NM_030594.4

SNP Detail

SNP ID Chromosome Location
rs898681323 Chr.15: 82571956-82571956 on Build GRCh38
rs533104462 Chr.15: 82571684-82571684 on Build GRCh38
rs568119088 Chr.15: 82571669-82571669 on Build GRCh38
rs950410188 Chr.15: 82571987-82571987 on Build GRCh38
rs936119889 Chr.15: 82571842-82571842 on Build GRCh38
rs146140459 Chr.15: 82571599-82571599 on Build GRCh38
rs114511543 Chr.15: 82572039-82572039 on Build GRCh38
rs118064941 Chr.15: 82571955-82571955 on Build GRCh38
rs548978034 Chr.15: 82571850-82571850 on Build GRCh38
rs983523725 Chr.15: 82571991-82571991 on Build GRCh38
rs1039552330 Chr.15: 82571696-82571696 on Build GRCh38
rs755577992 Chr.15: 82571986-82571986 on Build GRCh38
rs994580792 Chr.15: 82571979-82571979 on Build GRCh38
rs960795418 Chr.15: 82571959-82571959 on Build GRCh38
rs1045962041 Chr.15: 82571935-82571935 on Build GRCh38
rs1002310964 Chr.15: 82571953-82571953 on Build GRCh38
rs1050913230 Chr.15: 82571906-82571906 on Build GRCh38
rs911850598 Chr.15: 82571729-82571729 on Build GRCh38
rs190745150 Chr.15: 82571941-82571941 on Build GRCh38
rs1001699845 Chr.15: 82571609-82571609 on Build GRCh38
rs933838523 Chr.15: 82571895-82571895 on Build GRCh38
rs757138579 Chr.15: 82571843-82571843 on Build GRCh38
rs939616070 Chr.15: 82571635-82571635 on Build GRCh38
rs755739425 Chr.15: 82571943-82571943 on Build GRCh38
rs965964861 Chr.15: 82572012-82572012 on Build GRCh38
rs921745637 Chr.15: 82572023-82572023 on Build GRCh38
rs745599771 Chr.15: 82571890-82571890 on Build GRCh38
rs753875446 Chr.15: 82571590-82571590 on Build GRCh38
rs914154016 Chr.15: 82571840-82571840 on Build GRCh38
rs972464508 Chr.15: 82571816-82571824 on Build GRCh38
rs962826596 Chr.15: 82571778-82571778 on Build GRCh38
rs992746466 Chr.15: 82571981-82571981 on Build GRCh38
rs575086178 Chr.15: 82571596-82571596 on Build GRCh38
rs1029976649 Chr.15: 82571740-82571740 on Build GRCh38
rs974550611 Chr.15: 82572020-82572020 on Build GRCh38
rs925620763 Chr.15: 82571882-82571882 on Build GRCh38
rs550993262 Chr.15: 82571886-82571886 on Build GRCh38
rs1016905959 Chr.15: 82571777-82571777 on Build GRCh38
rs1027850536 Chr.15: 82571986-82571986 on Build GRCh38
rs1033352367 Chr.15: 82571636-82571636 on Build GRCh38
rs780998354 Chr.15: 82571848-82571848 on Build GRCh38
rs944483022 Chr.15: 82571733-82571733 on Build GRCh38
rs992314547 Chr.15: 82571829-82571829 on Build GRCh38
rs916944397 Chr.15: 82571640-82571640 on Build GRCh38
rs961140058 Chr.15: 82571822-82571822 on Build GRCh38
rs921981427 Chr.15: 82571819-82571819 on Build GRCh38
rs899700698 Chr.15: 82571893-82571893 on Build GRCh38
rs368165903 Chr.15: 82571656-82571656 on Build GRCh38
rs951299745 Chr.15: 82571750-82571750 on Build GRCh38
rs117188117 Chr.15: 82571691-82571691 on Build GRCh38
rs571781467 Chr.15: 82571604-82571606 on Build GRCh38
rs186230309 Chr.15: 82571702-82571702 on Build GRCh38
rs920253878 Chr.15: 82571678-82571678 on Build GRCh38
rs182146226 Chr.15: 82572035-82572035 on Build GRCh38
rs935786740 Chr.15: 82571884-82571884 on Build GRCh38
rs1018839242 Chr.15: 82572002-82572002 on Build GRCh38
rs1001626315 Chr.15: 82571954-82571954 on Build GRCh38
rs779739697 Chr.15: 82571951-82571951 on Build GRCh38
rs537004154 Chr.15: 82571878-82571878 on Build GRCh38
rs1010687760 Chr.15: 82571897-82571897 on Build GRCh38
rs887027303 Chr.15: 82571932-82571932 on Build GRCh38
rs959124391 Chr.15: 82571668-82571668 on Build GRCh38
rs1038892744 Chr.15: 82571751-82571751 on Build GRCh38
rs569135992 Chr.15: 82571859-82571859 on Build GRCh38
rs1004129991 Chr.15: 82571934-82571934 on Build GRCh38
rs768178780 Chr.15: 82572025-82572025 on Build GRCh38

View Full Product Details