Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11842130-11842634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSB
Entrez Gene ID
1508
Gene Name
cathepsin B
Gene Aliases
APPS, CPSB
UniGene
Hs.520898
Gene Chromosome Location
Chr.8: 11842524-11868150 on Build GRCh38
Transcripts
NM_001317237.1 NM_001908.4 NM_147780.3 NM_147781.3 NM_147782.3 NM_147783.3

SNP Detail

SNP ID Chromosome Location
rs918185074 Chr.8: 11842502-11842502 on Build GRCh38
rs1692822 Chr.8: 11842198-11842198 on Build GRCh38
rs570211266 Chr.8: 11842283-11842283 on Build GRCh38
rs931374959 Chr.8: 11842408-11842408 on Build GRCh38
rs935781195 Chr.8: 11842340-11842340 on Build GRCh38
rs552615383 Chr.8: 11842329-11842329 on Build GRCh38
rs1023596409 Chr.8: 11842495-11842495 on Build GRCh38
rs1013590945 Chr.8: 11842485-11842485 on Build GRCh38
rs912696566 Chr.8: 11842537-11842537 on Build GRCh38
rs964604955 Chr.8: 11842254-11842254 on Build GRCh38
rs1021442273 Chr.8: 11842470-11842470 on Build GRCh38
rs771111656 Chr.8: 11842433-11842434 on Build GRCh38
rs920698081 Chr.8: 11842174-11842174 on Build GRCh38
rs768191480 Chr.8: 11842427-11842427 on Build GRCh38
rs1040070923 Chr.8: 11842460-11842460 on Build GRCh38
rs539760046 Chr.8: 11842444-11842444 on Build GRCh38
rs78763986 Chr.8: 11842158-11842158 on Build GRCh38
rs1015586596 Chr.8: 11842163-11842163 on Build GRCh38
rs1040060274 Chr.8: 11842377-11842377 on Build GRCh38
rs762599789 Chr.8: 11842393-11842393 on Build GRCh38
rs938235768 Chr.8: 11842237-11842237 on Build GRCh38
rs112086464 Chr.8: 11842480-11842480 on Build GRCh38
rs6731 Chr.8: 11842592-11842592 on Build GRCh38
rs940192607 Chr.8: 11842398-11842398 on Build GRCh38
rs925485743 Chr.8: 11842434-11842434 on Build GRCh38
rs1008826034 Chr.8: 11842239-11842239 on Build GRCh38
rs895334078 Chr.8: 11842220-11842220 on Build GRCh38
rs969330705 Chr.8: 11842501-11842501 on Build GRCh38
rs4841601 Chr.8: 11842221-11842221 on Build GRCh38
rs891678974 Chr.8: 11842457-11842457 on Build GRCh38
rs1028713441 Chr.8: 11842303-11842303 on Build GRCh38
rs926916347 Chr.8: 11842231-11842231 on Build GRCh38
rs968527681 Chr.8: 11842474-11842474 on Build GRCh38
rs956972771 Chr.8: 11842516-11842516 on Build GRCh38
rs907889576 Chr.8: 11842299-11842299 on Build GRCh38
rs147542585 Chr.8: 11842413-11842413 on Build GRCh38
rs753560736 Chr.8: 11842182-11842182 on Build GRCh38
rs968852610 Chr.8: 11842368-11842368 on Build GRCh38
rs898543737 Chr.8: 11842300-11842300 on Build GRCh38
rs1056664743 Chr.8: 11842246-11842246 on Build GRCh38
rs931665288 Chr.8: 11842575-11842575 on Build GRCh38
rs570718685 Chr.8: 11842331-11842331 on Build GRCh38
rs1010146612 Chr.8: 11842609-11842609 on Build GRCh38
rs563434243 Chr.8: 11842545-11842545 on Build GRCh38
rs559417110 Chr.8: 11842484-11842484 on Build GRCh38
rs553106215 Chr.8: 11842466-11842466 on Build GRCh38
rs947081274 Chr.8: 11842228-11842228 on Build GRCh38
rs757890226 Chr.8: 11842419-11842419 on Build GRCh38
rs529882458 Chr.8: 11842271-11842271 on Build GRCh38
rs565345192 Chr.8: 11842219-11842219 on Build GRCh38
rs781702380 Chr.8: 11842503-11842503 on Build GRCh38
rs552678623 Chr.8: 11842593-11842593 on Build GRCh38
rs532275865 Chr.8: 11842203-11842203 on Build GRCh38
rs188206992 Chr.8: 11842232-11842232 on Build GRCh38
rs180821099 Chr.8: 11842297-11842297 on Build GRCh38
rs558150516 Chr.8: 11842471-11842471 on Build GRCh38
rs986664217 Chr.8: 11842199-11842199 on Build GRCh38
rs187321891 Chr.8: 11842376-11842376 on Build GRCh38
rs867399428 Chr.8: 11842355-11842355 on Build GRCh38
rs926941168 Chr.8: 11842391-11842391 on Build GRCh38
rs867073077 Chr.8: 11842421-11842421 on Build GRCh38
rs548343458 Chr.8: 11842281-11842281 on Build GRCh38
rs955850591 Chr.8: 11842346-11842346 on Build GRCh38
rs912386471 Chr.8: 11842205-11842205 on Build GRCh38
rs529998933 Chr.8: 11842493-11842493 on Build GRCh38
rs889843796 Chr.8: 11842567-11842567 on Build GRCh38
rs112267640 Chr.8: 11842161-11842165 on Build GRCh38
rs931905647 Chr.8: 11842469-11842469 on Build GRCh38
rs541699455 Chr.8: 11842499-11842499 on Build GRCh38
rs889781419 Chr.8: 11842423-11842423 on Build GRCh38
rs920069259 Chr.8: 11842544-11842544 on Build GRCh38
rs996049263 Chr.8: 11842273-11842273 on Build GRCh38
rs59291913 Chr.8: 11842352-11842352 on Build GRCh38
rs141503656 Chr.8: 11842358-11842358 on Build GRCh38
rs960806898 Chr.8: 11842412-11842412 on Build GRCh38
rs761374736 Chr.8: 11842473-11842473 on Build GRCh38
rs944023124 Chr.8: 11842589-11842589 on Build GRCh38
rs921611907 Chr.8: 11842560-11842560 on Build GRCh38
rs992160875 Chr.8: 11842425-11842425 on Build GRCh38
rs900453635 Chr.8: 11842387-11842387 on Build GRCh38
rs1027480231 Chr.8: 11842280-11842280 on Build GRCh38
rs891529200 Chr.8: 11842370-11842370 on Build GRCh38
rs369179289 Chr.8: 11842161-11842161 on Build GRCh38
rs979629745 Chr.8: 11842433-11842433 on Build GRCh38
rs969622376 Chr.8: 11842431-11842431 on Build GRCh38
rs73543173 Chr.8: 11842438-11842438 on Build GRCh38
rs891431299 Chr.8: 11842238-11842238 on Build GRCh38
rs1004287508 Chr.8: 11842278-11842278 on Build GRCh38
rs183869289 Chr.8: 11842177-11842177 on Build GRCh38
rs931452980 Chr.8: 11842551-11842551 on Build GRCh38
rs1044414353 Chr.8: 11842230-11842230 on Build GRCh38
rs191686846 Chr.8: 11842153-11842153 on Build GRCh38
rs116097524 Chr.8: 11842401-11842401 on Build GRCh38
rs896641522 Chr.8: 11842262-11842262 on Build GRCh38
rs1018899623 Chr.8: 11842250-11842250 on Build GRCh38
rs190621291 Chr.8: 11842392-11842392 on Build GRCh38
rs913012310 Chr.8: 11842453-11842453 on Build GRCh38
rs974560453 Chr.8: 11842168-11842168 on Build GRCh38
rs752213295 Chr.8: 11842402-11842402 on Build GRCh38
rs1014759774 Chr.8: 11842481-11842481 on Build GRCh38
rs995538479 Chr.8: 11842301-11842301 on Build GRCh38
rs1009000376 Chr.8: 11842374-11842374 on Build GRCh38
rs1010092502 Chr.8: 11842463-11842463 on Build GRCh38
rs372563624 Chr.8: 11842227-11842227 on Build GRCh38
rs116789205 Chr.8: 11842178-11842178 on Build GRCh38
rs535719837 Chr.8: 11842397-11842397 on Build GRCh38
rs972903884 Chr.8: 11842520-11842520 on Build GRCh38
rs980242585 Chr.8: 11842476-11842476 on Build GRCh38
rs1008542232 Chr.8: 11842332-11842332 on Build GRCh38
rs374729002 Chr.8: 11842342-11842342 on Build GRCh38
rs891732560 Chr.8: 11842605-11842605 on Build GRCh38
rs1037083751 Chr.8: 11842289-11842289 on Build GRCh38
rs1033029135 Chr.8: 11842512-11842512 on Build GRCh38
rs900128162 Chr.8: 11842465-11842465 on Build GRCh38
rs983904219 Chr.8: 11842291-11842291 on Build GRCh38
rs1036615665 Chr.8: 11842603-11842603 on Build GRCh38
rs28620463 Chr.8: 11842334-11842334 on Build GRCh38
rs1692821 Chr.8: 11842479-11842479 on Build GRCh38
rs1050265329 Chr.8: 11842564-11842564 on Build GRCh38
rs955796960 Chr.8: 11842216-11842216 on Build GRCh38

View Full Product Details