Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58995418-58995918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSZ
Entrez Gene ID
1522
Gene Name
cathepsin Z
Gene Aliases
CTSX
UniGene
Hs.252549
Gene Chromosome Location
Chr.20: 58995187-59007254 on Build GRCh38
Transcripts
NM_001336.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4911176 c.863A>G p.Y288C Chr.20: 58995698-58995698 on Build GRCh38
28388 c.852G>A p.K284K Chr.20: 58995709-58995709 on Build GRCh38
5190917 c.856G>A p.A286T Chr.20: 58995705-58995705 on Build GRCh38
6637851 c.804C>T p.G268G Chr.20: 58995757-58995757 on Build GRCh38
459715 c.819G>A p.L273L Chr.20: 58995742-58995742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377211201 Chr.20: 58995728-58995728 on Build GRCh38
rs1013036420 Chr.20: 58995841-58995841 on Build GRCh38
rs747831221 Chr.20: 58995530-58995530 on Build GRCh38
rs531118235 Chr.20: 58995836-58995836 on Build GRCh38
rs373057693 Chr.20: 58995776-58995776 on Build GRCh38
rs757409572 Chr.20: 58995692-58995692 on Build GRCh38
rs778830780 Chr.20: 58995696-58995696 on Build GRCh38
rs901765493 Chr.20: 58995517-58995517 on Build GRCh38
rs1035845013 Chr.20: 58995445-58995445 on Build GRCh38
rs768616983 Chr.20: 58995634-58995634 on Build GRCh38
rs753074949 Chr.20: 58995684-58995684 on Build GRCh38
rs1002513162 Chr.20: 58995833-58995833 on Build GRCh38
rs889783101 Chr.20: 58995550-58995550 on Build GRCh38
rs771559473 Chr.20: 58995798-58995798 on Build GRCh38
rs772282883 Chr.20: 58995707-58995707 on Build GRCh38
rs749203548 Chr.20: 58995504-58995504 on Build GRCh38
rs552511082 Chr.20: 58995555-58995555 on Build GRCh38
rs753994699 Chr.20: 58995689-58995689 on Build GRCh38
rs771918237 Chr.20: 58995549-58995549 on Build GRCh38
rs1043862133 Chr.20: 58995496-58995496 on Build GRCh38
rs1040210590 Chr.20: 58995515-58995515 on Build GRCh38
rs769169843 Chr.20: 58995803-58995803 on Build GRCh38
rs765511554 Chr.20: 58995750-58995750 on Build GRCh38
rs946363487 Chr.20: 58995661-58995661 on Build GRCh38
rs994175727 Chr.20: 58995755-58995755 on Build GRCh38
rs552691341 Chr.20: 58995873-58995873 on Build GRCh38
rs764347121 Chr.20: 58995743-58995743 on Build GRCh38
rs780100346 Chr.20: 58995613-58995613 on Build GRCh38
rs781168607 Chr.20: 58995642-58995642 on Build GRCh38
rs747899202 Chr.20: 58995643-58995643 on Build GRCh38
rs909139735 Chr.20: 58995897-58995897 on Build GRCh38
rs577223194 Chr.20: 58995777-58995777 on Build GRCh38
rs537622348 Chr.20: 58995527-58995527 on Build GRCh38
rs764445033 Chr.20: 58995687-58995687 on Build GRCh38
rs113505975 Chr.20: 58995778-58995778 on Build GRCh38
rs6064734 Chr.20: 58995718-58995718 on Build GRCh38
rs1039430565 Chr.20: 58995535-58995535 on Build GRCh38
rs370129546 Chr.20: 58995736-58995736 on Build GRCh38
rs200216191 Chr.20: 58995706-58995706 on Build GRCh38
rs113005730 Chr.20: 58995761-58995761 on Build GRCh38
rs772671004 Chr.20: 58995654-58995654 on Build GRCh38
rs931789474 Chr.20: 58995840-58995840 on Build GRCh38
rs778878200 Chr.20: 58995619-58995621 on Build GRCh38
rs914660362 Chr.20: 58995470-58995470 on Build GRCh38
rs770331757 Chr.20: 58995676-58995676 on Build GRCh38
rs886746195 Chr.20: 58995832-58995832 on Build GRCh38
rs374906677 Chr.20: 58995439-58995440 on Build GRCh38
rs777536692 Chr.20: 58995721-58995721 on Build GRCh38
rs778615569 Chr.20: 58995805-58995805 on Build GRCh38
rs776954962 Chr.20: 58995744-58995744 on Build GRCh38
rs958769851 Chr.20: 58995446-58995446 on Build GRCh38
rs530142909 Chr.20: 58995788-58995788 on Build GRCh38
rs575953808 Chr.20: 58995722-58995722 on Build GRCh38
rs916211878 Chr.20: 58995444-58995444 on Build GRCh38
rs774886423 Chr.20: 58995679-58995679 on Build GRCh38
rs200323067 Chr.20: 58995780-58995780 on Build GRCh38
rs761977049 Chr.20: 58995747-58995747 on Build GRCh38
rs931080980 Chr.20: 58995796-58995796 on Build GRCh38
rs372596273 Chr.20: 58995631-58995631 on Build GRCh38
rs764585799 Chr.20: 58995858-58995858 on Build GRCh38
rs538357999 Chr.20: 58995602-58995602 on Build GRCh38
rs865916774 Chr.20: 58995828-58995828 on Build GRCh38
rs773414005 Chr.20: 58995750-58995751 on Build GRCh38
rs946519766 Chr.20: 58995488-58995488 on Build GRCh38
rs748164483 Chr.20: 58995658-58995658 on Build GRCh38
rs750540427 Chr.20: 58995756-58995756 on Build GRCh38
rs368797360 Chr.20: 58995681-58995681 on Build GRCh38
rs796807785 Chr.20: 58995469-58995469 on Build GRCh38
rs776229887 Chr.20: 58995720-58995720 on Build GRCh38
rs141386152 Chr.20: 58995700-58995700 on Build GRCh38
rs894739014 Chr.20: 58995838-58995838 on Build GRCh38
rs761161044 Chr.20: 58995739-58995739 on Build GRCh38
rs376946513 Chr.20: 58995648-58995648 on Build GRCh38
rs773881826 Chr.20: 58995724-58995724 on Build GRCh38
rs200993587 Chr.20: 58995603-58995603 on Build GRCh38
rs947370139 Chr.20: 58995492-58995492 on Build GRCh38
rs368439024 Chr.20: 58995781-58995781 on Build GRCh38
rs1012474771 Chr.20: 58995769-58995769 on Build GRCh38
rs759384258 Chr.20: 58995803-58995809 on Build GRCh38
rs772730109 Chr.20: 58995551-58995551 on Build GRCh38
rs762587538 Chr.20: 58995655-58995655 on Build GRCh38
rs34069356 Chr.20: 58995705-58995705 on Build GRCh38
rs563935485 Chr.20: 58995830-58995830 on Build GRCh38
rs375173339 Chr.20: 58995806-58995806 on Build GRCh38
rs376895957 Chr.20: 58995779-58995779 on Build GRCh38
rs760962516 Chr.20: 58995685-58995685 on Build GRCh38
rs3787492 Chr.20: 58995799-58995799 on Build GRCh38
rs931925541 Chr.20: 58995717-58995717 on Build GRCh38
rs202209335 Chr.20: 58995735-58995735 on Build GRCh38
rs961455564 Chr.20: 58995557-58995557 on Build GRCh38
rs13720 Chr.20: 58995513-58995513 on Build GRCh38
rs150409478 Chr.20: 58995838-58995838 on Build GRCh38
rs972893900 Chr.20: 58995559-58995559 on Build GRCh38
rs758480638 Chr.20: 58995757-58995757 on Build GRCh38
rs199978196 Chr.20: 58995754-58995754 on Build GRCh38
rs1035109276 Chr.20: 58995567-58995567 on Build GRCh38
rs923598657 Chr.20: 58995784-58995784 on Build GRCh38
rs750495354 Chr.20: 58995698-58995698 on Build GRCh38
rs991505589 Chr.20: 58995464-58995464 on Build GRCh38
rs571003122 Chr.20: 58995898-58995898 on Build GRCh38
rs571279081 Chr.20: 58995578-58995578 on Build GRCh38
rs757281239 Chr.20: 58995608-58995608 on Build GRCh38
rs1050223948 Chr.20: 58995881-58995881 on Build GRCh38

View Full Product Details