Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108071848-108072396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL5
Entrez Gene ID
8065
Gene Name
cullin 5
Gene Aliases
VACM-1, VACM1
UniGene
Hs.440320
Gene Chromosome Location
Chr.11: 108008682-108107766 on Build GRCh38
Transcripts
NM_003478.3 XM_005271682.2 XM_011543013.1 XM_017018363.1 XM_017018364.1 XM_017018365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162924 c.917G>T p.G306V Chr.11: 108072374-108072374 on Build GRCh38
1704531 c.910C>T p.P304S Chr.11: 108072367-108072367 on Build GRCh38
4826658 c.878T>G p.L293* Chr.11: 108072335-108072335 on Build GRCh38
1627749 c.900G>A p.M300I Chr.11: 108072357-108072357 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761615310 Chr.11: 108072200-108072200 on Build GRCh38
rs951080525 Chr.11: 108071947-108071947 on Build GRCh38
rs73557142 Chr.11: 108072128-108072128 on Build GRCh38
rs867630223 Chr.11: 108072001-108072001 on Build GRCh38
rs745406143 Chr.11: 108072362-108072362 on Build GRCh38
rs547113683 Chr.11: 108072134-108072134 on Build GRCh38
rs139897854 Chr.11: 108072100-108072100 on Build GRCh38
rs958561690 Chr.11: 108072133-108072133 on Build GRCh38
rs552092979 Chr.11: 108071934-108071934 on Build GRCh38
rs925670162 Chr.11: 108071928-108071928 on Build GRCh38
rs925665643 Chr.11: 108072189-108072189 on Build GRCh38
rs765195678 Chr.11: 108072309-108072309 on Build GRCh38
rs577950268 Chr.11: 108072278-108072278 on Build GRCh38
rs916069574 Chr.11: 108072064-108072064 on Build GRCh38
rs921021531 Chr.11: 108072201-108072201 on Build GRCh38
rs754950193 Chr.11: 108071955-108071955 on Build GRCh38
rs567127926 Chr.11: 108071957-108071957 on Build GRCh38
rs1004599418 Chr.11: 108072019-108072019 on Build GRCh38
rs991224387 Chr.11: 108072039-108072039 on Build GRCh38
rs1051420681 Chr.11: 108072217-108072217 on Build GRCh38
rs998032463 Chr.11: 108072215-108072215 on Build GRCh38
rs750232747 Chr.11: 108072218-108072218 on Build GRCh38
rs982422199 Chr.11: 108071970-108071970 on Build GRCh38
rs779064591 Chr.11: 108072114-108072114 on Build GRCh38
rs768224932 Chr.11: 108071966-108071966 on Build GRCh38
rs767366473 Chr.11: 108072203-108072203 on Build GRCh38
rs779973742 Chr.11: 108072370-108072370 on Build GRCh38
rs1054456441 Chr.11: 108072143-108072143 on Build GRCh38
rs748236348 Chr.11: 108072283-108072283 on Build GRCh38
rs537934869 Chr.11: 108072206-108072206 on Build GRCh38
rs370972008 Chr.11: 108072311-108072311 on Build GRCh38
rs763502011 Chr.11: 108072322-108072322 on Build GRCh38
rs556644154 Chr.11: 108072258-108072258 on Build GRCh38
rs977479083 Chr.11: 108072179-108072179 on Build GRCh38
rs914599938 Chr.11: 108072149-108072149 on Build GRCh38
rs934123429 Chr.11: 108072205-108072205 on Build GRCh38
rs982060810 Chr.11: 108072127-108072127 on Build GRCh38
rs1002311009 Chr.11: 108072115-108072115 on Build GRCh38
rs1016947297 Chr.11: 108072040-108072040 on Build GRCh38
rs945667876 Chr.11: 108071889-108071889 on Build GRCh38
rs962436238 Chr.11: 108071994-108071994 on Build GRCh38
rs753411259 Chr.11: 108072342-108072342 on Build GRCh38
rs1016538221 Chr.11: 108072232-108072232 on Build GRCh38
rs879147541 Chr.11: 108072272-108072272 on Build GRCh38
rs146753263 Chr.11: 108072286-108072286 on Build GRCh38
rs373547119 Chr.11: 108072287-108072287 on Build GRCh38
rs766652147 Chr.11: 108072251-108072251 on Build GRCh38
rs1051291765 Chr.11: 108072221-108072221 on Build GRCh38
rs1039829858 Chr.11: 108071974-108071974 on Build GRCh38
rs1029599436 Chr.11: 108071936-108071936 on Build GRCh38
rs998386863 Chr.11: 108071982-108071982 on Build GRCh38
rs367766251 Chr.11: 108072348-108072348 on Build GRCh38
rs553755409 Chr.11: 108072324-108072324 on Build GRCh38
rs778248461 Chr.11: 108072352-108072352 on Build GRCh38
rs374199263 Chr.11: 108072323-108072323 on Build GRCh38
rs376271612 Chr.11: 108072298-108072298 on Build GRCh38
rs755373639 Chr.11: 108072294-108072298 on Build GRCh38
rs1012594151 Chr.11: 108072280-108072280 on Build GRCh38
rs199960553 Chr.11: 108072372-108072372 on Build GRCh38
rs934941152 Chr.11: 108072131-108072131 on Build GRCh38
rs992666939 Chr.11: 108072144-108072144 on Build GRCh38
rs760484703 Chr.11: 108072225-108072225 on Build GRCh38
rs967338295 Chr.11: 108072165-108072165 on Build GRCh38
rs758276900 Chr.11: 108072364-108072364 on Build GRCh38
rs935781881 Chr.11: 108071968-108071968 on Build GRCh38
rs998481689 Chr.11: 108071880-108071880 on Build GRCh38
rs112575935 Chr.11: 108072162-108072162 on Build GRCh38
rs534498806 Chr.11: 108071988-108071988 on Build GRCh38
rs200926051 Chr.11: 108072190-108072192 on Build GRCh38
rs1041386246 Chr.11: 108071909-108071909 on Build GRCh38
rs750544241 Chr.11: 108072317-108072317 on Build GRCh38

View Full Product Details