Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119539597-119540146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
UniGene
Hs.248572
Gene Chromosome Location
Chr.X: 119538149-119565434 on Build GRCh38
Transcripts
NM_001170569.1 NM_001170570.1 NM_022101.3

SNP Detail

SNP ID Chromosome Location
rs775160550 Chr.X: 119540068-119540068 on Build GRCh38
rs750276905 Chr.X: 119539648-119539648 on Build GRCh38
rs764134233 Chr.X: 119539767-119539767 on Build GRCh38
rs771234107 Chr.X: 119539721-119539721 on Build GRCh38
rs745788287 Chr.X: 119539766-119539766 on Build GRCh38
rs749869271 Chr.X: 119539815-119539815 on Build GRCh38
rs766950247 Chr.X: 119539813-119539813 on Build GRCh38
rs771688031 Chr.X: 119540040-119540040 on Build GRCh38
rs918193208 Chr.X: 119540035-119540035 on Build GRCh38
rs916815585 Chr.X: 119539976-119539976 on Build GRCh38
rs780473535 Chr.X: 119539802-119539802 on Build GRCh38
rs768685919 Chr.X: 119539890-119539890 on Build GRCh38
rs994637087 Chr.X: 119539650-119539650 on Build GRCh38
rs200402905 Chr.X: 119540108-119540110 on Build GRCh38
rs779254765 Chr.X: 119539682-119539682 on Build GRCh38
rs566138042 Chr.X: 119540105-119540105 on Build GRCh38
rs749363300 Chr.X: 119539798-119539798 on Build GRCh38
rs867286271 Chr.X: 119540016-119540016 on Build GRCh38
rs754962708 Chr.X: 119539809-119539810 on Build GRCh38
rs761447422 Chr.X: 119539806-119539806 on Build GRCh38
rs958160680 Chr.X: 119539963-119539963 on Build GRCh38
rs763328354 Chr.X: 119540037-119540037 on Build GRCh38
rs190223379 Chr.X: 119539831-119539831 on Build GRCh38
rs775444597 Chr.X: 119539731-119539731 on Build GRCh38
rs768463891 Chr.X: 119540061-119540061 on Build GRCh38
rs749146098 Chr.X: 119539952-119539952 on Build GRCh38
rs967133891 Chr.X: 119539861-119539861 on Build GRCh38
rs770338201 Chr.X: 119539972-119539972 on Build GRCh38
rs905210861 Chr.X: 119540066-119540066 on Build GRCh38
rs1050696691 Chr.X: 119540117-119540117 on Build GRCh38
rs370514854 Chr.X: 119539685-119539685 on Build GRCh38
rs34888518 Chr.X: 119539719-119539720 on Build GRCh38
rs927993283 Chr.X: 119540033-119540033 on Build GRCh38
rs372047133 Chr.X: 119539681-119539681 on Build GRCh38
rs781716344 Chr.X: 119539899-119539899 on Build GRCh38
rs940771026 Chr.X: 119540039-119540039 on Build GRCh38
rs773406776 Chr.X: 119540015-119540015 on Build GRCh38
rs1036370187 Chr.X: 119540060-119540060 on Build GRCh38
rs748544983 Chr.X: 119539941-119539941 on Build GRCh38
rs899258838 Chr.X: 119540100-119540100 on Build GRCh38
rs781317262 Chr.X: 119539797-119539798 on Build GRCh38
rs948319546 Chr.X: 119539989-119539989 on Build GRCh38
rs756134299 Chr.X: 119539845-119539845 on Build GRCh38
rs936628458 Chr.X: 119540075-119540075 on Build GRCh38
rs373908361 Chr.X: 119539711-119539711 on Build GRCh38
rs781421862 Chr.X: 119539643-119539643 on Build GRCh38
rs747235166 Chr.X: 119539716-119539716 on Build GRCh38
rs368172135 Chr.X: 119539852-119539852 on Build GRCh38
rs755795873 Chr.X: 119539821-119539821 on Build GRCh38
rs748325295 Chr.X: 119539683-119539683 on Build GRCh38

View Full Product Details