Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61356728-61357228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561A3
Entrez Gene ID
220002
Gene Name
cytochrome b561 family member A3
Gene Aliases
CYBASC3, LCYTB
UniGene
Hs.22546
Gene Chromosome Location
Chr.11: 61348748-61362299 on Build GRCh38
Transcripts
NM_001161452.1 NM_001161454.1 NM_001300763.1 NM_153611.4 XM_011544821.1 XM_011544823.1 XM_011544824.1 XM_017017345.1 XM_017017346.1 XM_017017347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5970231 c.28C>T p.L10L Chr.11: 61357211-61357211 on Build GRCh38
5512793 c.37-5_37-3delTCT p.? Chr.11: 61356731-61356733 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945971267 Chr.11: 61357095-61357095 on Build GRCh38
rs961630228 Chr.11: 61356888-61356888 on Build GRCh38
rs972856023 Chr.11: 61356894-61356894 on Build GRCh38
rs913119174 Chr.11: 61357085-61357085 on Build GRCh38
rs980198785 Chr.11: 61356993-61356993 on Build GRCh38
rs1011162981 Chr.11: 61357074-61357074 on Build GRCh38
rs757249515 Chr.11: 61356772-61356772 on Build GRCh38
rs904243408 Chr.11: 61356813-61356813 on Build GRCh38
rs892758732 Chr.11: 61357032-61357032 on Build GRCh38
rs757621666 Chr.11: 61357111-61357111 on Build GRCh38
rs749254249 Chr.11: 61356771-61356771 on Build GRCh38
rs929861010 Chr.11: 61356868-61356868 on Build GRCh38
rs543197868 Chr.11: 61356863-61356863 on Build GRCh38
rs752606159 Chr.11: 61356753-61356755 on Build GRCh38
rs1021321474 Chr.11: 61357084-61357084 on Build GRCh38
rs965460509 Chr.11: 61357198-61357198 on Build GRCh38
rs764383982 Chr.11: 61357009-61357009 on Build GRCh38
rs2290388 Chr.11: 61356831-61356831 on Build GRCh38
rs906588245 Chr.11: 61356958-61356959 on Build GRCh38
rs1019834723 Chr.11: 61356883-61356883 on Build GRCh38
rs1057445568 Chr.11: 61357083-61357083 on Build GRCh38
rs952892224 Chr.11: 61356978-61356978 on Build GRCh38
rs530605885 Chr.11: 61356875-61356875 on Build GRCh38
rs1045168604 Chr.11: 61356879-61356879 on Build GRCh38
rs890213426 Chr.11: 61357172-61357172 on Build GRCh38
rs765970690 Chr.11: 61357031-61357031 on Build GRCh38
rs577994166 Chr.11: 61356829-61356829 on Build GRCh38
rs758399713 Chr.11: 61357155-61357157 on Build GRCh38
rs532903165 Chr.11: 61357195-61357195 on Build GRCh38
rs954382350 Chr.11: 61356872-61356872 on Build GRCh38
rs760302280 Chr.11: 61356968-61356968 on Build GRCh38
rs12291349 Chr.11: 61356874-61356874 on Build GRCh38
rs547657560 Chr.11: 61356857-61356857 on Build GRCh38
rs904351250 Chr.11: 61357099-61357099 on Build GRCh38
rs751942711 Chr.11: 61357096-61357096 on Build GRCh38
rs781536094 Chr.11: 61357126-61357126 on Build GRCh38
rs560804273 Chr.11: 61356843-61356843 on Build GRCh38
rs750116768 Chr.11: 61356962-61356962 on Build GRCh38
rs368149383 Chr.11: 61357131-61357131 on Build GRCh38
rs753323262 Chr.11: 61357097-61357097 on Build GRCh38
rs1008528292 Chr.11: 61357191-61357191 on Build GRCh38
rs938672353 Chr.11: 61357081-61357081 on Build GRCh38
rs1003719090 Chr.11: 61356992-61356992 on Build GRCh38
rs550723461 Chr.11: 61357125-61357125 on Build GRCh38
rs745685214 Chr.11: 61357201-61357201 on Build GRCh38

View Full Product Details