Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144464084-144464589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYHR1
Entrez Gene ID
50626
Gene Name
cysteine and histidine rich 1
Gene Aliases
CHRP
UniGene
Hs.459379
Gene Chromosome Location
Chr.8: 144449582-144465692 on Build GRCh38
Transcripts
NM_001129888.1 NM_032687.3 NM_138496.1 XM_006716560.3 XM_006716561.3 XM_017013463.1 XM_017013464.1 XM_017013465.1 XM_017013466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4977123 c.469G>A p.E157K Chr.8: 144464237-144464237 on Build GRCh38
5503439 c.307G>A p.A103T Chr.8: 144464399-144464399 on Build GRCh38
5430204 c.446C>G p.A149G Chr.8: 144464260-144464260 on Build GRCh38
5835523 c.484delC p.L162fs*14 Chr.8: 144464222-144464222 on Build GRCh38
5835522 c.545_546insT p.A183fs*>11 Chr.8: 144464160-144464161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770873506 Chr.8: 144464390-144464390 on Build GRCh38
rs370071758 Chr.8: 144464136-144464136 on Build GRCh38
rs371644139 Chr.8: 144464324-144464324 on Build GRCh38
rs754370713 Chr.8: 144464428-144464428 on Build GRCh38
rs762099793 Chr.8: 144464276-144464276 on Build GRCh38
rs770367627 Chr.8: 144464199-144464199 on Build GRCh38
rs1035774356 Chr.8: 144464445-144464445 on Build GRCh38
rs543509475 Chr.8: 144464162-144464162 on Build GRCh38
rs757794751 Chr.8: 144464267-144464267 on Build GRCh38
rs764367953 Chr.8: 144464233-144464235 on Build GRCh38
rs78088422 Chr.8: 144464373-144464373 on Build GRCh38
rs764849123 Chr.8: 144464443-144464443 on Build GRCh38
rs749590903 Chr.8: 144464120-144464120 on Build GRCh38
rs761863174 Chr.8: 144464232-144464232 on Build GRCh38
rs77204173 Chr.8: 144464214-144464214 on Build GRCh38
rs576227276 Chr.8: 144464112-144464112 on Build GRCh38
rs750326550 Chr.8: 144464543-144464543 on Build GRCh38
rs754526850 Chr.8: 144464394-144464394 on Build GRCh38
rs778756479 Chr.8: 144464329-144464329 on Build GRCh38
rs764965148 Chr.8: 144464153-144464153 on Build GRCh38
rs752221160 Chr.8: 144464531-144464531 on Build GRCh38
rs779440226 Chr.8: 144464127-144464127 on Build GRCh38
rs775672668 Chr.8: 144464358-144464358 on Build GRCh38
rs751572780 Chr.8: 144464158-144464158 on Build GRCh38
rs772258449 Chr.8: 144464499-144464499 on Build GRCh38
rs774490079 Chr.8: 144464406-144464406 on Build GRCh38
rs760139500 Chr.8: 144464527-144464527 on Build GRCh38
rs747803036 Chr.8: 144464385-144464385 on Build GRCh38
rs199539352 Chr.8: 144464302-144464302 on Build GRCh38
rs748648310 Chr.8: 144464133-144464133 on Build GRCh38
rs889414226 Chr.8: 144464475-144464475 on Build GRCh38
rs762037494 Chr.8: 144464416-144464416 on Build GRCh38
rs144387246 Chr.8: 144464537-144464537 on Build GRCh38
rs966082253 Chr.8: 144464167-144464167 on Build GRCh38
rs780014273 Chr.8: 144464550-144464550 on Build GRCh38
rs146960526 Chr.8: 144464281-144464281 on Build GRCh38
rs757062164 Chr.8: 144464464-144464464 on Build GRCh38
rs1021522989 Chr.8: 144464498-144464498 on Build GRCh38
rs948382010 Chr.8: 144464234-144464234 on Build GRCh38
rs368527220 Chr.8: 144464108-144464108 on Build GRCh38
rs762990071 Chr.8: 144464216-144464217 on Build GRCh38
rs766525030 Chr.8: 144464240-144464240 on Build GRCh38
rs764797730 Chr.8: 144464529-144464529 on Build GRCh38
rs149259609 Chr.8: 144464280-144464280 on Build GRCh38
rs752016150 Chr.8: 144464262-144464262 on Build GRCh38
rs766572350 Chr.8: 144464164-144464164 on Build GRCh38
rs759737579 Chr.8: 144464244-144464244 on Build GRCh38
rs759681184 Chr.8: 144464285-144464285 on Build GRCh38
rs747942669 Chr.8: 144464332-144464332 on Build GRCh38
rs756077860 Chr.8: 144464548-144464548 on Build GRCh38
rs776484785 Chr.8: 144464391-144464391 on Build GRCh38
rs756644723 Chr.8: 144464313-144464316 on Build GRCh38
rs958556235 Chr.8: 144464453-144464453 on Build GRCh38
rs769710867 Chr.8: 144464357-144464357 on Build GRCh38
rs758206016 Chr.8: 144464386-144464386 on Build GRCh38
rs761161002 Chr.8: 144464144-144464144 on Build GRCh38
rs374378728 Chr.8: 144464309-144464309 on Build GRCh38
rs766833653 Chr.8: 144464377-144464377 on Build GRCh38
rs2721167 Chr.8: 144464532-144464532 on Build GRCh38
rs750123440 Chr.8: 144464170-144464170 on Build GRCh38
rs750926515 Chr.8: 144464284-144464285 on Build GRCh38
rs765359523 Chr.8: 144464289-144464289 on Build GRCh38
rs200369634 Chr.8: 144464360-144464360 on Build GRCh38
rs753937256 Chr.8: 144464242-144464242 on Build GRCh38
rs779632554 Chr.8: 144464321-144464321 on Build GRCh38
rs765360904 Chr.8: 144464245-144464245 on Build GRCh38
rs377084522 Chr.8: 144464124-144464124 on Build GRCh38
rs773213525 Chr.8: 144464349-144464349 on Build GRCh38
rs748211450 Chr.8: 144464563-144464563 on Build GRCh38
rs753364848 Chr.8: 144464384-144464384 on Build GRCh38
rs952212804 Chr.8: 144464565-144464565 on Build GRCh38
rs376180792 Chr.8: 144464538-144464538 on Build GRCh38
rs143628277 Chr.8: 144464283-144464283 on Build GRCh38
rs547376015 Chr.8: 144464275-144464276 on Build GRCh38
rs764328897 Chr.8: 144464298-144464298 on Build GRCh38
rs767597784 Chr.8: 144464104-144464104 on Build GRCh38
rs371837585 Chr.8: 144464522-144464522 on Build GRCh38
rs148681176 Chr.8: 144464304-144464304 on Build GRCh38
rs777134791 Chr.8: 144464516-144464516 on Build GRCh38
rs75845238 Chr.8: 144464536-144464536 on Build GRCh38
rs368285970 Chr.8: 144464311-144464311 on Build GRCh38
rs375882093 Chr.8: 144464292-144464292 on Build GRCh38
rs545265637 Chr.8: 144464116-144464116 on Build GRCh38
rs1018944792 Chr.8: 144464159-144464159 on Build GRCh38
rs762642971 Chr.8: 144464215-144464215 on Build GRCh38
rs578183711 Chr.8: 144464198-144464198 on Build GRCh38
rs753025595 Chr.8: 144464253-144464253 on Build GRCh38
rs758621338 Chr.8: 144464256-144464256 on Build GRCh38
rs775184067 Chr.8: 144464154-144464154 on Build GRCh38
rs199783284 Chr.8: 144464423-144464423 on Build GRCh38
rs752345868 Chr.8: 144464447-144464447 on Build GRCh38
rs774091305 Chr.8: 144464224-144464224 on Build GRCh38
rs919087758 Chr.8: 144464184-144464184 on Build GRCh38
rs762849718 Chr.8: 144464155-144464155 on Build GRCh38
rs772055597 Chr.8: 144464340-144464340 on Build GRCh38
rs768795345 Chr.8: 144464398-144464398 on Build GRCh38
rs139696517 Chr.8: 144464457-144464457 on Build GRCh38
rs768898712 Chr.8: 144464160-144464160 on Build GRCh38
rs915862154 Chr.8: 144464337-144464337 on Build GRCh38
rs764944855 Chr.8: 144464138-144464138 on Build GRCh38
rs778210137 Chr.8: 144464259-144464259 on Build GRCh38
rs201021717 Chr.8: 144464194-144464194 on Build GRCh38
rs199699589 Chr.8: 144464394-144464394 on Build GRCh38
rs748331926 Chr.8: 144464490-144464490 on Build GRCh38
rs781697158 Chr.8: 144464268-144464268 on Build GRCh38
rs771204323 Chr.8: 144464507-144464507 on Build GRCh38
rs367711533 Chr.8: 144464426-144464426 on Build GRCh38
rs142152732 Chr.8: 144464284-144464284 on Build GRCh38
rs748438702 Chr.8: 144464185-144464185 on Build GRCh38
rs954366035 Chr.8: 144464547-144464547 on Build GRCh38
rs773532604 Chr.8: 144464425-144464425 on Build GRCh38
rs761095771 Chr.8: 144464504-144464504 on Build GRCh38
rs776952885 Chr.8: 144464191-144464191 on Build GRCh38
rs763917343 Chr.8: 144464156-144464156 on Build GRCh38
rs1007756521 Chr.8: 144464494-144464494 on Build GRCh38
rs146814251 Chr.8: 144464478-144464478 on Build GRCh38
rs753434966 Chr.8: 144464391-144464392 on Build GRCh38
rs777729434 Chr.8: 144464388-144464388 on Build GRCh38
rs745798293 Chr.8: 144464476-144464476 on Build GRCh38
rs772426239 Chr.8: 144464134-144464134 on Build GRCh38
rs139843668 Chr.8: 144464282-144464282 on Build GRCh38
rs745363210 Chr.8: 144464276-144464276 on Build GRCh38
rs763256167 Chr.8: 144464296-144464296 on Build GRCh38
rs746879021 Chr.8: 144464389-144464389 on Build GRCh38
rs372867926 Chr.8: 144464486-144464486 on Build GRCh38
rs762160865 Chr.8: 144464369-144464369 on Build GRCh38
rs199972231 Chr.8: 144464202-144464202 on Build GRCh38
rs542765903 Chr.8: 144464518-144464518 on Build GRCh38
rs781188457 Chr.8: 144464471-144464471 on Build GRCh38
rs756668440 Chr.8: 144464376-144464376 on Build GRCh38
rs776196363 Chr.8: 144464145-144464145 on Build GRCh38
rs767873228 Chr.8: 144464371-144464371 on Build GRCh38
rs766896345 Chr.8: 144464427-144464427 on Build GRCh38
rs759202673 Chr.8: 144464152-144464152 on Build GRCh38
rs746800437 Chr.8: 144464460-144464460 on Build GRCh38
rs373353154 Chr.8: 144464166-144464166 on Build GRCh38
rs571153730 Chr.8: 144464359-144464359 on Build GRCh38
rs148121201 Chr.8: 144464384-144464385 on Build GRCh38
rs773814209 Chr.8: 144464135-144464135 on Build GRCh38
rs774618679 Chr.8: 144464362-144464362 on Build GRCh38
rs35095467 Chr.8: 144464374-144464374 on Build GRCh38
rs868278812 Chr.8: 144464491-144464491 on Build GRCh38
rs560617378 Chr.8: 144464205-144464205 on Build GRCh38
rs756311768 Chr.8: 144464110-144464110 on Build GRCh38
rs754062946 Chr.8: 144464168-144464168 on Build GRCh38
rs775498418 Chr.8: 144464133-144464134 on Build GRCh38
rs750996356 Chr.8: 144464310-144464310 on Build GRCh38
rs776144196 Chr.8: 144464201-144464201 on Build GRCh38
rs760672462 Chr.8: 144464354-144464354 on Build GRCh38
rs898772897 Chr.8: 144464331-144464331 on Build GRCh38
rs769405048 Chr.8: 144464279-144464279 on Build GRCh38
rs768664885 Chr.8: 144464480-144464480 on Build GRCh38
rs573534662 Chr.8: 144464483-144464483 on Build GRCh38
rs773363700 Chr.8: 144464503-144464503 on Build GRCh38
rs1044264615 Chr.8: 144464432-144464432 on Build GRCh38
rs536518241 Chr.8: 144464436-144464436 on Build GRCh38
rs754439967 Chr.8: 144464441-144464441 on Build GRCh38
rs1045415312 Chr.8: 144464235-144464235 on Build GRCh38

View Full Product Details