Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41096066-41096587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A13
Entrez Gene ID
1553
Gene Name
cytochrome P450 family 2 subfamily A member 13
Gene Aliases
CPAD, CYP2A, CYPIIA13
UniGene
Hs.567252
Gene Chromosome Location
Chr.19: 41088451-41096195 on Build GRCh38
Transcripts
NM_000766.4

SNP Detail

SNP ID Chromosome Location
rs969971437 Chr.19: 41096271-41096271 on Build GRCh38
rs1013793745 Chr.19: 41096242-41096242 on Build GRCh38
rs563647043 Chr.19: 41096240-41096240 on Build GRCh38
rs1006786989 Chr.19: 41096434-41096434 on Build GRCh38
rs111472610 Chr.19: 41096375-41096375 on Build GRCh38
rs903927646 Chr.19: 41096373-41096373 on Build GRCh38
rs777665388 Chr.19: 41096381-41096381 on Build GRCh38
rs978998169 Chr.19: 41096540-41096540 on Build GRCh38
rs113572237 Chr.19: 41096534-41096534 on Build GRCh38
rs924471185 Chr.19: 41096544-41096544 on Build GRCh38
rs552858039 Chr.19: 41096199-41096199 on Build GRCh38
rs933192235 Chr.19: 41096332-41096332 on Build GRCh38
rs954378808 Chr.19: 41096489-41096489 on Build GRCh38
rs73563347 Chr.19: 41096327-41096327 on Build GRCh38
rs142385410 Chr.19: 41096299-41096299 on Build GRCh38
rs910447683 Chr.19: 41096521-41096521 on Build GRCh38
rs8192790 Chr.19: 41096339-41096339 on Build GRCh38
rs150022558 Chr.19: 41096403-41096403 on Build GRCh38
rs903960994 Chr.19: 41096213-41096213 on Build GRCh38
rs971723353 Chr.19: 41096315-41096315 on Build GRCh38
rs761290013 Chr.19: 41096254-41096254 on Build GRCh38
rs1016407573 Chr.19: 41096300-41096300 on Build GRCh38
rs774493945 Chr.19: 41096090-41096090 on Build GRCh38
rs768668892 Chr.19: 41096502-41096502 on Build GRCh38
rs1709084 Chr.19: 41096565-41096565 on Build GRCh38
rs151309514 Chr.19: 41096333-41096333 on Build GRCh38
rs1052346373 Chr.19: 41096396-41096396 on Build GRCh38
rs192657620 Chr.19: 41096431-41096431 on Build GRCh38
rs896739330 Chr.19: 41096236-41096236 on Build GRCh38
rs77913771 Chr.19: 41096483-41096483 on Build GRCh38
rs549633782 Chr.19: 41096170-41096170 on Build GRCh38
rs1044216899 Chr.19: 41096126-41096126 on Build GRCh38
rs187681654 Chr.19: 41096266-41096266 on Build GRCh38
rs8192791 Chr.19: 41096351-41096351 on Build GRCh38
rs570325924 Chr.19: 41096387-41096387 on Build GRCh38
rs1013760746 Chr.19: 41096406-41096406 on Build GRCh38
rs961753984 Chr.19: 41096472-41096472 on Build GRCh38
rs1024323051 Chr.19: 41096417-41096417 on Build GRCh38
rs541737986 Chr.19: 41096229-41096229 on Build GRCh38
rs570875313 Chr.19: 41096457-41096457 on Build GRCh38
rs73563350 Chr.19: 41096539-41096539 on Build GRCh38
rs1029976297 Chr.19: 41096485-41096485 on Build GRCh38
rs148803262 Chr.19: 41096226-41096226 on Build GRCh38
rs749393990 Chr.19: 41096501-41096501 on Build GRCh38
rs763424203 Chr.19: 41096243-41096247 on Build GRCh38
rs961848505 Chr.19: 41096312-41096312 on Build GRCh38
rs553461983 Chr.19: 41096510-41096510 on Build GRCh38
rs762183074 Chr.19: 41096151-41096151 on Build GRCh38
rs183253628 Chr.19: 41096093-41096093 on Build GRCh38

View Full Product Details