Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41122795-41123194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2F1
Entrez Gene ID
1572
Gene Name
cytochrome P450 family 2 subfamily F member 1
Gene Aliases
C2F1, CYP2F, CYPIIF1
UniGene
Hs.558318
Gene Chromosome Location
Chr.19: 41114292-41128381 on Build GRCh38
Transcripts
NM_000774.4 NR_135528.1 XM_011526551.2 XM_011526552.2 XM_011526553.2 XM_011526554.2 XM_011526555.2 XM_017026384.1 XM_017026385.1 XM_017026386.1 XM_017026387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1316827 c.949T>A p.Y317N Chr.19: 41122948-41122948 on Build GRCh38
1316826 c.947A>C p.K316T Chr.19: 41122946-41122946 on Build GRCh38
1316829 c.951C>A p.Y317* Chr.19: 41122950-41122950 on Build GRCh38
1316828 c.950A>G p.Y317C Chr.19: 41122949-41122949 on Build GRCh38
6525025 c.861C>A p.T287T Chr.19: 41122860-41122860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775255579 Chr.19: 41123031-41123031 on Build GRCh38
rs558960916 Chr.19: 41123072-41123072 on Build GRCh38
rs972713547 Chr.19: 41122824-41122824 on Build GRCh38
rs777262251 Chr.19: 41123105-41123105 on Build GRCh38
rs372508830 Chr.19: 41122992-41122992 on Build GRCh38
rs777056947 Chr.19: 41123035-41123035 on Build GRCh38
rs770780630 Chr.19: 41123016-41123016 on Build GRCh38
rs745710850 Chr.19: 41123114-41123114 on Build GRCh38
rs757660495 Chr.19: 41122922-41122922 on Build GRCh38
rs58894601 Chr.19: 41123110-41123110 on Build GRCh38
rs144438871 Chr.19: 41123153-41123153 on Build GRCh38
rs755708391 Chr.19: 41123082-41123082 on Build GRCh38
rs149278083 Chr.19: 41122856-41122856 on Build GRCh38
rs762128864 Chr.19: 41123039-41123039 on Build GRCh38
rs199736867 Chr.19: 41122950-41122950 on Build GRCh38
rs768645487 Chr.19: 41123154-41123154 on Build GRCh38
rs762482043 Chr.19: 41123083-41123083 on Build GRCh38
rs747551722 Chr.19: 41122938-41122938 on Build GRCh38
rs571508288 Chr.19: 41122816-41122816 on Build GRCh38
rs754087330 Chr.19: 41122976-41122976 on Build GRCh38
rs201357530 Chr.19: 41122927-41122927 on Build GRCh38
rs774372143 Chr.19: 41122825-41122825 on Build GRCh38
rs528392605 Chr.19: 41123113-41123113 on Build GRCh38
rs751469261 Chr.19: 41122894-41122894 on Build GRCh38
rs761918918 Chr.19: 41122909-41122910 on Build GRCh38
rs765690510 Chr.19: 41122987-41122987 on Build GRCh38
rs760853657 Chr.19: 41123033-41123033 on Build GRCh38
rs759466316 Chr.19: 41122953-41122953 on Build GRCh38
rs764412091 Chr.19: 41122970-41122970 on Build GRCh38
rs778198076 Chr.19: 41122830-41122830 on Build GRCh38
rs578082249 Chr.19: 41123070-41123070 on Build GRCh38
rs772699453 Chr.19: 41122939-41122939 on Build GRCh38
rs139430706 Chr.19: 41122884-41122884 on Build GRCh38
rs749122119 Chr.19: 41122942-41122942 on Build GRCh38
rs780092368 Chr.19: 41123132-41123132 on Build GRCh38
rs562735786 Chr.19: 41123034-41123034 on Build GRCh38
rs775451080 Chr.19: 41123161-41123161 on Build GRCh38
rs770690821 Chr.19: 41122949-41122949 on Build GRCh38
rs771737659 Chr.19: 41123117-41123117 on Build GRCh38
rs146496115 Chr.19: 41122926-41122926 on Build GRCh38
rs780375537 Chr.19: 41122996-41122996 on Build GRCh38
rs771583336 Chr.19: 41122837-41122837 on Build GRCh38
rs372975136 Chr.19: 41123013-41123013 on Build GRCh38
rs750838904 Chr.19: 41123041-41123041 on Build GRCh38
rs369146753 Chr.19: 41122916-41122916 on Build GRCh38
rs374776224 Chr.19: 41122893-41122893 on Build GRCh38
rs543160199 Chr.19: 41123022-41123022 on Build GRCh38
rs554179755 Chr.19: 41122900-41122900 on Build GRCh38
rs140997784 Chr.19: 41122960-41122960 on Build GRCh38
rs538835299 Chr.19: 41122817-41122817 on Build GRCh38
rs772073383 Chr.19: 41123029-41123029 on Build GRCh38
rs149668363 Chr.19: 41122839-41122839 on Build GRCh38
rs756494789 Chr.19: 41122827-41122827 on Build GRCh38
rs1012834241 Chr.19: 41123107-41123107 on Build GRCh38
rs758955642 Chr.19: 41122993-41122993 on Build GRCh38
rs923573842 Chr.19: 41122833-41122833 on Build GRCh38
rs763185947 Chr.19: 41123064-41123064 on Build GRCh38
rs773071416 Chr.19: 41122877-41122877 on Build GRCh38
rs564173383 Chr.19: 41122859-41122859 on Build GRCh38
rs369957652 Chr.19: 41123008-41123008 on Build GRCh38
rs756919598 Chr.19: 41123112-41123112 on Build GRCh38
rs144137084 Chr.19: 41122904-41122904 on Build GRCh38
rs745595856 Chr.19: 41123025-41123025 on Build GRCh38
rs895479178 Chr.19: 41123100-41123100 on Build GRCh38
rs776862361 Chr.19: 41123160-41123160 on Build GRCh38
rs747723892 Chr.19: 41123007-41123007 on Build GRCh38
rs767293784 Chr.19: 41122959-41122959 on Build GRCh38
rs376675412 Chr.19: 41122995-41122995 on Build GRCh38
rs776202770 Chr.19: 41122869-41122869 on Build GRCh38
rs752943800 Chr.19: 41122822-41122822 on Build GRCh38
rs756365554 Chr.19: 41122905-41122905 on Build GRCh38
rs746554629 Chr.19: 41122845-41122845 on Build GRCh38
rs372625854 Chr.19: 41122870-41122870 on Build GRCh38
rs754281294 Chr.19: 41122921-41122921 on Build GRCh38
rs780756537 Chr.19: 41122929-41122929 on Build GRCh38
rs371209937 Chr.19: 41122836-41122836 on Build GRCh38
rs777451701 Chr.19: 41123010-41123010 on Build GRCh38
rs765495577 Chr.19: 41123040-41123040 on Build GRCh38
rs200603349 Chr.19: 41122986-41122986 on Build GRCh38
rs769262528 Chr.19: 41122876-41122876 on Build GRCh38
rs779607821 Chr.19: 41122923-41122923 on Build GRCh38

View Full Product Details