Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99844095-99844619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A43
Entrez Gene ID
64816
Gene Name
cytochrome P450 family 3 subfamily A member 43
Gene Aliases
-
UniGene
Hs.306220
Gene Chromosome Location
Chr.7: 99828013-99867138 on Build GRCh38
Transcripts
NM_001278921.1 NM_022820.4 NM_057095.2 NM_057096.3 NR_103868.1 NR_103869.1 XM_017012542.1 XM_017012543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093888 c.220C>A p.L74M Chr.7: 99844144-99844144 on Build GRCh38
3883886 c.274A>G p.T92A Chr.7: 99844198-99844198 on Build GRCh38
6871399 c.293G>A p.C98Y Chr.7: 99844217-99844217 on Build GRCh38
5408386 c.273A>G p.K91K Chr.7: 99844197-99844197 on Build GRCh38
7100368 c.285G>A p.V95V Chr.7: 99844209-99844209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191989738 Chr.7: 99844476-99844476 on Build GRCh38
rs975180383 Chr.7: 99844182-99844182 on Build GRCh38
rs772177902 Chr.7: 99844221-99844221 on Build GRCh38
rs891949649 Chr.7: 99844474-99844474 on Build GRCh38
rs773210923 Chr.7: 99844263-99844263 on Build GRCh38
rs754186707 Chr.7: 99844123-99844123 on Build GRCh38
rs935717534 Chr.7: 99844410-99844410 on Build GRCh38
rs148195838 Chr.7: 99844142-99844142 on Build GRCh38
rs957585400 Chr.7: 99844321-99844321 on Build GRCh38
rs747927876 Chr.7: 99844159-99844159 on Build GRCh38
rs45573333 Chr.7: 99844359-99844359 on Build GRCh38
rs147384596 Chr.7: 99844206-99844206 on Build GRCh38
rs760774417 Chr.7: 99844180-99844180 on Build GRCh38
rs551395170 Chr.7: 99844402-99844402 on Build GRCh38
rs368393851 Chr.7: 99844155-99844155 on Build GRCh38
rs1038182227 Chr.7: 99844127-99844127 on Build GRCh38
rs150654722 Chr.7: 99844347-99844347 on Build GRCh38
rs538636991 Chr.7: 99844358-99844358 on Build GRCh38
rs1019198122 Chr.7: 99844535-99844535 on Build GRCh38
rs1031826582 Chr.7: 99844317-99844317 on Build GRCh38
rs117077404 Chr.7: 99844545-99844545 on Build GRCh38
rs374867022 Chr.7: 99844165-99844165 on Build GRCh38
rs899629424 Chr.7: 99844464-99844464 on Build GRCh38
rs757979352 Chr.7: 99844144-99844144 on Build GRCh38
rs778567825 Chr.7: 99844140-99844140 on Build GRCh38
rs540428216 Chr.7: 99844302-99844302 on Build GRCh38
rs764333132 Chr.7: 99844121-99844121 on Build GRCh38
rs1052888354 Chr.7: 99844446-99844446 on Build GRCh38
rs1038218210 Chr.7: 99844459-99844459 on Build GRCh38
rs764610145 Chr.7: 99844185-99844185 on Build GRCh38
rs533668916 Chr.7: 99844499-99844499 on Build GRCh38
rs757871892 Chr.7: 99844135-99844135 on Build GRCh38
rs746924665 Chr.7: 99844148-99844148 on Build GRCh38
rs892893165 Chr.7: 99844561-99844561 on Build GRCh38
rs769491358 Chr.7: 99844162-99844162 on Build GRCh38
rs769615848 Chr.7: 99844247-99844247 on Build GRCh38
rs748042277 Chr.7: 99844238-99844238 on Build GRCh38
rs754311708 Chr.7: 99844186-99844186 on Build GRCh38
rs998650681 Chr.7: 99844315-99844315 on Build GRCh38
rs575211777 Chr.7: 99844491-99844491 on Build GRCh38
rs929283249 Chr.7: 99844361-99844361 on Build GRCh38
rs942098335 Chr.7: 99844450-99844450 on Build GRCh38
rs142965961 Chr.7: 99844163-99844163 on Build GRCh38
rs1052764573 Chr.7: 99844555-99844555 on Build GRCh38
rs565953842 Chr.7: 99844329-99844329 on Build GRCh38
rs952187634 Chr.7: 99844213-99844213 on Build GRCh38
rs757573296 Chr.7: 99844193-99844193 on Build GRCh38
rs527828976 Chr.7: 99844237-99844237 on Build GRCh38
rs909306249 Chr.7: 99844435-99844435 on Build GRCh38
rs757580967 Chr.7: 99844125-99844125 on Build GRCh38
rs779683976 Chr.7: 99844216-99844216 on Build GRCh38
rs181668317 Chr.7: 99844364-99844364 on Build GRCh38
rs775586166 Chr.7: 99844176-99844176 on Build GRCh38
rs765762180 Chr.7: 99844200-99844201 on Build GRCh38
rs376007798 Chr.7: 99844147-99844147 on Build GRCh38
rs529186603 Chr.7: 99844346-99844346 on Build GRCh38
rs990744579 Chr.7: 99844323-99844323 on Build GRCh38
rs201274854 Chr.7: 99844134-99844134 on Build GRCh38
rs908353844 Chr.7: 99844280-99844280 on Build GRCh38
rs1011293092 Chr.7: 99844575-99844575 on Build GRCh38
rs754469265 Chr.7: 99844236-99844236 on Build GRCh38
rs767561695 Chr.7: 99844170-99844170 on Build GRCh38
rs746478882 Chr.7: 99844226-99844226 on Build GRCh38
rs890824732 Chr.7: 99844501-99844501 on Build GRCh38
rs746658949 Chr.7: 99844352-99844352 on Build GRCh38
rs765590792 Chr.7: 99844200-99844200 on Build GRCh38
rs141131428 Chr.7: 99844201-99844201 on Build GRCh38
rs187199266 Chr.7: 99844400-99844400 on Build GRCh38

View Full Product Details