Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3959167-3959660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPK3
Entrez Gene ID
1613
Gene Name
death associated protein kinase 3
Gene Aliases
DLK, ZIP, ZIPK
UniGene
Hs.631844
Gene Chromosome Location
Chr.19: 3958453-3971123 on Build GRCh38
Transcripts
NM_001348.2 XM_005259508.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4077904 c.850C>T p.R284C Chr.19: 3959616-3959616 on Build GRCh38
5365487 c.1065G>A p.A355A Chr.19: 3959401-3959401 on Build GRCh38
7271224 c.830C>T p.A277V Chr.19: 3959636-3959636 on Build GRCh38
7238954 c.1137G>T p.R379R Chr.19: 3959329-3959329 on Build GRCh38
6409219 c.1285T>A p.S429T Chr.19: 3959181-3959181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750281566 Chr.19: 3959201-3959201 on Build GRCh38
rs746361778 Chr.19: 3959347-3959347 on Build GRCh38
rs764416419 Chr.19: 3959281-3959281 on Build GRCh38
rs759513536 Chr.19: 3959383-3959383 on Build GRCh38
rs368652868 Chr.19: 3959461-3959461 on Build GRCh38
rs750145142 Chr.19: 3959289-3959289 on Build GRCh38
rs866821340 Chr.19: 3959526-3959526 on Build GRCh38
rs973700357 Chr.19: 3959615-3959615 on Build GRCh38
rs77999895 Chr.19: 3959188-3959188 on Build GRCh38
rs768789939 Chr.19: 3959431-3959431 on Build GRCh38
rs767578366 Chr.19: 3959263-3959263 on Build GRCh38
rs200263027 Chr.19: 3959524-3959524 on Build GRCh38
rs773577945 Chr.19: 3959558-3959558 on Build GRCh38
rs920776635 Chr.19: 3959538-3959562 on Build GRCh38
rs111719490 Chr.19: 3959477-3959477 on Build GRCh38
rs200352688 Chr.19: 3959301-3959301 on Build GRCh38
rs769532059 Chr.19: 3959232-3959232 on Build GRCh38
rs200817833 Chr.19: 3959628-3959628 on Build GRCh38
rs748198560 Chr.19: 3959635-3959635 on Build GRCh38
rs746472976 Chr.19: 3959212-3959212 on Build GRCh38
rs1034652061 Chr.19: 3959518-3959518 on Build GRCh38
rs1025063393 Chr.19: 3959564-3959564 on Build GRCh38
rs531962501 Chr.19: 3959346-3959346 on Build GRCh38
rs780476853 Chr.19: 3959489-3959489 on Build GRCh38
rs759543762 Chr.19: 3959256-3959256 on Build GRCh38
rs755678108 Chr.19: 3959323-3959323 on Build GRCh38
rs577336858 Chr.19: 3959611-3959611 on Build GRCh38
rs777499933 Chr.19: 3959329-3959329 on Build GRCh38
rs779229348 Chr.19: 3959409-3959409 on Build GRCh38
rs370168020 Chr.19: 3959422-3959422 on Build GRCh38
rs201287470 Chr.19: 3959322-3959322 on Build GRCh38
rs780562643 Chr.19: 3959427-3959427 on Build GRCh38
rs760637791 Chr.19: 3959597-3959597 on Build GRCh38
rs752202554 Chr.19: 3959387-3959387 on Build GRCh38
rs56251209 Chr.19: 3959446-3959446 on Build GRCh38
rs768714914 Chr.19: 3959358-3959358 on Build GRCh38
rs749691110 Chr.19: 3959243-3959243 on Build GRCh38
rs772529170 Chr.19: 3959418-3959418 on Build GRCh38
rs552081970 Chr.19: 3959436-3959436 on Build GRCh38
rs568467540 Chr.19: 3959509-3959509 on Build GRCh38
rs201446546 Chr.19: 3959487-3959487 on Build GRCh38
rs906434876 Chr.19: 3959392-3959392 on Build GRCh38
rs147011394 Chr.19: 3959348-3959348 on Build GRCh38
rs958771540 Chr.19: 3959349-3959349 on Build GRCh38
rs775204534 Chr.19: 3959591-3959591 on Build GRCh38
rs780075017 Chr.19: 3959211-3959211 on Build GRCh38
rs985748144 Chr.19: 3959317-3959317 on Build GRCh38
rs767590822 Chr.19: 3959386-3959386 on Build GRCh38
rs942297952 Chr.19: 3959420-3959420 on Build GRCh38
rs368187481 Chr.19: 3959614-3959614 on Build GRCh38
rs775606509 Chr.19: 3959271-3959271 on Build GRCh38
rs958583859 Chr.19: 3959519-3959519 on Build GRCh38
rs1034377541 Chr.19: 3959205-3959205 on Build GRCh38
rs760594069 Chr.19: 3959464-3959464 on Build GRCh38
rs747786647 Chr.19: 3959223-3959223 on Build GRCh38
rs759204602 Chr.19: 3959544-3959544 on Build GRCh38
rs774935995 Chr.19: 3959253-3959253 on Build GRCh38
rs778568680 Chr.19: 3959484-3959484 on Build GRCh38
rs756397002 Chr.19: 3959200-3959200 on Build GRCh38
rs922141244 Chr.19: 3959619-3959619 on Build GRCh38
rs774762127 Chr.19: 3959339-3959339 on Build GRCh38
rs199804056 Chr.19: 3959602-3959602 on Build GRCh38
rs759423522 Chr.19: 3959458-3959458 on Build GRCh38
rs769104946 Chr.19: 3959521-3959521 on Build GRCh38
rs755036580 Chr.19: 3959310-3959310 on Build GRCh38
rs1012048987 Chr.19: 3959236-3959236 on Build GRCh38
rs763980607 Chr.19: 3959600-3959600 on Build GRCh38
rs529645237 Chr.19: 3959375-3959375 on Build GRCh38
rs758239546 Chr.19: 3959297-3959297 on Build GRCh38
rs375140792 Chr.19: 3959365-3959365 on Build GRCh38
rs537660810 Chr.19: 3959460-3959460 on Build GRCh38
rs560829558 Chr.19: 3959364-3959364 on Build GRCh38
rs781570365 Chr.19: 3959631-3959631 on Build GRCh38
rs761336141 Chr.19: 3959610-3959610 on Build GRCh38
rs967919275 Chr.19: 3959261-3959261 on Build GRCh38
rs377026101 Chr.19: 3959415-3959415 on Build GRCh38
rs775516777 Chr.19: 3959352-3959352 on Build GRCh38
rs878855905 Chr.19: 3959636-3959636 on Build GRCh38
rs954890165 Chr.19: 3959506-3959506 on Build GRCh38
rs369672101 Chr.19: 3959234-3959234 on Build GRCh38
rs771329507 Chr.19: 3959335-3959335 on Build GRCh38
rs753339617 Chr.19: 3959473-3959473 on Build GRCh38
rs983531332 Chr.19: 3959595-3959595 on Build GRCh38
rs751550544 Chr.19: 3959302-3959302 on Build GRCh38
rs910807209 Chr.19: 3959527-3959527 on Build GRCh38
rs756917160 Chr.19: 3959403-3959403 on Build GRCh38
rs181345641 Chr.19: 3959394-3959394 on Build GRCh38
rs776930359 Chr.19: 3959367-3959367 on Build GRCh38
rs201738241 Chr.19: 3959612-3959612 on Build GRCh38
rs546562367 Chr.19: 3959231-3959231 on Build GRCh38
rs770089058 Chr.19: 3959549-3959549 on Build GRCh38
rs751921245 Chr.19: 3959627-3959627 on Build GRCh38
rs372041714 Chr.19: 3959471-3959471 on Build GRCh38
rs559756449 Chr.19: 3959379-3959379 on Build GRCh38
rs745590940 Chr.19: 3959580-3959580 on Build GRCh38
rs993029019 Chr.19: 3959548-3959548 on Build GRCh38
rs766719686 Chr.19: 3959620-3959620 on Build GRCh38
rs141435903 Chr.19: 3959246-3959246 on Build GRCh38
rs199762113 Chr.19: 3959330-3959330 on Build GRCh38
rs900684606 Chr.19: 3959435-3959435 on Build GRCh38
rs1047724607 Chr.19: 3959412-3959412 on Build GRCh38
rs771988226 Chr.19: 3959590-3959590 on Build GRCh38
rs753522344 Chr.19: 3959398-3959398 on Build GRCh38
rs546274811 Chr.19: 3959378-3959378 on Build GRCh38
rs138075090 Chr.19: 3959353-3959353 on Build GRCh38
rs754551409 Chr.19: 3959222-3959222 on Build GRCh38
rs777223988 Chr.19: 3959396-3959396 on Build GRCh38
rs758239056 Chr.19: 3959206-3959206 on Build GRCh38
rs563048257 Chr.19: 3959273-3959273 on Build GRCh38
rs763032380 Chr.19: 3959321-3959321 on Build GRCh38
rs771292370 Chr.19: 3959245-3959245 on Build GRCh38
rs774246128 Chr.19: 3959451-3959451 on Build GRCh38
rs763645506 Chr.19: 3959468-3959468 on Build GRCh38

View Full Product Details