Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100192934-100193449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
UniGene
Hs.709187
Gene Chromosome Location
Chr.1: 100186922-100249864 on Build GRCh38
Transcripts
NM_001918.3

SNP Detail

SNP ID Chromosome Location
rs935458434 Chr.1: 100193366-100193366 on Build GRCh38
rs886044952 Chr.1: 100193068-100193068 on Build GRCh38
rs982399085 Chr.1: 100193183-100193183 on Build GRCh38
rs886044954 Chr.1: 100193228-100193228 on Build GRCh38
rs909584791 Chr.1: 100193200-100193200 on Build GRCh38
rs1009944021 Chr.1: 100193391-100193391 on Build GRCh38
rs752971591 Chr.1: 100193085-100193085 on Build GRCh38
rs907984674 Chr.1: 100193275-100193275 on Build GRCh38
rs182210964 Chr.1: 100193100-100193100 on Build GRCh38
rs923269758 Chr.1: 100193294-100193294 on Build GRCh38
rs902228621 Chr.1: 100193068-100193068 on Build GRCh38
rs969923994 Chr.1: 100193075-100193075 on Build GRCh38
rs1052427880 Chr.1: 100193385-100193385 on Build GRCh38
rs891586098 Chr.1: 100193356-100193356 on Build GRCh38
rs904028975 Chr.1: 100192978-100192978 on Build GRCh38
rs999909646 Chr.1: 100192956-100192956 on Build GRCh38
rs1034241013 Chr.1: 100193192-100193192 on Build GRCh38
rs904081965 Chr.1: 100193326-100193326 on Build GRCh38
rs1052460576 Chr.1: 100192966-100192966 on Build GRCh38
rs1023974357 Chr.1: 100193404-100193404 on Build GRCh38
rs1050661116 Chr.1: 100192959-100192959 on Build GRCh38
rs1051133771 Chr.1: 100193322-100193322 on Build GRCh38
rs886044953 Chr.1: 100193120-100193120 on Build GRCh38
rs1035155094 Chr.1: 100193159-100193159 on Build GRCh38
rs1011197393 Chr.1: 100192996-100192996 on Build GRCh38
rs951988309 Chr.1: 100193268-100193268 on Build GRCh38
rs548212429 Chr.1: 100193132-100193132 on Build GRCh38
rs921197858 Chr.1: 100193285-100193286 on Build GRCh38
rs891528882 Chr.1: 100193071-100193071 on Build GRCh38
rs894050029 Chr.1: 100192990-100192990 on Build GRCh38
rs2784174 Chr.1: 100193022-100193022 on Build GRCh38
rs984601866 Chr.1: 100193271-100193271 on Build GRCh38
rs971047446 Chr.1: 100193147-100193147 on Build GRCh38
rs528597114 Chr.1: 100193093-100193093 on Build GRCh38
rs933789529 Chr.1: 100193291-100193291 on Build GRCh38
rs951184118 Chr.1: 100193166-100193166 on Build GRCh38
rs893851234 Chr.1: 100193386-100193386 on Build GRCh38
rs936865959 Chr.1: 100193332-100193332 on Build GRCh38
rs149526077 Chr.1: 100193278-100193278 on Build GRCh38
rs758747432 Chr.1: 100193190-100193190 on Build GRCh38
rs1032742499 Chr.1: 100193045-100193045 on Build GRCh38
rs185645415 Chr.1: 100193303-100193303 on Build GRCh38
rs567851497 Chr.1: 100193259-100193259 on Build GRCh38
rs1052540127 Chr.1: 100193340-100193340 on Build GRCh38
rs549247028 Chr.1: 100193286-100193286 on Build GRCh38
rs906876017 Chr.1: 100193412-100193412 on Build GRCh38
rs886044951 Chr.1: 100193054-100193054 on Build GRCh38
rs962597737 Chr.1: 100193266-100193273 on Build GRCh38

View Full Product Details