Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100194772-100195268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
UniGene
Hs.709187
Gene Chromosome Location
Chr.1: 100186922-100249864 on Build GRCh38
Transcripts
NM_001918.3 XM_005270545.3 XM_017000468.1 XM_017000469.1

SNP Detail

SNP ID Chromosome Location
rs1009607918 Chr.1: 100195021-100195021 on Build GRCh38
rs570360415 Chr.1: 100195160-100195160 on Build GRCh38
rs969842468 Chr.1: 100194889-100194889 on Build GRCh38
rs528151186 Chr.1: 100194975-100194975 on Build GRCh38
rs191584096 Chr.1: 100194923-100194923 on Build GRCh38
rs112470859 Chr.1: 100194909-100194909 on Build GRCh38
rs1020906663 Chr.1: 100194836-100194836 on Build GRCh38
rs765873871 Chr.1: 100194912-100194912 on Build GRCh38
rs751799908 Chr.1: 100195103-100195103 on Build GRCh38
rs559530772 Chr.1: 100194974-100194974 on Build GRCh38
rs982395561 Chr.1: 100194867-100194867 on Build GRCh38
rs1013664323 Chr.1: 100195121-100195121 on Build GRCh38
rs1020963681 Chr.1: 100195077-100195077 on Build GRCh38
rs886044959 Chr.1: 100195190-100195191 on Build GRCh38
rs908582625 Chr.1: 100194932-100194932 on Build GRCh38
rs941752024 Chr.1: 100194933-100194933 on Build GRCh38
rs764457050 Chr.1: 100195031-100195031 on Build GRCh38
rs1003932483 Chr.1: 100195149-100195149 on Build GRCh38
rs918933820 Chr.1: 100194961-100194961 on Build GRCh38
rs144220855 Chr.1: 100195014-100195014 on Build GRCh38
rs932951938 Chr.1: 100194967-100194967 on Build GRCh38
rs553113077 Chr.1: 100195191-100195191 on Build GRCh38
rs947041211 Chr.1: 100195057-100195057 on Build GRCh38
rs971098649 Chr.1: 100194845-100194845 on Build GRCh38
rs548286585 Chr.1: 100195004-100195004 on Build GRCh38
rs974257403 Chr.1: 100194948-100194948 on Build GRCh38
rs758699159 Chr.1: 100195016-100195016 on Build GRCh38
rs961325946 Chr.1: 100195187-100195187 on Build GRCh38
rs959877704 Chr.1: 100195177-100195177 on Build GRCh38
rs974058127 Chr.1: 100195206-100195206 on Build GRCh38
rs187360848 Chr.1: 100195244-100195244 on Build GRCh38
rs971762953 Chr.1: 100194958-100194958 on Build GRCh38
rs1051703335 Chr.1: 100194970-100194970 on Build GRCh38
rs183934265 Chr.1: 100194926-100194926 on Build GRCh38
rs1053540812 Chr.1: 100194999-100194999 on Build GRCh38
rs532897370 Chr.1: 100195222-100195222 on Build GRCh38
rs1033961722 Chr.1: 100195173-100195173 on Build GRCh38
rs550450511 Chr.1: 100195114-100195114 on Build GRCh38
rs1001357197 Chr.1: 100195109-100195109 on Build GRCh38
rs776256682 Chr.1: 100194874-100194874 on Build GRCh38
rs530528870 Chr.1: 100195113-100195113 on Build GRCh38
rs1001785444 Chr.1: 100194904-100194904 on Build GRCh38

View Full Product Details