Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41984523-41985030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF4L1
Entrez Gene ID
285429
Gene Name
DDB1 and CUL4 associated factor 4 like 1
Gene Aliases
WDR21B
UniGene
Hs.213307
Gene Chromosome Location
Chr.4: 41981696-41986467 on Build GRCh38
Transcripts
NM_001029955.3

SNP Detail

SNP ID Chromosome Location
rs1036001632 Chr.4: 41984563-41984563 on Build GRCh38
rs936649788 Chr.4: 41984709-41984709 on Build GRCh38
rs1014147200 Chr.4: 41984994-41984994 on Build GRCh38
rs1038744102 Chr.4: 41984853-41984853 on Build GRCh38
rs182745422 Chr.4: 41984774-41984774 on Build GRCh38
rs956684274 Chr.4: 41984768-41984768 on Build GRCh38
rs551794000 Chr.4: 41984833-41984833 on Build GRCh38
rs16853848 Chr.4: 41984775-41984775 on Build GRCh38
rs894262760 Chr.4: 41984619-41984619 on Build GRCh38
rs531895663 Chr.4: 41984688-41984688 on Build GRCh38
rs950614197 Chr.4: 41984595-41984595 on Build GRCh38
rs747692778 Chr.4: 41984568-41984568 on Build GRCh38
rs1026646998 Chr.4: 41984583-41984583 on Build GRCh38
rs78170005 Chr.4: 41984567-41984567 on Build GRCh38
rs993078342 Chr.4: 41984603-41984603 on Build GRCh38
rs1022304605 Chr.4: 41984777-41984777 on Build GRCh38
rs898877090 Chr.4: 41984867-41984867 on Build GRCh38
rs953492468 Chr.4: 41984822-41984822 on Build GRCh38
rs567628561 Chr.4: 41984776-41984776 on Build GRCh38
rs144441211 Chr.4: 41984938-41984938 on Build GRCh38
rs971957389 Chr.4: 41984865-41984865 on Build GRCh38
rs773088170 Chr.4: 41984841-41984844 on Build GRCh38
rs576165153 Chr.4: 41984897-41984897 on Build GRCh38
rs149814888 Chr.4: 41984969-41984972 on Build GRCh38
rs986347518 Chr.4: 41984837-41984837 on Build GRCh38
rs953328402 Chr.4: 41984548-41984548 on Build GRCh38
rs556611082 Chr.4: 41984843-41984843 on Build GRCh38
rs970819562 Chr.4: 41984619-41984620 on Build GRCh38
rs562982859 Chr.4: 41984653-41984653 on Build GRCh38
rs758885140 Chr.4: 41984575-41984575 on Build GRCh38
rs985991797 Chr.4: 41984558-41984558 on Build GRCh38
rs1012794134 Chr.4: 41984621-41984621 on Build GRCh38
rs890596661 Chr.4: 41984829-41984829 on Build GRCh38
rs764604149 Chr.4: 41984647-41984647 on Build GRCh38
rs571942983 Chr.4: 41984760-41984760 on Build GRCh38
rs906753459 Chr.4: 41984735-41984735 on Build GRCh38
rs917049980 Chr.4: 41984612-41984612 on Build GRCh38
rs746400814 Chr.4: 41984973-41984973 on Build GRCh38
rs191229779 Chr.4: 41984743-41984743 on Build GRCh38
rs915605660 Chr.4: 41985001-41985001 on Build GRCh38
rs911760646 Chr.4: 41984561-41984561 on Build GRCh38
rs1054080074 Chr.4: 41984750-41984750 on Build GRCh38
rs540593442 Chr.4: 41984545-41984545 on Build GRCh38
rs1003810658 Chr.4: 41984739-41984739 on Build GRCh38
rs560662961 Chr.4: 41984553-41984553 on Build GRCh38
rs4410557 Chr.4: 41984900-41984900 on Build GRCh38
rs771294490 Chr.4: 41984683-41984683 on Build GRCh38
rs1048839361 Chr.4: 41984576-41984576 on Build GRCh38
rs984462781 Chr.4: 41984998-41984998 on Build GRCh38
rs994904213 Chr.4: 41984920-41984924 on Build GRCh38
rs2581439 Chr.4: 41984840-41984840 on Build GRCh38
rs549848516 Chr.4: 41984638-41984638 on Build GRCh38
rs944659852 Chr.4: 41984562-41984562 on Build GRCh38

View Full Product Details