Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18920215-18920711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX49
Entrez Gene ID
54555
Gene Name
DEAD-box helicase 49
Gene Aliases
R27090_2
UniGene
Hs.143187
Gene Chromosome Location
Chr.19: 18919675-18928633 on Build GRCh38
Transcripts
NM_019070.4 NR_033677.1 XM_011528083.1 XM_011528084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993186 c.164C>T p.A55V Chr.19: 18920628-18920628 on Build GRCh38
5389145 c.203C>T p.P68L Chr.19: 18920667-18920667 on Build GRCh38
3100621 c.189G>A p.K63K Chr.19: 18920653-18920653 on Build GRCh38
5013402 c.116-1G>T p.? Chr.19: 18920579-18920579 on Build GRCh38
6579957 c.196G>A p.E66K Chr.19: 18920660-18920660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148771932 Chr.19: 18920646-18920646 on Build GRCh38
rs758914473 Chr.19: 18920550-18920550 on Build GRCh38
rs775712338 Chr.19: 18920582-18920582 on Build GRCh38
rs139365572 Chr.19: 18920365-18920365 on Build GRCh38
rs757589847 Chr.19: 18920670-18920670 on Build GRCh38
rs994953441 Chr.19: 18920406-18920406 on Build GRCh38
rs759200566 Chr.19: 18920633-18920633 on Build GRCh38
rs1023957279 Chr.19: 18920567-18920567 on Build GRCh38
rs887242748 Chr.19: 18920385-18920385 on Build GRCh38
rs757400865 Chr.19: 18920572-18920572 on Build GRCh38
rs750603622 Chr.19: 18920687-18920687 on Build GRCh38
rs760556288 Chr.19: 18920615-18920615 on Build GRCh38
rs369608763 Chr.19: 18920629-18920629 on Build GRCh38
rs748093851 Chr.19: 18920594-18920594 on Build GRCh38
rs1053655769 Chr.19: 18920342-18920342 on Build GRCh38
rs776218735 Chr.19: 18920631-18920631 on Build GRCh38
rs901460322 Chr.19: 18920332-18920332 on Build GRCh38
rs915248864 Chr.19: 18920310-18920310 on Build GRCh38
rs758380147 Chr.19: 18920497-18920497 on Build GRCh38
rs558074299 Chr.19: 18920237-18920237 on Build GRCh38
rs763359848 Chr.19: 18920641-18920641 on Build GRCh38
rs75693456 Chr.19: 18920392-18920392 on Build GRCh38
rs764713644 Chr.19: 18920644-18920644 on Build GRCh38
rs781434295 Chr.19: 18920575-18920575 on Build GRCh38
rs1044432777 Chr.19: 18920400-18920400 on Build GRCh38
rs946488072 Chr.19: 18920288-18920288 on Build GRCh38
rs905842239 Chr.19: 18920402-18920402 on Build GRCh38
rs754956564 Chr.19: 18920337-18920337 on Build GRCh38
rs759169492 Chr.19: 18920602-18920602 on Build GRCh38
rs142390134 Chr.19: 18920686-18920686 on Build GRCh38
rs145421176 Chr.19: 18920529-18920529 on Build GRCh38
rs376039187 Chr.19: 18920312-18920312 on Build GRCh38
rs748059078 Chr.19: 18920429-18920429 on Build GRCh38
rs935431619 Chr.19: 18920258-18920258 on Build GRCh38
rs778933813 Chr.19: 18920348-18920348 on Build GRCh38
rs950650032 Chr.19: 18920451-18920451 on Build GRCh38
rs34332105 Chr.19: 18920635-18920635 on Build GRCh38
rs1005323068 Chr.19: 18920404-18920404 on Build GRCh38
rs950301399 Chr.19: 18920645-18920645 on Build GRCh38
rs746112583 Chr.19: 18920578-18920578 on Build GRCh38
rs186669281 Chr.19: 18920420-18920420 on Build GRCh38
rs375276251 Chr.19: 18920531-18920531 on Build GRCh38
rs573301432 Chr.19: 18920363-18920363 on Build GRCh38
rs772104304 Chr.19: 18920598-18920598 on Build GRCh38
rs2231994 Chr.19: 18920457-18920457 on Build GRCh38
rs531275843 Chr.19: 18920549-18920549 on Build GRCh38
rs116507095 Chr.19: 18920636-18920636 on Build GRCh38
rs977282335 Chr.19: 18920596-18920596 on Build GRCh38
rs769933093 Chr.19: 18920579-18920579 on Build GRCh38
rs1000245 Chr.19: 18920278-18920278 on Build GRCh38
rs1019160884 Chr.19: 18920480-18920480 on Build GRCh38
rs868122514 Chr.19: 18920282-18920282 on Build GRCh38
rs1039698916 Chr.19: 18920330-18920330 on Build GRCh38
rs147735065 Chr.19: 18920639-18920639 on Build GRCh38
rs746789708 Chr.19: 18920576-18920576 on Build GRCh38
rs772859126 Chr.19: 18920606-18920606 on Build GRCh38
rs181710631 Chr.19: 18920338-18920338 on Build GRCh38
rs747468651 Chr.19: 18920565-18920565 on Build GRCh38
rs369580612 Chr.19: 18920440-18920440 on Build GRCh38

View Full Product Details