Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33025896-33026409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC7
Entrez Gene ID
91614
Gene Name
DEP domain containing 7
Gene Aliases
TR2, dJ85M6.4
UniGene
Hs.280990
Gene Chromosome Location
Chr.11: 33015864-33033582 on Build GRCh38
Transcripts
NM_001077242.1 NM_139160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889573 c.365C>G p.P122R Chr.11: 33025950-33025950 on Build GRCh38
2114816 c.437+2T>C p.? Chr.11: 33026051-33026051 on Build GRCh38
5799868 c.320T>G p.F107C Chr.11: 33025932-33025932 on Build GRCh38
136827 c.349G>A p.D117N Chr.11: 33025961-33025961 on Build GRCh38
6537296 c.355A>G p.S119G Chr.11: 33025967-33025967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769462869 Chr.11: 33026151-33026151 on Build GRCh38
rs561100082 Chr.11: 33026149-33026149 on Build GRCh38
rs781700415 Chr.11: 33025949-33025949 on Build GRCh38
rs754187737 Chr.11: 33025940-33025940 on Build GRCh38
rs957023667 Chr.11: 33026155-33026155 on Build GRCh38
rs773955423 Chr.11: 33026018-33026018 on Build GRCh38
rs550128153 Chr.11: 33026192-33026192 on Build GRCh38
rs749467283 Chr.11: 33026013-33026016 on Build GRCh38
rs530218104 Chr.11: 33026160-33026160 on Build GRCh38
rs756019898 Chr.11: 33026117-33026117 on Build GRCh38
rs750559110 Chr.11: 33025960-33025960 on Build GRCh38
rs1011384063 Chr.11: 33026323-33026323 on Build GRCh38
rs903078675 Chr.11: 33026382-33026382 on Build GRCh38
rs866171135 Chr.11: 33025989-33025989 on Build GRCh38
rs888419252 Chr.11: 33025934-33025934 on Build GRCh38
rs930059338 Chr.11: 33026317-33026317 on Build GRCh38
rs765593581 Chr.11: 33026069-33026069 on Build GRCh38
rs764361404 Chr.11: 33025931-33025931 on Build GRCh38
rs764600089 Chr.11: 33026062-33026062 on Build GRCh38
rs796274095 Chr.11: 33026378-33026382 on Build GRCh38
rs977086642 Chr.11: 33025948-33025948 on Build GRCh38
rs528768783 Chr.11: 33026170-33026170 on Build GRCh38
rs750740480 Chr.11: 33025997-33025997 on Build GRCh38
rs556333291 Chr.11: 33025978-33025978 on Build GRCh38
rs950277361 Chr.11: 33026373-33026375 on Build GRCh38
rs771872138 Chr.11: 33025972-33025972 on Build GRCh38
rs202173561 Chr.11: 33025975-33025975 on Build GRCh38
rs183105931 Chr.11: 33026335-33026335 on Build GRCh38
rs769070608 Chr.11: 33026180-33026180 on Build GRCh38
rs757731519 Chr.11: 33025942-33025943 on Build GRCh38
rs749113884 Chr.11: 33026123-33026123 on Build GRCh38
rs771468155 Chr.11: 33026042-33026042 on Build GRCh38
rs370122271 Chr.11: 33026075-33026075 on Build GRCh38
rs770684464 Chr.11: 33026133-33026133 on Build GRCh38
rs894753189 Chr.11: 33026364-33026364 on Build GRCh38
rs998609921 Chr.11: 33026108-33026108 on Build GRCh38
rs779997442 Chr.11: 33026078-33026078 on Build GRCh38
rs888957298 Chr.11: 33026296-33026296 on Build GRCh38
rs919089274 Chr.11: 33026286-33026286 on Build GRCh38
rs991189900 Chr.11: 33026024-33026024 on Build GRCh38
rs776339757 Chr.11: 33026176-33026176 on Build GRCh38
rs777244672 Chr.11: 33026195-33026195 on Build GRCh38
rs756567724 Chr.11: 33026008-33026019 on Build GRCh38
rs780285915 Chr.11: 33026008-33026011 on Build GRCh38
rs781243212 Chr.11: 33025968-33025968 on Build GRCh38
rs374941692 Chr.11: 33026076-33026076 on Build GRCh38
rs200889394 Chr.11: 33026065-33026065 on Build GRCh38
rs374632275 Chr.11: 33026036-33026036 on Build GRCh38
rs142562030 Chr.11: 33026058-33026058 on Build GRCh38
rs760593713 Chr.11: 33026144-33026144 on Build GRCh38
rs933427582 Chr.11: 33026203-33026203 on Build GRCh38
rs541433489 Chr.11: 33026142-33026142 on Build GRCh38
rs568264163 Chr.11: 33026306-33026314 on Build GRCh38
rs975924855 Chr.11: 33026238-33026238 on Build GRCh38
rs531309178 Chr.11: 33026045-33026045 on Build GRCh38
rs754273487 Chr.11: 33026063-33026063 on Build GRCh38
rs376814687 Chr.11: 33025956-33025956 on Build GRCh38
rs576295157 Chr.11: 33026032-33026032 on Build GRCh38
rs879708135 Chr.11: 33026090-33026090 on Build GRCh38
rs1050499113 Chr.11: 33026225-33026225 on Build GRCh38
rs573702016 Chr.11: 33025944-33025944 on Build GRCh38
rs770594618 Chr.11: 33026017-33026017 on Build GRCh38
rs749025119 Chr.11: 33025984-33025984 on Build GRCh38
rs371496330 Chr.11: 33025965-33025965 on Build GRCh38
rs1055792174 Chr.11: 33026336-33026336 on Build GRCh38
rs751595199 Chr.11: 33026104-33026104 on Build GRCh38
rs777639817 Chr.11: 33026120-33026120 on Build GRCh38
rs377735139 Chr.11: 33026052-33026052 on Build GRCh38
rs748025899 Chr.11: 33025974-33025974 on Build GRCh38
rs982846776 Chr.11: 33026318-33026318 on Build GRCh38

View Full Product Details