Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23835959-23836464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL3
Entrez Gene ID
91319
Gene Name
derlin 3
Gene Aliases
C22orf14, IZP6, LLN2, derlin-3
UniGene
Hs.593679
Gene Chromosome Location
Chr.22: 23834503-23839344 on Build GRCh38
Transcripts
NM_001002862.2 NM_001135751.1 NM_198440.3 XM_011530506.2 XM_017029082.1

SNP Detail

SNP ID Chromosome Location
rs527714153 Chr.22: 23836445-23836445 on Build GRCh38
rs35483188 Chr.22: 23836009-23836009 on Build GRCh38
rs948927452 Chr.22: 23836442-23836442 on Build GRCh38
rs759500700 Chr.22: 23836071-23836071 on Build GRCh38
rs888820824 Chr.22: 23836285-23836285 on Build GRCh38
rs1044670261 Chr.22: 23836088-23836088 on Build GRCh38
rs986048490 Chr.22: 23836294-23836294 on Build GRCh38
rs1026120843 Chr.22: 23836358-23836358 on Build GRCh38
rs775077193 Chr.22: 23836293-23836293 on Build GRCh38
rs893307660 Chr.22: 23836096-23836096 on Build GRCh38
rs1046119617 Chr.22: 23836021-23836021 on Build GRCh38
rs776420357 Chr.22: 23836207-23836207 on Build GRCh38
rs993369752 Chr.22: 23836431-23836431 on Build GRCh38
rs1011713932 Chr.22: 23836108-23836108 on Build GRCh38
rs776528048 Chr.22: 23836035-23836035 on Build GRCh38
rs929444834 Chr.22: 23836013-23836013 on Build GRCh38
rs1030179907 Chr.22: 23836222-23836222 on Build GRCh38
rs981581108 Chr.22: 23836419-23836419 on Build GRCh38
rs1041917315 Chr.22: 23836131-23836131 on Build GRCh38
rs565748402 Chr.22: 23836075-23836075 on Build GRCh38
rs948979653 Chr.22: 23836080-23836080 on Build GRCh38
rs928678820 Chr.22: 23836078-23836078 on Build GRCh38
rs566893269 Chr.22: 23836318-23836318 on Build GRCh38
rs5760061 Chr.22: 23836092-23836092 on Build GRCh38
rs547170405 Chr.22: 23836209-23836209 on Build GRCh38
rs892240817 Chr.22: 23836133-23836133 on Build GRCh38
rs551310409 Chr.22: 23836091-23836091 on Build GRCh38
rs34564314 Chr.22: 23836119-23836119 on Build GRCh38
rs5760062 Chr.22: 23836403-23836403 on Build GRCh38
rs973424494 Chr.22: 23836344-23836344 on Build GRCh38
rs1017573746 Chr.22: 23836297-23836297 on Build GRCh38
rs35961970 Chr.22: 23836352-23836352 on Build GRCh38
rs929419883 Chr.22: 23836426-23836426 on Build GRCh38
rs879218692 Chr.22: 23836253-23836253 on Build GRCh38
rs531304611 Chr.22: 23836027-23836027 on Build GRCh38
rs971840258 Chr.22: 23836353-23836353 on Build GRCh38
rs536450679 Chr.22: 23836099-23836099 on Build GRCh38
rs963271764 Chr.22: 23836348-23836348 on Build GRCh38
rs954752977 Chr.22: 23836261-23836261 on Build GRCh38
rs902907022 Chr.22: 23836192-23836192 on Build GRCh38
rs919377255 Chr.22: 23836010-23836010 on Build GRCh38
rs896027769 Chr.22: 23836057-23836057 on Build GRCh38
rs1009793681 Chr.22: 23836141-23836141 on Build GRCh38
rs917424894 Chr.22: 23836440-23836440 on Build GRCh38
rs904864288 Chr.22: 23836089-23836089 on Build GRCh38

View Full Product Details