Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19132533-19133038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
UniGene
Hs.517407
Gene Chromosome Location
Chr.22: 19130279-19144726 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Gene Symbol
TSSK2
Entrez Gene ID
23617
Gene Name
testis specific serine kinase 2
Gene Aliases
DGS-G, SPOGA2, STK22B, TSK2
UniGene
Hs.694070
Gene Chromosome Location
Chr.22: 19130808-19132623 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

SNP Detail

SNP ID Chromosome Location
rs528388321 Chr.22: 19132776-19132776 on Build GRCh38
rs928101548 Chr.22: 19132629-19132629 on Build GRCh38
rs368842661 Chr.22: 19132573-19132573 on Build GRCh38
rs936289352 Chr.22: 19132835-19132835 on Build GRCh38
rs144849879 Chr.22: 19132750-19132750 on Build GRCh38
rs937994053 Chr.22: 19132647-19132647 on Build GRCh38
rs973312138 Chr.22: 19132817-19132818 on Build GRCh38
rs1016657564 Chr.22: 19133013-19133013 on Build GRCh38
rs1003082166 Chr.22: 19132975-19132975 on Build GRCh38
rs560231390 Chr.22: 19132906-19132906 on Build GRCh38
rs1026488807 Chr.22: 19132981-19132981 on Build GRCh38
rs148153676 Chr.22: 19132756-19132756 on Build GRCh38
rs902185654 Chr.22: 19132783-19132783 on Build GRCh38
rs938110654 Chr.22: 19132885-19132885 on Build GRCh38
rs894871963 Chr.22: 19132877-19132877 on Build GRCh38
rs748542953 Chr.22: 19132598-19132598 on Build GRCh38
rs531089437 Chr.22: 19132557-19132557 on Build GRCh38
rs572425503 Chr.22: 19132946-19132946 on Build GRCh38
rs910447956 Chr.22: 19132564-19132564 on Build GRCh38
rs950679565 Chr.22: 19132678-19132678 on Build GRCh38
rs1007201326 Chr.22: 19132799-19132799 on Build GRCh38
rs1013056839 Chr.22: 19132608-19132608 on Build GRCh38
rs973197084 Chr.22: 19132595-19132595 on Build GRCh38
rs928017723 Chr.22: 19132872-19132872 on Build GRCh38
rs551345502 Chr.22: 19132914-19132914 on Build GRCh38
rs902787469 Chr.22: 19133006-19133006 on Build GRCh38
rs923086662 Chr.22: 19132757-19132757 on Build GRCh38
rs894032490 Chr.22: 19132723-19132723 on Build GRCh38
rs8141243 Chr.22: 19133011-19133011 on Build GRCh38
rs933260862 Chr.22: 19132763-19132763 on Build GRCh38
rs1023657467 Chr.22: 19132614-19132614 on Build GRCh38
rs1055069269 Chr.22: 19132694-19132694 on Build GRCh38
rs756699267 Chr.22: 19132624-19132624 on Build GRCh38
rs1026311463 Chr.22: 19132651-19132651 on Build GRCh38
rs372479336 Chr.22: 19132597-19132597 on Build GRCh38
rs571198298 Chr.22: 19133005-19133005 on Build GRCh38
rs1042328210 Chr.22: 19132779-19132779 on Build GRCh38
rs749643455 Chr.22: 19132995-19132995 on Build GRCh38
rs981650897 Chr.22: 19132622-19132622 on Build GRCh38
rs984846751 Chr.22: 19132685-19132685 on Build GRCh38
rs907378054 Chr.22: 19132970-19132970 on Build GRCh38
rs1012391208 Chr.22: 19132913-19132913 on Build GRCh38
rs971701457 Chr.22: 19132621-19132621 on Build GRCh38
rs778253800 Chr.22: 19132748-19132748 on Build GRCh38
rs774675457 Chr.22: 19133007-19133007 on Build GRCh38
rs1053960225 Chr.22: 19132865-19132865 on Build GRCh38
rs749722866 Chr.22: 19132849-19132849 on Build GRCh38
rs565507174 Chr.22: 19132759-19132759 on Build GRCh38
rs563420605 Chr.22: 19132749-19132749 on Build GRCh38
rs1032052420 Chr.22: 19132563-19132563 on Build GRCh38
rs919216851 Chr.22: 19132838-19132838 on Build GRCh38
rs1017286965 Chr.22: 19132801-19132801 on Build GRCh38
rs1042605252 Chr.22: 19132993-19132993 on Build GRCh38
rs544813621 Chr.22: 19132658-19132658 on Build GRCh38
rs941936934 Chr.22: 19132588-19132588 on Build GRCh38
rs963249842 Chr.22: 19132813-19132813 on Build GRCh38
rs535028759 Chr.22: 19132992-19132992 on Build GRCh38
rs981993435 Chr.22: 19132856-19132856 on Build GRCh38
rs998392491 Chr.22: 19132768-19132768 on Build GRCh38

View Full Product Details