Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 958781-959277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKQ
Entrez Gene ID
1609
Gene Name
diacylglycerol kinase theta
Gene Aliases
DAGK, DAGK4, DAGK7
UniGene
Hs.584858
Gene Chromosome Location
Chr.4: 958884-973560 on Build GRCh38
Transcripts
NM_001347.3 XM_011513411.1

SNP Detail

SNP ID Chromosome Location
rs1056331783 Chr.4: 959005-959005 on Build GRCh38
rs1012732897 Chr.4: 958902-958902 on Build GRCh38
rs780301846 Chr.4: 959202-959202 on Build GRCh38
rs563239851 Chr.4: 959127-959128 on Build GRCh38
rs920021448 Chr.4: 958968-958968 on Build GRCh38
rs974028498 Chr.4: 959161-959161 on Build GRCh38
rs138854884 Chr.4: 959040-959040 on Build GRCh38
rs961397074 Chr.4: 959159-959159 on Build GRCh38
rs112332402 Chr.4: 958884-958884 on Build GRCh38
rs984805176 Chr.4: 958967-958967 on Build GRCh38
rs981298288 Chr.4: 958936-958936 on Build GRCh38
rs919371387 Chr.4: 958801-958801 on Build GRCh38
rs111544975 Chr.4: 959005-959008 on Build GRCh38
rs1027515155 Chr.4: 959164-959164 on Build GRCh38
rs1044771299 Chr.4: 958987-958987 on Build GRCh38
rs764057442 Chr.4: 958993-958993 on Build GRCh38
rs1002657131 Chr.4: 959251-959251 on Build GRCh38
rs117925245 Chr.4: 959029-959029 on Build GRCh38
rs374498286 Chr.4: 959153-959153 on Build GRCh38
rs1038698936 Chr.4: 959200-959200 on Build GRCh38
rs781716597 Chr.4: 959233-959233 on Build GRCh38
rs1050471803 Chr.4: 958845-958845 on Build GRCh38
rs929394892 Chr.4: 958948-958948 on Build GRCh38
rs978375306 Chr.4: 959197-959197 on Build GRCh38
rs891362209 Chr.4: 958849-958849 on Build GRCh38
rs974160711 Chr.4: 958963-958963 on Build GRCh38
rs1022815489 Chr.4: 959100-959100 on Build GRCh38
rs902827208 Chr.4: 958873-958873 on Build GRCh38
rs550045681 Chr.4: 959199-959199 on Build GRCh38
rs971352769 Chr.4: 958932-958932 on Build GRCh38
rs941282323 Chr.4: 958838-958838 on Build GRCh38
rs913812829 Chr.4: 959152-959152 on Build GRCh38
rs528608185 Chr.4: 959035-959035 on Build GRCh38
rs765443625 Chr.4: 959118-959118 on Build GRCh38
rs865836427 Chr.4: 959044-959044 on Build GRCh38
rs892769859 Chr.4: 959213-959213 on Build GRCh38
rs929363626 Chr.4: 958821-958821 on Build GRCh38
rs1038375977 Chr.4: 958802-958802 on Build GRCh38
rs60461049 Chr.4: 959072-959072 on Build GRCh38
rs528908648 Chr.4: 959049-959049 on Build GRCh38
rs980141453 Chr.4: 959132-959132 on Build GRCh38
rs753622445 Chr.4: 959039-959039 on Build GRCh38
rs542403269 Chr.4: 959165-959165 on Build GRCh38
rs951241768 Chr.4: 958829-958829 on Build GRCh38
rs961324275 Chr.4: 958947-958947 on Build GRCh38
rs115657987 Chr.4: 959166-959166 on Build GRCh38
rs189158494 Chr.4: 959012-959012 on Build GRCh38
rs957467812 Chr.4: 958901-958901 on Build GRCh38
rs909320451 Chr.4: 959106-959106 on Build GRCh38
rs903599164 Chr.4: 959047-959047 on Build GRCh38
rs912734097 Chr.4: 959191-959191 on Build GRCh38
rs757057929 Chr.4: 959080-959080 on Build GRCh38
rs940124939 Chr.4: 958800-958800 on Build GRCh38
rs552535616 Chr.4: 959136-959136 on Build GRCh38
rs535481791 Chr.4: 959230-959230 on Build GRCh38
rs944290025 Chr.4: 959055-959055 on Build GRCh38
rs146299481 Chr.4: 959075-959075 on Build GRCh38
rs372876115 Chr.4: 958855-958855 on Build GRCh38
rs1009329236 Chr.4: 959024-959024 on Build GRCh38
rs1015455853 Chr.4: 959137-959137 on Build GRCh38
rs892267347 Chr.4: 959019-959019 on Build GRCh38
rs747055356 Chr.4: 959203-959203 on Build GRCh38
rs774935686 Chr.4: 958943-958943 on Build GRCh38
rs73064477 Chr.4: 958856-958856 on Build GRCh38
rs370920706 Chr.4: 959120-959120 on Build GRCh38
rs1008598294 Chr.4: 958853-958853 on Build GRCh38
rs1044502569 Chr.4: 959222-959222 on Build GRCh38
rs184400698 Chr.4: 958916-958916 on Build GRCh38
rs545836419 Chr.4: 959121-959121 on Build GRCh38
rs944154490 Chr.4: 959192-959192 on Build GRCh38
rs998515441 Chr.4: 959073-959073 on Build GRCh38
rs1042624154 Chr.4: 959069-959069 on Build GRCh38
rs574472129 Chr.4: 958823-958823 on Build GRCh38
rs531204056 Chr.4: 959214-959214 on Build GRCh38
rs1021972493 Chr.4: 958887-958887 on Build GRCh38
rs535587961 Chr.4: 958824-958824 on Build GRCh38
rs1007251162 Chr.4: 959048-959048 on Build GRCh38
rs948786241 Chr.4: 958986-958986 on Build GRCh38
rs1056363041 Chr.4: 959150-959150 on Build GRCh38
rs966682141 Chr.4: 958928-958928 on Build GRCh38
rs760633370 Chr.4: 958964-958964 on Build GRCh38
rs529817686 Chr.4: 959162-959162 on Build GRCh38
rs191616491 Chr.4: 959224-959224 on Build GRCh38
rs75456209 Chr.4: 958899-958899 on Build GRCh38
rs950916914 Chr.4: 959078-959078 on Build GRCh38
rs1025325670 Chr.4: 958822-958822 on Build GRCh38
rs1054636241 Chr.4: 959074-959074 on Build GRCh38
rs1054140364 Chr.4: 959204-959204 on Build GRCh38
rs1003402727 Chr.4: 958870-958870 on Build GRCh38
rs866867968 Chr.4: 959250-959250 on Build GRCh38
rs950949037 Chr.4: 959237-959237 on Build GRCh38
rs1034840733 Chr.4: 958872-958872 on Build GRCh38

View Full Product Details