Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12677095-12677633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHPS
Entrez Gene ID
1725
Gene Name
deoxyhypusine synthase
Gene Aliases
DHS, DS, MIG13
UniGene
Hs.79064
Gene Chromosome Location
Chr.19: 12672164-12681887 on Build GRCh38
Transcripts
NM_001206974.1 NM_001930.3 NM_013406.2 NR_038192.1 XM_011527770.1 XM_011527771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982861 c.715G>T p.G239C Chr.19: 12677360-12677360 on Build GRCh38
438620 c.697A>C p.S233R Chr.19: 12677378-12677378 on Build GRCh38
6643294 c.696T>C p.F232F Chr.19: 12677379-12677379 on Build GRCh38
991400 c.824G>A p.C275Y Chr.19: 12677172-12677172 on Build GRCh38
4507461 c.746C>T p.S249F Chr.19: 12677329-12677329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764549648 Chr.19: 12677366-12677366 on Build GRCh38
rs761549753 Chr.19: 12677345-12677345 on Build GRCh38
rs765769748 Chr.19: 12677452-12677452 on Build GRCh38
rs189109985 Chr.19: 12677283-12677283 on Build GRCh38
rs772922784 Chr.19: 12677408-12677408 on Build GRCh38
rs376275007 Chr.19: 12677423-12677423 on Build GRCh38
rs758418180 Chr.19: 12677190-12677190 on Build GRCh38
rs931417522 Chr.19: 12677587-12677587 on Build GRCh38
rs978898685 Chr.19: 12677158-12677158 on Build GRCh38
rs767613292 Chr.19: 12677162-12677162 on Build GRCh38
rs1012142857 Chr.19: 12677387-12677387 on Build GRCh38
rs750117682 Chr.19: 12677436-12677438 on Build GRCh38
rs535857791 Chr.19: 12677360-12677360 on Build GRCh38
rs745985350 Chr.19: 12677228-12677228 on Build GRCh38
rs974776882 Chr.19: 12677252-12677252 on Build GRCh38
rs919971126 Chr.19: 12677344-12677344 on Build GRCh38
rs759035741 Chr.19: 12677439-12677439 on Build GRCh38
rs911917434 Chr.19: 12677464-12677464 on Build GRCh38
rs1021333751 Chr.19: 12677463-12677463 on Build GRCh38
rs746075617 Chr.19: 12677317-12677317 on Build GRCh38
rs773694823 Chr.19: 12677361-12677361 on Build GRCh38
rs780759776 Chr.19: 12677385-12677386 on Build GRCh38
rs954382476 Chr.19: 12677531-12677531 on Build GRCh38
rs760044789 Chr.19: 12677183-12677183 on Build GRCh38
rs912160587 Chr.19: 12677214-12677214 on Build GRCh38
rs975024793 Chr.19: 12677467-12677467 on Build GRCh38
rs751094326 Chr.19: 12677334-12677336 on Build GRCh38
rs938956428 Chr.19: 12677370-12677370 on Build GRCh38
rs758656990 Chr.19: 12677385-12677385 on Build GRCh38
rs1014820500 Chr.19: 12677538-12677538 on Build GRCh38
rs371375475 Chr.19: 12677262-12677262 on Build GRCh38
rs934680485 Chr.19: 12677178-12677178 on Build GRCh38
rs761265606 Chr.19: 12677365-12677365 on Build GRCh38
rs779347086 Chr.19: 12677316-12677316 on Build GRCh38
rs576946487 Chr.19: 12677540-12677540 on Build GRCh38
rs940339073 Chr.19: 12677596-12677596 on Build GRCh38
rs753137312 Chr.19: 12677216-12677216 on Build GRCh38
rs781054297 Chr.19: 12677394-12677394 on Build GRCh38
rs377182954 Chr.19: 12677268-12677268 on Build GRCh38
rs760127859 Chr.19: 12677575-12677575 on Build GRCh38
rs759569057 Chr.19: 12677151-12677151 on Build GRCh38
rs749455821 Chr.19: 12677146-12677146 on Build GRCh38
rs143117686 Chr.19: 12677355-12677355 on Build GRCh38
rs748558868 Chr.19: 12677139-12677139 on Build GRCh38
rs747003113 Chr.19: 12677337-12677337 on Build GRCh38
rs761309007 Chr.19: 12677187-12677187 on Build GRCh38
rs149307460 Chr.19: 12677115-12677115 on Build GRCh38
rs925940285 Chr.19: 12677175-12677175 on Build GRCh38
rs758888080 Chr.19: 12677473-12677473 on Build GRCh38
rs1028652737 Chr.19: 12677254-12677254 on Build GRCh38
rs181620939 Chr.19: 12677402-12677402 on Build GRCh38
rs550355399 Chr.19: 12677183-12677183 on Build GRCh38
rs776604750 Chr.19: 12677340-12677340 on Build GRCh38
rs763786266 Chr.19: 12677177-12677177 on Build GRCh38
rs754130948 Chr.19: 12677149-12677150 on Build GRCh38
rs776254007 Chr.19: 12677267-12677267 on Build GRCh38
rs75392871 Chr.19: 12677225-12677225 on Build GRCh38
rs960496806 Chr.19: 12677118-12677118 on Build GRCh38
rs767041900 Chr.19: 12677440-12677440 on Build GRCh38
rs754366240 Chr.19: 12677371-12677371 on Build GRCh38
rs757878296 Chr.19: 12677191-12677191 on Build GRCh38
rs774023747 Chr.19: 12677427-12677427 on Build GRCh38
rs1044659893 Chr.19: 12677127-12677127 on Build GRCh38
rs748062148 Chr.19: 12677397-12677397 on Build GRCh38
rs748969853 Chr.19: 12677415-12677415 on Build GRCh38
rs368781559 Chr.19: 12677131-12677131 on Build GRCh38
rs370385403 Chr.19: 12677272-12677272 on Build GRCh38
rs376157066 Chr.19: 12677284-12677284 on Build GRCh38
rs922090415 Chr.19: 12677508-12677508 on Build GRCh38
rs750681585 Chr.19: 12677375-12677375 on Build GRCh38
rs778230502 Chr.19: 12677300-12677300 on Build GRCh38
rs904313226 Chr.19: 12677480-12677480 on Build GRCh38
rs756762989 Chr.19: 12677365-12677366 on Build GRCh38
rs575787787 Chr.19: 12677189-12677189 on Build GRCh38
rs753258751 Chr.19: 12677297-12677297 on Build GRCh38
rs534945961 Chr.19: 12677545-12677545 on Build GRCh38
rs764915726 Chr.19: 12677271-12677271 on Build GRCh38
rs766382313 Chr.19: 12677474-12677474 on Build GRCh38
rs756511088 Chr.19: 12677298-12677298 on Build GRCh38
rs749528954 Chr.19: 12677224-12677224 on Build GRCh38
rs547740533 Chr.19: 12677372-12677372 on Build GRCh38
rs78624364 Chr.19: 12677590-12677590 on Build GRCh38
rs765716761 Chr.19: 12677266-12677268 on Build GRCh38
rs754205298 Chr.19: 12677309-12677309 on Build GRCh38
rs187418820 Chr.19: 12677426-12677426 on Build GRCh38
rs568560231 Chr.19: 12677230-12677230 on Build GRCh38
rs757666183 Chr.19: 12677314-12677314 on Build GRCh38
rs181350133 Chr.19: 12677138-12677138 on Build GRCh38
rs561221935 Chr.19: 12677585-12677585 on Build GRCh38
rs113571246 Chr.19: 12677342-12677342 on Build GRCh38
rs747522720 Chr.19: 12677119-12677119 on Build GRCh38
rs979253304 Chr.19: 12677412-12677412 on Build GRCh38
rs777019316 Chr.19: 12677445-12677445 on Build GRCh38
rs367826439 Chr.19: 12677388-12677388 on Build GRCh38
rs374500589 Chr.19: 12677373-12677373 on Build GRCh38
rs780219273 Chr.19: 12677328-12677328 on Build GRCh38
rs781692345 Chr.19: 12677193-12677193 on Build GRCh38
rs147232397 Chr.19: 12677550-12677550 on Build GRCh38
rs772294853 Chr.19: 12677326-12677326 on Build GRCh38
rs1033105341 Chr.19: 12677413-12677413 on Build GRCh38
rs988829468 Chr.19: 12677453-12677453 on Build GRCh38
rs565337083 Chr.19: 12677223-12677223 on Build GRCh38
rs762534456 Chr.19: 12677356-12677356 on Build GRCh38
rs755051406 Chr.19: 12677391-12677391 on Build GRCh38
rs944925441 Chr.19: 12677465-12677465 on Build GRCh38
rs140764692 Chr.19: 12677147-12677147 on Build GRCh38
rs770020319 Chr.19: 12677143-12677143 on Build GRCh38
rs373000198 Chr.19: 12677227-12677227 on Build GRCh38
rs116774671 Chr.19: 12677581-12677581 on Build GRCh38
rs774505760 Chr.19: 12677150-12677150 on Build GRCh38
rs775628435 Chr.19: 12677235-12677235 on Build GRCh38
rs756557667 Chr.19: 12677222-12677222 on Build GRCh38
rs751589042 Chr.19: 12677482-12677482 on Build GRCh38
rs574751661 Chr.19: 12677579-12677579 on Build GRCh38
rs995883582 Chr.19: 12677123-12677123 on Build GRCh38
rs750922335 Chr.19: 12677471-12677472 on Build GRCh38
rs987402768 Chr.19: 12677534-12677534 on Build GRCh38
rs764762126 Chr.19: 12677188-12677188 on Build GRCh38
rs371100778 Chr.19: 12677349-12677349 on Build GRCh38
rs144283239 Chr.19: 12677144-12677144 on Build GRCh38
rs762347442 Chr.19: 12677447-12677447 on Build GRCh38
rs775538805 Chr.19: 12677167-12677167 on Build GRCh38
rs760586806 Chr.19: 12677168-12677168 on Build GRCh38
rs995929106 Chr.19: 12677501-12677501 on Build GRCh38

View Full Product Details