Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11963457-11963963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK3
Entrez Gene ID
27122
Gene Name
dickkopf WNT signaling pathway inhibitor 3
Gene Aliases
REIC, RIG
UniGene
Hs.292156
Gene Chromosome Location
Chr.11: 11962996-12009370 on Build GRCh38
Transcripts
NM_001018057.1 NM_013253.4 NM_015881.5 XM_006718178.2 XM_017017553.1 XM_017017554.1 XM_017017555.1

SNP Detail

SNP ID Chromosome Location
rs200966495 Chr.11: 11963585-11963590 on Build GRCh38
rs71720144 Chr.11: 11963591-11963596 on Build GRCh38
rs979008190 Chr.11: 11963750-11963750 on Build GRCh38
rs887112724 Chr.11: 11963602-11963602 on Build GRCh38
rs934025764 Chr.11: 11963828-11963828 on Build GRCh38
rs118111563 Chr.11: 11963686-11963686 on Build GRCh38
rs547174692 Chr.11: 11963829-11963829 on Build GRCh38
rs754774401 Chr.11: 11963813-11963813 on Build GRCh38
rs185027615 Chr.11: 11963489-11963489 on Build GRCh38
rs554019287 Chr.11: 11963775-11963775 on Build GRCh38
rs182001705 Chr.11: 11963705-11963705 on Build GRCh38
rs1038708982 Chr.11: 11963659-11963659 on Build GRCh38
rs1020578933 Chr.11: 11963736-11963736 on Build GRCh38
rs977495740 Chr.11: 11963786-11963786 on Build GRCh38
rs749842345 Chr.11: 11963586-11963587 on Build GRCh38
rs941255218 Chr.11: 11963656-11963656 on Build GRCh38
rs769818616 Chr.11: 11963842-11963843 on Build GRCh38
rs199949466 Chr.11: 11963591-11963591 on Build GRCh38
rs929483215 Chr.11: 11963920-11963920 on Build GRCh38
rs941379180 Chr.11: 11963918-11963918 on Build GRCh38
rs1047092576 Chr.11: 11963592-11963592 on Build GRCh38
rs751760580 Chr.11: 11963855-11963855 on Build GRCh38
rs898008781 Chr.11: 11963646-11963646 on Build GRCh38
rs1005663885 Chr.11: 11963699-11963699 on Build GRCh38
rs1029473168 Chr.11: 11963753-11963753 on Build GRCh38
rs998005906 Chr.11: 11963741-11963741 on Build GRCh38
rs575949655 Chr.11: 11963876-11963876 on Build GRCh38
rs979425513 Chr.11: 11963506-11963506 on Build GRCh38
rs142843448 Chr.11: 11963587-11963592 on Build GRCh38
rs896231431 Chr.11: 11963713-11963713 on Build GRCh38
rs536278976 Chr.11: 11963824-11963824 on Build GRCh38
rs569599830 Chr.11: 11963603-11963603 on Build GRCh38
rs138940704 Chr.11: 11963697-11963697 on Build GRCh38
rs904999571 Chr.11: 11963688-11963688 on Build GRCh38
rs950272496 Chr.11: 11963847-11963847 on Build GRCh38
rs922669160 Chr.11: 11963579-11963579 on Build GRCh38
rs753645667 Chr.11: 11963634-11963634 on Build GRCh38
rs565019537 Chr.11: 11963522-11963523 on Build GRCh38
rs549682860 Chr.11: 11963545-11963545 on Build GRCh38
rs757360565 Chr.11: 11963908-11963908 on Build GRCh38
rs186387336 Chr.11: 11963796-11963796 on Build GRCh38
rs529756397 Chr.11: 11963526-11963526 on Build GRCh38
rs993853120 Chr.11: 11963681-11963681 on Build GRCh38
rs189543796 Chr.11: 11963508-11963508 on Build GRCh38
rs963979793 Chr.11: 11963724-11963724 on Build GRCh38
rs964318305 Chr.11: 11963791-11963791 on Build GRCh38
rs942865117 Chr.11: 11963534-11963534 on Build GRCh38
rs188439232 Chr.11: 11963849-11963849 on Build GRCh38
rs957388735 Chr.11: 11963818-11963818 on Build GRCh38
rs1018187927 Chr.11: 11963707-11963707 on Build GRCh38
rs534302111 Chr.11: 11963720-11963720 on Build GRCh38

View Full Product Details