Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4448328-4448858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
UniGene
Hs.459779
Gene Chromosome Location
Chr.16: 4425805-4456775 on Build GRCh38
Transcripts
NM_001135110.2 NM_001286516.1 NM_005147.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2920499 c.1210C>T p.H404Y Chr.16: 4448817-4448817 on Build GRCh38
3817986 c.1167T>G p.G389G Chr.16: 4448774-4448774 on Build GRCh38
2920493 c.1184G>A p.R395Q Chr.16: 4448791-4448791 on Build GRCh38
6051307 c.1198G>A p.G400S Chr.16: 4448805-4448805 on Build GRCh38
7309673 c.1137G>A p.G379G Chr.16: 4448744-4448744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754903303 Chr.16: 4448780-4448780 on Build GRCh38
rs577822007 Chr.16: 4448395-4448395 on Build GRCh38
rs186560686 Chr.16: 4448368-4448368 on Build GRCh38
rs537804316 Chr.16: 4448602-4448602 on Build GRCh38
rs374949136 Chr.16: 4448706-4448706 on Build GRCh38
rs367945866 Chr.16: 4448683-4448683 on Build GRCh38
rs748410800 Chr.16: 4448693-4448693 on Build GRCh38
rs8048039 Chr.16: 4448485-4448485 on Build GRCh38
rs529005130 Chr.16: 4448429-4448429 on Build GRCh38
rs183740870 Chr.16: 4448614-4448614 on Build GRCh38
rs759995110 Chr.16: 4448809-4448809 on Build GRCh38
rs749067505 Chr.16: 4448533-4448533 on Build GRCh38
rs779591735 Chr.16: 4448768-4448768 on Build GRCh38
rs965966614 Chr.16: 4448635-4448635 on Build GRCh38
rs767814911 Chr.16: 4448741-4448741 on Build GRCh38
rs144382022 Chr.16: 4448593-4448593 on Build GRCh38
rs966002818 Chr.16: 4448656-4448656 on Build GRCh38
rs770091535 Chr.16: 4448696-4448696 on Build GRCh38
rs984417808 Chr.16: 4448710-4448710 on Build GRCh38
rs375478818 Chr.16: 4448582-4448582 on Build GRCh38
rs750067919 Chr.16: 4448766-4448766 on Build GRCh38
rs957160894 Chr.16: 4448413-4448413 on Build GRCh38
rs149535474 Chr.16: 4448457-4448457 on Build GRCh38
rs144171308 Chr.16: 4448466-4448466 on Build GRCh38
rs567434142 Chr.16: 4448578-4448578 on Build GRCh38
rs920181628 Chr.16: 4448789-4448789 on Build GRCh38
rs560729194 Chr.16: 4448398-4448398 on Build GRCh38
rs894713778 Chr.16: 4448361-4448361 on Build GRCh38
rs757958184 Chr.16: 4448767-4448767 on Build GRCh38
rs1028662371 Chr.16: 4448669-4448669 on Build GRCh38
rs908957603 Chr.16: 4448461-4448461 on Build GRCh38
rs148777688 Chr.16: 4448828-4448828 on Build GRCh38
rs887176245 Chr.16: 4448604-4448604 on Build GRCh38
rs773439058 Chr.16: 4448700-4448700 on Build GRCh38
rs764799382 Chr.16: 4448758-4448758 on Build GRCh38
rs981469060 Chr.16: 4448786-4448786 on Build GRCh38
rs918193138 Chr.16: 4448556-4448556 on Build GRCh38
rs764715883 Chr.16: 4448394-4448394 on Build GRCh38
rs147164370 Chr.16: 4448739-4448739 on Build GRCh38
rs968160187 Chr.16: 4448678-4448678 on Build GRCh38
rs950420416 Chr.16: 4448703-4448703 on Build GRCh38
rs542364234 Chr.16: 4448782-4448782 on Build GRCh38
rs577959957 Chr.16: 4448659-4448659 on Build GRCh38
rs764425444 Chr.16: 4448819-4448819 on Build GRCh38
rs894754181 Chr.16: 4448498-4448498 on Build GRCh38
rs540518960 Chr.16: 4448397-4448397 on Build GRCh38
rs761091031 Chr.16: 4448814-4448814 on Build GRCh38
rs868315319 Chr.16: 4448637-4448637 on Build GRCh38
rs756823839 Chr.16: 4448646-4448650 on Build GRCh38
rs535599820 Chr.16: 4448589-4448589 on Build GRCh38
rs771039488 Chr.16: 4448729-4448729 on Build GRCh38
rs918889498 Chr.16: 4448453-4448453 on Build GRCh38
rs1032850187 Chr.16: 4448412-4448412 on Build GRCh38
rs995516921 Chr.16: 4448594-4448594 on Build GRCh38
rs199799945 Chr.16: 4448743-4448743 on Build GRCh38
rs749509441 Chr.16: 4448795-4448795 on Build GRCh38
rs985956377 Chr.16: 4448414-4448414 on Build GRCh38
rs372419915 Chr.16: 4448360-4448360 on Build GRCh38
rs1045454433 Chr.16: 4448373-4448373 on Build GRCh38
rs562479853 Chr.16: 4448451-4448451 on Build GRCh38
rs8047438 Chr.16: 4448532-4448532 on Build GRCh38
rs1004709571 Chr.16: 4448436-4448436 on Build GRCh38
rs770967257 Chr.16: 4448804-4448804 on Build GRCh38
rs778848053 Chr.16: 4448810-4448810 on Build GRCh38
rs368706161 Chr.16: 4448750-4448750 on Build GRCh38
rs948278318 Chr.16: 4448577-4448577 on Build GRCh38
rs1014278311 Chr.16: 4448371-4448371 on Build GRCh38
rs1018763480 Chr.16: 4448616-4448616 on Build GRCh38
rs751489092 Chr.16: 4448771-4448771 on Build GRCh38
rs1027490343 Chr.16: 4448396-4448396 on Build GRCh38
rs756016318 Chr.16: 4448492-4448492 on Build GRCh38
rs191191272 Chr.16: 4448599-4448599 on Build GRCh38
rs996464724 Chr.16: 4448644-4448644 on Build GRCh38
rs992305284 Chr.16: 4448762-4448762 on Build GRCh38
rs754873799 Chr.16: 4448559-4448559 on Build GRCh38
rs542676990 Chr.16: 4448448-4448448 on Build GRCh38
rs373320261 Chr.16: 4448352-4448352 on Build GRCh38
rs896736085 Chr.16: 4448367-4448367 on Build GRCh38
rs775850018 Chr.16: 4448813-4448813 on Build GRCh38
rs747893201 Chr.16: 4448790-4448790 on Build GRCh38
rs1017843826 Chr.16: 4448642-4448642 on Build GRCh38
rs774460631 Chr.16: 4448805-4448805 on Build GRCh38
rs540908577 Chr.16: 4448715-4448715 on Build GRCh38
rs937749066 Chr.16: 4448748-4448748 on Build GRCh38
rs921568044 Chr.16: 4448460-4448460 on Build GRCh38
rs774781065 Chr.16: 4448737-4448737 on Build GRCh38
rs898592419 Chr.16: 4448374-4448374 on Build GRCh38
rs868528749 Chr.16: 4448353-4448353 on Build GRCh38
rs547309662 Chr.16: 4448572-4448572 on Build GRCh38
rs967834406 Chr.16: 4448462-4448462 on Build GRCh38
rs770146336 Chr.16: 4448791-4448791 on Build GRCh38

View Full Product Details