Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99946573-99947079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB14
Entrez Gene ID
79982
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Gene Aliases
EGNR9427, PRO34683
UniGene
Hs.577426
Gene Chromosome Location
Chr.4: 99896248-99946726 on Build GRCh38
Transcripts
NM_001031723.3 NM_001278310.1 NM_001278311.1 XM_017008628.1

SNP Detail

SNP ID Chromosome Location
rs546092392 Chr.4: 99946600-99946600 on Build GRCh38
rs1015127291 Chr.4: 99946670-99946670 on Build GRCh38
rs1021558373 Chr.4: 99946718-99946718 on Build GRCh38
rs926938402 Chr.4: 99947010-99947010 on Build GRCh38
rs564398598 Chr.4: 99946650-99946650 on Build GRCh38
rs1010546909 Chr.4: 99946714-99946714 on Build GRCh38
rs951594191 Chr.4: 99946791-99946791 on Build GRCh38
rs1018315065 Chr.4: 99946866-99946866 on Build GRCh38
rs976959920 Chr.4: 99946632-99946632 on Build GRCh38
rs889281852 Chr.4: 99946963-99946963 on Build GRCh38
rs779832127 Chr.4: 99946617-99946617 on Build GRCh38
rs960650095 Chr.4: 99947029-99947029 on Build GRCh38
rs923825989 Chr.4: 99946705-99946705 on Build GRCh38
rs1031312535 Chr.4: 99946865-99946865 on Build GRCh38
rs192640615 Chr.4: 99947040-99947040 on Build GRCh38
rs550031527 Chr.4: 99946706-99946706 on Build GRCh38
rs1056128606 Chr.4: 99946611-99946611 on Build GRCh38
rs547856536 Chr.4: 99946911-99946911 on Build GRCh38
rs903202951 Chr.4: 99946987-99946987 on Build GRCh38
rs966595748 Chr.4: 99946671-99946671 on Build GRCh38
rs768359313 Chr.4: 99946622-99946622 on Build GRCh38
rs905621511 Chr.4: 99946674-99946674 on Build GRCh38
rs965499276 Chr.4: 99946608-99946608 on Build GRCh38
rs941719076 Chr.4: 99946661-99946661 on Build GRCh38
rs1029057375 Chr.4: 99946673-99946673 on Build GRCh38
rs1011845249 Chr.4: 99946634-99946634 on Build GRCh38
rs978058559 Chr.4: 99946864-99946864 on Build GRCh38
rs757493938 Chr.4: 99946602-99946602 on Build GRCh38
rs945869672 Chr.4: 99947026-99947026 on Build GRCh38
rs908898433 Chr.4: 99946902-99946902 on Build GRCh38
rs558904157 Chr.4: 99947017-99947017 on Build GRCh38
rs368625421 Chr.4: 99946855-99946855 on Build GRCh38
rs968033606 Chr.4: 99946699-99946699 on Build GRCh38
rs976904279 Chr.4: 99946769-99946769 on Build GRCh38
rs779280281 Chr.4: 99946859-99946859 on Build GRCh38
rs990117747 Chr.4: 99946690-99946690 on Build GRCh38
rs1018370832 Chr.4: 99946752-99946752 on Build GRCh38
rs991913645 Chr.4: 99946886-99946886 on Build GRCh38
rs938363175 Chr.4: 99947013-99947013 on Build GRCh38
rs1011667970 Chr.4: 99946842-99946842 on Build GRCh38
rs1042931162 Chr.4: 99947033-99947033 on Build GRCh38
rs111410193 Chr.4: 99946887-99946887 on Build GRCh38
rs1017600384 Chr.4: 99946955-99946955 on Build GRCh38
rs940324619 Chr.4: 99946916-99946916 on Build GRCh38
rs984750516 Chr.4: 99946800-99946800 on Build GRCh38
rs751749129 Chr.4: 99946595-99946595 on Build GRCh38
rs1022110495 Chr.4: 99946977-99946977 on Build GRCh38
rs757556588 Chr.4: 99946857-99946857 on Build GRCh38
rs556310701 Chr.4: 99947047-99947047 on Build GRCh38
rs947725537 Chr.4: 99946781-99946781 on Build GRCh38
rs781421850 Chr.4: 99946610-99946610 on Build GRCh38
rs112389785 Chr.4: 99946824-99946824 on Build GRCh38
rs916296939 Chr.4: 99946766-99946766 on Build GRCh38
rs891595058 Chr.4: 99946836-99946836 on Build GRCh38
rs1017693113 Chr.4: 99946663-99946663 on Build GRCh38
rs918832592 Chr.4: 99946643-99946643 on Build GRCh38
rs1005542824 Chr.4: 99946667-99946667 on Build GRCh38
rs942615171 Chr.4: 99947056-99947056 on Build GRCh38
rs997654537 Chr.4: 99946831-99946831 on Build GRCh38
rs1050624778 Chr.4: 99946960-99946960 on Build GRCh38
rs1049784443 Chr.4: 99946655-99946655 on Build GRCh38
rs887995272 Chr.4: 99946665-99946665 on Build GRCh38
rs939138767 Chr.4: 99946596-99946596 on Build GRCh38
rs1048758622 Chr.4: 99946790-99946790 on Build GRCh38
rs918779946 Chr.4: 99946779-99946779 on Build GRCh38
rs756177038 Chr.4: 99946616-99946616 on Build GRCh38
rs1029091940 Chr.4: 99946832-99946832 on Build GRCh38
rs568277286 Chr.4: 99946924-99946924 on Build GRCh38
rs775790779 Chr.4: 99946965-99946965 on Build GRCh38
rs953371118 Chr.4: 99946686-99946686 on Build GRCh38
rs887432983 Chr.4: 99946806-99946806 on Build GRCh38
rs550327006 Chr.4: 99946717-99946717 on Build GRCh38
rs570699788 Chr.4: 99947022-99947022 on Build GRCh38
rs745938379 Chr.4: 99946615-99946615 on Build GRCh38
rs960543303 Chr.4: 99946880-99946880 on Build GRCh38
rs933479908 Chr.4: 99946945-99946945 on Build GRCh38
rs951596260 Chr.4: 99946922-99946922 on Build GRCh38
rs930247581 Chr.4: 99946645-99946645 on Build GRCh38
rs938459084 Chr.4: 99946825-99946825 on Build GRCh38
rs531827004 Chr.4: 99946695-99946695 on Build GRCh38
rs749146046 Chr.4: 99946618-99946618 on Build GRCh38
rs947839752 Chr.4: 99946894-99946894 on Build GRCh38
rs997621944 Chr.4: 99946672-99946672 on Build GRCh38
rs371077668 Chr.4: 99946968-99946968 on Build GRCh38
rs1021664742 Chr.4: 99946850-99946850 on Build GRCh38
rs1044079266 Chr.4: 99946819-99946819 on Build GRCh38
rs912985793 Chr.4: 99947025-99947025 on Build GRCh38
rs1037001504 Chr.4: 99946817-99946817 on Build GRCh38
rs964734228 Chr.4: 99946681-99946681 on Build GRCh38
rs984933929 Chr.4: 99946899-99946899 on Build GRCh38
rs979721734 Chr.4: 99947004-99947004 on Build GRCh38
rs1056924091 Chr.4: 99946677-99946677 on Build GRCh38
rs984307144 Chr.4: 99946952-99946952 on Build GRCh38
rs978409285 Chr.4: 99946704-99946704 on Build GRCh38
rs940979647 Chr.4: 99946812-99946812 on Build GRCh38
rs1006900412 Chr.4: 99946728-99946728 on Build GRCh38
rs1047369877 Chr.4: 99946648-99946648 on Build GRCh38
rs941652051 Chr.4: 99946811-99946816 on Build GRCh38
rs991889241 Chr.4: 99946719-99946719 on Build GRCh38
rs969092371 Chr.4: 99946727-99946727 on Build GRCh38
rs902061267 Chr.4: 99946942-99946942 on Build GRCh38
rs889167341 Chr.4: 99946839-99946839 on Build GRCh38
rs1025740066 Chr.4: 99946915-99946915 on Build GRCh38

View Full Product Details