Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128218888-128219287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
UniGene
Hs.522413
Gene Chromosome Location
Chr.9: 128203355-128255248 on Build GRCh38
Transcripts
NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654561 c.574G>A p.E192K Chr.9: 128219237-128219237 on Build GRCh38
5558796 c.542T>C p.L181P Chr.9: 128219205-128219205 on Build GRCh38
1674857 c.394C>A p.L132M Chr.9: 128219057-128219057 on Build GRCh38
3716216 c.438G>A p.G146G Chr.9: 128219101-128219101 on Build GRCh38
4390687 c.536C>T p.S179F Chr.9: 128219199-128219199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11788598 Chr.9: 128219016-128219016 on Build GRCh38
rs763487889 Chr.9: 128219020-128219020 on Build GRCh38
rs1004906778 Chr.9: 128219092-128219092 on Build GRCh38
rs762841745 Chr.9: 128219239-128219239 on Build GRCh38
rs367581351 Chr.9: 128219011-128219011 on Build GRCh38
rs183885342 Chr.9: 128219053-128219053 on Build GRCh38
rs911827492 Chr.9: 128219040-128219040 on Build GRCh38
rs1022351123 Chr.9: 128219263-128219263 on Build GRCh38
rs1057518655 Chr.9: 128219130-128219131 on Build GRCh38
rs776275818 Chr.9: 128219204-128219204 on Build GRCh38
rs956432874 Chr.9: 128219005-128219005 on Build GRCh38
rs747255317 Chr.9: 128219001-128219001 on Build GRCh38
rs780514615 Chr.9: 128219247-128219247 on Build GRCh38
rs765080648 Chr.9: 128219218-128219218 on Build GRCh38
rs141695422 Chr.9: 128219035-128219035 on Build GRCh38
rs927628227 Chr.9: 128219002-128219002 on Build GRCh38
rs1051766292 Chr.9: 128219074-128219074 on Build GRCh38
rs746270475 Chr.9: 128219015-128219015 on Build GRCh38
rs1003737693 Chr.9: 128218954-128218954 on Build GRCh38
rs987855255 Chr.9: 128219262-128219262 on Build GRCh38
rs181308722 Chr.9: 128218935-128218935 on Build GRCh38
rs994732590 Chr.9: 128219139-128219139 on Build GRCh38
rs1003369790 Chr.9: 128219245-128219245 on Build GRCh38
rs746030356 Chr.9: 128219089-128219089 on Build GRCh38
rs772763746 Chr.9: 128219025-128219025 on Build GRCh38
rs748627087 Chr.9: 128219012-128219012 on Build GRCh38
rs36009729 Chr.9: 128219051-128219051 on Build GRCh38
rs568247101 Chr.9: 128219137-128219137 on Build GRCh38
rs749870166 Chr.9: 128219114-128219114 on Build GRCh38
rs777767122 Chr.9: 128219065-128219065 on Build GRCh38
rs548983793 Chr.9: 128218976-128218976 on Build GRCh38
rs772636298 Chr.9: 128219141-128219141 on Build GRCh38
rs587777860 Chr.9: 128219192-128219192 on Build GRCh38
rs758325864 Chr.9: 128219050-128219050 on Build GRCh38
rs981816587 Chr.9: 128219000-128219000 on Build GRCh38
rs904992382 Chr.9: 128219236-128219236 on Build GRCh38
rs146242842 Chr.9: 128219191-128219191 on Build GRCh38
rs759118746 Chr.9: 128219206-128219206 on Build GRCh38
rs548927481 Chr.9: 128218987-128218987 on Build GRCh38
rs781503957 Chr.9: 128219081-128219081 on Build GRCh38
rs143834670 Chr.9: 128219113-128219113 on Build GRCh38
rs766023182 Chr.9: 128219032-128219032 on Build GRCh38
rs757372393 Chr.9: 128219257-128219257 on Build GRCh38
rs535456711 Chr.9: 128219267-128219267 on Build GRCh38
rs550052311 Chr.9: 128219230-128219230 on Build GRCh38
rs771549971 Chr.9: 128219021-128219021 on Build GRCh38
rs751407306 Chr.9: 128219256-128219256 on Build GRCh38
rs772469890 Chr.9: 128219030-128219030 on Build GRCh38
rs988210313 Chr.9: 128219008-128219008 on Build GRCh38
rs764950748 Chr.9: 128219041-128219041 on Build GRCh38
rs935840154 Chr.9: 128219140-128219140 on Build GRCh38
rs767639839 Chr.9: 128219266-128219266 on Build GRCh38
rs746559414 Chr.9: 128219175-128219175 on Build GRCh38
rs141748215 Chr.9: 128219077-128219077 on Build GRCh38
rs141079649 Chr.9: 128219225-128219225 on Build GRCh38
rs770287739 Chr.9: 128219111-128219111 on Build GRCh38
rs759343711 Chr.9: 128219039-128219039 on Build GRCh38
rs568834961 Chr.9: 128218979-128218979 on Build GRCh38
rs776155686 Chr.9: 128219018-128219018 on Build GRCh38

View Full Product Details