Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32679281-32679783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1L
Entrez Gene ID
10059
Gene Name
dynamin 1 like
Gene Aliases
DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV
UniGene
Hs.556296
Gene Chromosome Location
Chr.12: 32679200-32745650 on Build GRCh38
Transcripts
NM_001278463.1 NM_001278464.1 NM_001278465.1 NM_001278466.1 NM_005690.4 NM_012062.4 NM_012063.3 XM_005253282.4 XM_011520543.2 XM_011520544.2 XM_017018663.1 XM_017018664.1 XM_017018665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3739614 c.58G>A p.A20T Chr.12: 32679421-32679421 on Build GRCh38
3396069 c.56G>T p.G19V Chr.12: 32679419-32679419 on Build GRCh38
3396070 c.56G>T p.G19V Chr.12: 32679419-32679419 on Build GRCh38
7429252 c.38A>T p.D13V Chr.12: 32679401-32679401 on Build GRCh38
3739613 c.58G>A p.A20T Chr.12: 32679421-32679421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896148786 Chr.12: 32679409-32679409 on Build GRCh38
rs767578893 Chr.12: 32679354-32679354 on Build GRCh38
rs766636851 Chr.12: 32679311-32679311 on Build GRCh38
rs747115615 Chr.12: 32679347-32679347 on Build GRCh38
rs1009004153 Chr.12: 32679603-32679603 on Build GRCh38
rs151285821 Chr.12: 32679363-32679363 on Build GRCh38
rs771990382 Chr.12: 32679438-32679438 on Build GRCh38
rs754614254 Chr.12: 32679357-32679357 on Build GRCh38
rs201968952 Chr.12: 32679350-32679350 on Build GRCh38
rs191084732 Chr.12: 32679339-32679339 on Build GRCh38
rs201348675 Chr.12: 32679399-32679399 on Build GRCh38
rs781103166 Chr.12: 32679388-32679388 on Build GRCh38
rs200885280 Chr.12: 32679552-32679569 on Build GRCh38
rs760304487 Chr.12: 32679355-32679355 on Build GRCh38
rs996170934 Chr.12: 32679677-32679677 on Build GRCh38
rs771767552 Chr.12: 32679488-32679488 on Build GRCh38
rs758693731 Chr.12: 32679361-32679361 on Build GRCh38
rs867424801 Chr.12: 32679534-32679551 on Build GRCh38
rs772586756 Chr.12: 32679344-32679344 on Build GRCh38
rs200938430 Chr.12: 32679411-32679411 on Build GRCh38
rs772176772 Chr.12: 32679308-32679308 on Build GRCh38
rs374067166 Chr.12: 32679348-32679348 on Build GRCh38
rs771090587 Chr.12: 32679380-32679380 on Build GRCh38
rs748628452 Chr.12: 32679307-32679307 on Build GRCh38
rs776650839 Chr.12: 32679313-32679313 on Build GRCh38
rs766295842 Chr.12: 32679492-32679492 on Build GRCh38
rs757231215 Chr.12: 32679322-32679322 on Build GRCh38
rs771381138 Chr.12: 32679389-32679389 on Build GRCh38
rs554801764 Chr.12: 32679561-32679561 on Build GRCh38
rs374414603 Chr.12: 32679316-32679316 on Build GRCh38
rs770521657 Chr.12: 32679326-32679326 on Build GRCh38
rs997430334 Chr.12: 32679581-32679581 on Build GRCh38
rs367813907 Chr.12: 32679317-32679317 on Build GRCh38
rs753070922 Chr.12: 32679360-32679360 on Build GRCh38
rs915515186 Chr.12: 32679330-32679330 on Build GRCh38
rs900480276 Chr.12: 32679661-32679661 on Build GRCh38
rs1046941909 Chr.12: 32679546-32679546 on Build GRCh38
rs114505859 Chr.12: 32679332-32679332 on Build GRCh38
rs748468670 Chr.12: 32679343-32679343 on Build GRCh38
rs550823466 Chr.12: 32679310-32679310 on Build GRCh38
rs778486856 Chr.12: 32679359-32679359 on Build GRCh38
rs760510351 Chr.12: 32679491-32679491 on Build GRCh38
rs372402179 Chr.12: 32679340-32679340 on Build GRCh38
rs1030287075 Chr.12: 32679594-32679594 on Build GRCh38
rs1045095164 Chr.12: 32679746-32679746 on Build GRCh38
rs574313739 Chr.12: 32679633-32679633 on Build GRCh38
rs759025359 Chr.12: 32679500-32679500 on Build GRCh38
rs1019474939 Chr.12: 32679647-32679647 on Build GRCh38
rs58142640 Chr.12: 32679305-32679305 on Build GRCh38
rs764639933 Chr.12: 32679495-32679495 on Build GRCh38
rs764871053 Chr.12: 32679501-32679501 on Build GRCh38
rs774908821 Chr.12: 32679306-32679306 on Build GRCh38
rs370381346 Chr.12: 32679356-32679356 on Build GRCh38
rs968278258 Chr.12: 32679698-32679698 on Build GRCh38
rs748892584 Chr.12: 32679338-32679338 on Build GRCh38
rs2649574 Chr.12: 32679705-32679705 on Build GRCh38
rs960705619 Chr.12: 32679718-32679718 on Build GRCh38
rs371570335 Chr.12: 32679319-32679319 on Build GRCh38
rs887498165 Chr.12: 32679515-32679515 on Build GRCh38
rs968505110 Chr.12: 32679638-32679638 on Build GRCh38
rs1022492454 Chr.12: 32679679-32679679 on Build GRCh38
rs761652857 Chr.12: 32679352-32679352 on Build GRCh38
rs961875049 Chr.12: 32679560-32679560 on Build GRCh38
rs1005980990 Chr.12: 32679519-32679519 on Build GRCh38
rs776969387 Chr.12: 32679349-32679349 on Build GRCh38
rs979489732 Chr.12: 32679651-32679651 on Build GRCh38
rs918189484 Chr.12: 32679712-32679712 on Build GRCh38
rs929787276 Chr.12: 32679539-32679539 on Build GRCh38
rs745921568 Chr.12: 32679391-32679391 on Build GRCh38
rs761707963 Chr.12: 32679416-32679416 on Build GRCh38
rs752341107 Chr.12: 32679502-32679502 on Build GRCh38
rs111956384 Chr.12: 32679695-32679695 on Build GRCh38
rs760693266 Chr.12: 32679309-32679309 on Build GRCh38
rs540976767 Chr.12: 32679493-32679493 on Build GRCh38
rs543177370 Chr.12: 32679732-32679732 on Build GRCh38
rs886485933 Chr.12: 32679550-32679550 on Build GRCh38
rs767523355 Chr.12: 32679325-32679325 on Build GRCh38
rs755525061 Chr.12: 32679329-32679329 on Build GRCh38
rs886049282 Chr.12: 32679314-32679314 on Build GRCh38
rs775798240 Chr.12: 32679414-32679414 on Build GRCh38
rs1031147360 Chr.12: 32679707-32679707 on Build GRCh38
rs887456524 Chr.12: 32679752-32679752 on Build GRCh38
rs747457741 Chr.12: 32679372-32679372 on Build GRCh38
rs773648742 Chr.12: 32679346-32679346 on Build GRCh38
rs985841546 Chr.12: 32679607-32679607 on Build GRCh38

View Full Product Details