Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93875392-93875891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP2
Entrez Gene ID
30836
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2
UniGene
Hs.601998
Gene Chromosome Location
Chr.1: 93869458-93879206 on Build GRCh38
Transcripts
NM_014597.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6865484 c.1749_1750insT p.K584fs*1 Chr.1: 93875701-93875702 on Build GRCh38
5689941 c.1755_1760delACAGTC p.Q586_S587delQS Chr.1: 93875691-93875696 on Build GRCh38
4530518 c.1704G>A p.L568L Chr.1: 93875747-93875747 on Build GRCh38
7218642 c.1740T>C p.F580F Chr.1: 93875711-93875711 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1029744264 Chr.1: 93875838-93875838 on Build GRCh38
rs753227748 Chr.1: 93875611-93875611 on Build GRCh38
rs776883430 Chr.1: 93875763-93875763 on Build GRCh38
rs2273165 Chr.1: 93875711-93875711 on Build GRCh38
rs927821888 Chr.1: 93875683-93875683 on Build GRCh38
rs996953150 Chr.1: 93875431-93875431 on Build GRCh38
rs371640394 Chr.1: 93875735-93875735 on Build GRCh38
rs758853165 Chr.1: 93875627-93875627 on Build GRCh38
rs761311700 Chr.1: 93875814-93875814 on Build GRCh38
rs751130310 Chr.1: 93875803-93875803 on Build GRCh38
rs200835030 Chr.1: 93875647-93875647 on Build GRCh38
rs750222636 Chr.1: 93875662-93875662 on Build GRCh38
rs749276200 Chr.1: 93875709-93875709 on Build GRCh38
rs767121828 Chr.1: 93875817-93875817 on Build GRCh38
rs377355673 Chr.1: 93875684-93875684 on Build GRCh38
rs763625772 Chr.1: 93875629-93875629 on Build GRCh38
rs899582909 Chr.1: 93875815-93875815 on Build GRCh38
rs955617724 Chr.1: 93875450-93875450 on Build GRCh38
rs893540651 Chr.1: 93875868-93875868 on Build GRCh38
rs893592483 Chr.1: 93875469-93875469 on Build GRCh38
rs775678976 Chr.1: 93875781-93875781 on Build GRCh38
rs1063601 Chr.1: 93875753-93875753 on Build GRCh38
rs1004843380 Chr.1: 93875853-93875853 on Build GRCh38
rs755913175 Chr.1: 93875663-93875663 on Build GRCh38
rs763458729 Chr.1: 93875792-93875792 on Build GRCh38
rs184813048 Chr.1: 93875550-93875550 on Build GRCh38
rs939212006 Chr.1: 93875685-93875685 on Build GRCh38
rs948809882 Chr.1: 93875590-93875590 on Build GRCh38
rs759868544 Chr.1: 93875765-93875765 on Build GRCh38
rs770217550 Chr.1: 93875773-93875773 on Build GRCh38
rs765704130 Chr.1: 93875605-93875605 on Build GRCh38
rs756920025 Chr.1: 93875641-93875641 on Build GRCh38
rs538150006 Chr.1: 93875583-93875583 on Build GRCh38
rs753498722 Chr.1: 93875618-93875619 on Build GRCh38
rs548165042 Chr.1: 93875553-93875553 on Build GRCh38
rs908502122 Chr.1: 93875508-93875508 on Build GRCh38
rs12082462 Chr.1: 93875418-93875418 on Build GRCh38
rs962755581 Chr.1: 93875540-93875540 on Build GRCh38
rs565750616 Chr.1: 93875489-93875489 on Build GRCh38
rs202212966 Chr.1: 93875422-93875423 on Build GRCh38
rs746763890 Chr.1: 93875589-93875589 on Build GRCh38
rs12046789 Chr.1: 93875704-93875704 on Build GRCh38
rs1035366745 Chr.1: 93875575-93875576 on Build GRCh38
rs1020640976 Chr.1: 93875500-93875500 on Build GRCh38
rs547153623 Chr.1: 93875448-93875448 on Build GRCh38
rs532266491 Chr.1: 93875419-93875419 on Build GRCh38
rs1010572302 Chr.1: 93875493-93875493 on Build GRCh38
rs1003433125 Chr.1: 93875559-93875559 on Build GRCh38
rs368806814 Chr.1: 93875696-93875696 on Build GRCh38
rs996590102 Chr.1: 93875823-93875823 on Build GRCh38
rs751008383 Chr.1: 93875640-93875640 on Build GRCh38
rs553865698 Chr.1: 93875839-93875839 on Build GRCh38
rs2273166 Chr.1: 93875581-93875581 on Build GRCh38
rs188843224 Chr.1: 93875799-93875799 on Build GRCh38
rs772527663 Chr.1: 93875743-93875743 on Build GRCh38
rs771344446 Chr.1: 93875739-93875739 on Build GRCh38
rs1022998868 Chr.1: 93875628-93875628 on Build GRCh38
rs943537462 Chr.1: 93875441-93875441 on Build GRCh38
rs982995994 Chr.1: 93875452-93875452 on Build GRCh38
rs376936090 Chr.1: 93875734-93875734 on Build GRCh38
rs750109852 Chr.1: 93875832-93875832 on Build GRCh38
rs572167310 Chr.1: 93875862-93875862 on Build GRCh38
rs754587702 Chr.1: 93875604-93875604 on Build GRCh38

View Full Product Details