Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36225203-36225735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY2
Entrez Gene ID
9980
Gene Name
dopey family member 2
Gene Aliases
21orf5, C21orf5
UniGene
Hs.204575
Gene Chromosome Location
Chr.21: 36156781-36294274 on Build GRCh38
Transcripts
NM_001320714.1 NM_005128.3 XM_017028509.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030487 c.1464C>A p.V488V Chr.21: 36225658-36225658 on Build GRCh38
6253528 c.1398G>T p.R466S Chr.21: 36225592-36225592 on Build GRCh38
5519762 c.1404C>T p.S468S Chr.21: 36225598-36225598 on Build GRCh38
3912060 c.1440C>T p.A480A Chr.21: 36225634-36225634 on Build GRCh38
6891606 c.1468C>T p.P490S Chr.21: 36225662-36225662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541132993 Chr.21: 36225253-36225254 on Build GRCh38
rs754060532 Chr.21: 36225616-36225616 on Build GRCh38
rs372484886 Chr.21: 36225695-36225695 on Build GRCh38
rs781402818 Chr.21: 36225535-36225535 on Build GRCh38
rs756099172 Chr.21: 36225668-36225668 on Build GRCh38
rs139220161 Chr.21: 36225598-36225598 on Build GRCh38
rs763946178 Chr.21: 36225368-36225368 on Build GRCh38
rs939128428 Chr.21: 36225292-36225292 on Build GRCh38
rs747793961 Chr.21: 36225644-36225644 on Build GRCh38
rs910592449 Chr.21: 36225547-36225547 on Build GRCh38
rs1002566945 Chr.21: 36225630-36225630 on Build GRCh38
rs969035136 Chr.21: 36225705-36225705 on Build GRCh38
rs779751459 Chr.21: 36225697-36225697 on Build GRCh38
rs760122066 Chr.21: 36225540-36225541 on Build GRCh38
rs762487784 Chr.21: 36225672-36225672 on Build GRCh38
rs146962277 Chr.21: 36225483-36225483 on Build GRCh38
rs578180903 Chr.21: 36225313-36225313 on Build GRCh38
rs182321260 Chr.21: 36225621-36225621 on Build GRCh38
rs1047948153 Chr.21: 36225593-36225593 on Build GRCh38
rs549968514 Chr.21: 36225522-36225522 on Build GRCh38
rs775136760 Chr.21: 36225591-36225591 on Build GRCh38
rs535902103 Chr.21: 36225599-36225599 on Build GRCh38
rs79714985 Chr.21: 36225246-36225246 on Build GRCh38
rs142730557 Chr.21: 36225622-36225622 on Build GRCh38
rs987590364 Chr.21: 36225420-36225420 on Build GRCh38
rs981198478 Chr.21: 36225455-36225455 on Build GRCh38
rs970216462 Chr.21: 36225432-36225432 on Build GRCh38
rs920497199 Chr.21: 36225231-36225231 on Build GRCh38
rs538538998 Chr.21: 36225617-36225617 on Build GRCh38
rs371338959 Chr.21: 36225587-36225587 on Build GRCh38
rs756265773 Chr.21: 36225551-36225551 on Build GRCh38
rs1040560805 Chr.21: 36225279-36225279 on Build GRCh38
rs955842866 Chr.21: 36225690-36225690 on Build GRCh38
rs764218338 Chr.21: 36225373-36225373 on Build GRCh38
rs763274848 Chr.21: 36225322-36225322 on Build GRCh38
rs138343054 Chr.21: 36225611-36225611 on Build GRCh38
rs756943716 Chr.21: 36225509-36225509 on Build GRCh38
rs143255868 Chr.21: 36225623-36225623 on Build GRCh38
rs145667945 Chr.21: 36225586-36225586 on Build GRCh38
rs761100361 Chr.21: 36225677-36225677 on Build GRCh38
rs1035428487 Chr.21: 36225655-36225655 on Build GRCh38
rs904784033 Chr.21: 36225356-36225356 on Build GRCh38
rs140670312 Chr.21: 36225578-36225578 on Build GRCh38
rs776017850 Chr.21: 36225675-36225675 on Build GRCh38
rs962066037 Chr.21: 36225473-36225473 on Build GRCh38
rs527709649 Chr.21: 36225386-36225386 on Build GRCh38
rs2284631 Chr.21: 36225383-36225383 on Build GRCh38
rs943426015 Chr.21: 36225574-36225574 on Build GRCh38
rs1020530210 Chr.21: 36225323-36225323 on Build GRCh38
rs1056594267 Chr.21: 36225293-36225293 on Build GRCh38
rs988663563 Chr.21: 36225520-36225520 on Build GRCh38
rs905572702 Chr.21: 36225627-36225627 on Build GRCh38
rs1056245510 Chr.21: 36225577-36225577 on Build GRCh38
rs142732043 Chr.21: 36225388-36225388 on Build GRCh38
rs889205976 Chr.21: 36225251-36225251 on Build GRCh38
rs953640873 Chr.21: 36225400-36225400 on Build GRCh38
rs55892541 Chr.21: 36225564-36225564 on Build GRCh38
rs776411695 Chr.21: 36225640-36225640 on Build GRCh38
rs761271177 Chr.21: 36225519-36225519 on Build GRCh38
rs920549596 Chr.21: 36225492-36225492 on Build GRCh38
rs570655881 Chr.21: 36225318-36225318 on Build GRCh38
rs769492617 Chr.21: 36225654-36225654 on Build GRCh38
rs754324598 Chr.21: 36225693-36225693 on Build GRCh38
rs778099707 Chr.21: 36225552-36225552 on Build GRCh38
rs945086367 Chr.21: 36225579-36225579 on Build GRCh38
rs779131422 Chr.21: 36225618-36225618 on Build GRCh38
rs532293436 Chr.21: 36225546-36225546 on Build GRCh38
rs199879429 Chr.21: 36225613-36225613 on Build GRCh38
rs746938685 Chr.21: 36225631-36225631 on Build GRCh38
rs75698155 Chr.21: 36225457-36225457 on Build GRCh38
rs759213725 Chr.21: 36225232-36225232 on Build GRCh38
rs760570178 Chr.21: 36225594-36225594 on Build GRCh38
rs563067875 Chr.21: 36225268-36225268 on Build GRCh38
rs755477088 Chr.21: 36225523-36225523 on Build GRCh38
rs865913338 Chr.21: 36225254-36225254 on Build GRCh38
rs368171607 Chr.21: 36225659-36225659 on Build GRCh38
rs765839013 Chr.21: 36225673-36225673 on Build GRCh38
rs1019426155 Chr.21: 36225443-36225443 on Build GRCh38
rs75091966 Chr.21: 36225245-36225245 on Build GRCh38
rs750339936 Chr.21: 36225632-36225632 on Build GRCh38
rs771089082 Chr.21: 36225580-36225580 on Build GRCh38
rs571113551 Chr.21: 36225542-36225542 on Build GRCh38
rs199987084 Chr.21: 36225615-36225615 on Build GRCh38
rs368081231 Chr.21: 36225521-36225521 on Build GRCh38
rs560884449 Chr.21: 36225440-36225440 on Build GRCh38
rs375177464 Chr.21: 36225696-36225696 on Build GRCh38
rs374472573 Chr.21: 36225608-36225608 on Build GRCh38
rs764452963 Chr.21: 36225686-36225686 on Build GRCh38
rs962118828 Chr.21: 36225678-36225678 on Build GRCh38
rs904984552 Chr.21: 36225285-36225285 on Build GRCh38
rs768365405 Chr.21: 36225596-36225596 on Build GRCh38

View Full Product Details