Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74519457-74519955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DQX1
Entrez Gene ID
165545
Gene Name
DEAQ-box RNA dependent ATPase 1
Gene Aliases
-
UniGene
Hs.191705
Gene Chromosome Location
Chr.2: 74518131-74526281 on Build GRCh38
Transcripts
NM_133637.2 XM_011532644.1 XM_011532645.1 XM_017003520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1690724 c.1733C>T p.S578F Chr.2: 74519629-74519629 on Build GRCh38
1690725 c.1379C>T p.S460F Chr.2: 74519629-74519629 on Build GRCh38
3727034 c.1355T>A p.M452K Chr.2: 74519653-74519653 on Build GRCh38
3739924 c.1452+8G>T p.? Chr.2: 74519548-74519548 on Build GRCh38
5742514 c.1630G>T p.A544S Chr.2: 74519732-74519732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767873000 Chr.2: 74519662-74519664 on Build GRCh38
rs145654502 Chr.2: 74519691-74519691 on Build GRCh38
rs751937661 Chr.2: 74519678-74519678 on Build GRCh38
rs753343867 Chr.2: 74519873-74519873 on Build GRCh38
rs769414746 Chr.2: 74519613-74519613 on Build GRCh38
rs372419081 Chr.2: 74519907-74519907 on Build GRCh38
rs759725464 Chr.2: 74519652-74519652 on Build GRCh38
rs764283600 Chr.2: 74519760-74519760 on Build GRCh38
rs377186424 Chr.2: 74519912-74519912 on Build GRCh38
rs747931015 Chr.2: 74519888-74519888 on Build GRCh38
rs753085539 Chr.2: 74519541-74519541 on Build GRCh38
rs754034455 Chr.2: 74519528-74519528 on Build GRCh38
rs367904898 Chr.2: 74519607-74519607 on Build GRCh38
rs779232287 Chr.2: 74519530-74519530 on Build GRCh38
rs199690861 Chr.2: 74519731-74519731 on Build GRCh38
rs778097007 Chr.2: 74519592-74519592 on Build GRCh38
rs145106899 Chr.2: 74519648-74519648 on Build GRCh38
rs774087472 Chr.2: 74519636-74519636 on Build GRCh38
rs990228771 Chr.2: 74519544-74519544 on Build GRCh38
rs746951176 Chr.2: 74519903-74519903 on Build GRCh38
rs773318125 Chr.2: 74519743-74519743 on Build GRCh38
rs752874772 Chr.2: 74519756-74519757 on Build GRCh38
rs267599457 Chr.2: 74519576-74519576 on Build GRCh38
rs749377736 Chr.2: 74519931-74519931 on Build GRCh38
rs1034391645 Chr.2: 74519807-74519807 on Build GRCh38
rs779621715 Chr.2: 74519717-74519717 on Build GRCh38
rs369663123 Chr.2: 74519919-74519919 on Build GRCh38
rs762205722 Chr.2: 74519765-74519765 on Build GRCh38
rs774756227 Chr.2: 74519764-74519764 on Build GRCh38
rs150473457 Chr.2: 74519620-74519620 on Build GRCh38
rs372690839 Chr.2: 74519900-74519900 on Build GRCh38
rs141367262 Chr.2: 74519744-74519744 on Build GRCh38
rs931767714 Chr.2: 74519708-74519708 on Build GRCh38
rs374700854 Chr.2: 74519521-74519521 on Build GRCh38
rs983831139 Chr.2: 74519768-74519772 on Build GRCh38
rs765520131 Chr.2: 74519657-74519657 on Build GRCh38
rs374931711 Chr.2: 74519776-74519776 on Build GRCh38
rs750941458 Chr.2: 74519780-74519780 on Build GRCh38
rs778267619 Chr.2: 74519674-74519674 on Build GRCh38
rs770010456 Chr.2: 74519747-74519747 on Build GRCh38
rs151136945 Chr.2: 74519599-74519599 on Build GRCh38
rs200151764 Chr.2: 74519555-74519555 on Build GRCh38
rs781273773 Chr.2: 74519588-74519588 on Build GRCh38
rs760972225 Chr.2: 74519896-74519896 on Build GRCh38
rs201414755 Chr.2: 74519709-74519709 on Build GRCh38
rs754650935 Chr.2: 74519727-74519727 on Build GRCh38
rs754633804 Chr.2: 74519876-74519876 on Build GRCh38
rs910834737 Chr.2: 74519929-74519929 on Build GRCh38
rs138905883 Chr.2: 74519675-74519675 on Build GRCh38
rs117622212 Chr.2: 74519644-74519644 on Build GRCh38
rs768508488 Chr.2: 74519630-74519630 on Build GRCh38
rs368612069 Chr.2: 74519895-74519895 on Build GRCh38
rs548201031 Chr.2: 74519822-74519822 on Build GRCh38
rs958947864 Chr.2: 74519887-74519887 on Build GRCh38
rs755644140 Chr.2: 74519716-74519716 on Build GRCh38
rs755552519 Chr.2: 74519868-74519868 on Build GRCh38
rs199814471 Chr.2: 74519560-74519560 on Build GRCh38
rs756657595 Chr.2: 74519597-74519597 on Build GRCh38
rs200935798 Chr.2: 74519518-74519518 on Build GRCh38
rs764526320 Chr.2: 74519677-74519677 on Build GRCh38
rs745440948 Chr.2: 74519600-74519600 on Build GRCh38
rs961190054 Chr.2: 74519796-74519796 on Build GRCh38
rs778466905 Chr.2: 74519884-74519884 on Build GRCh38
rs758293958 Chr.2: 74519893-74519893 on Build GRCh38
rs753263732 Chr.2: 74519647-74519647 on Build GRCh38
rs765601572 Chr.2: 74519532-74519532 on Build GRCh38
rs775511545 Chr.2: 74519748-74519748 on Build GRCh38
rs772100580 Chr.2: 74519732-74519732 on Build GRCh38
rs762847165 Chr.2: 74519506-74519506 on Build GRCh38
rs267599458 Chr.2: 74519577-74519577 on Build GRCh38
rs778753797 Chr.2: 74519730-74519730 on Build GRCh38
rs200550092 Chr.2: 74519751-74519751 on Build GRCh38
rs370687787 Chr.2: 74519513-74519513 on Build GRCh38
rs1003286889 Chr.2: 74519781-74519781 on Build GRCh38
rs748973277 Chr.2: 74519724-74519724 on Build GRCh38
rs377273869 Chr.2: 74519871-74519871 on Build GRCh38
rs150191467 Chr.2: 74519596-74519596 on Build GRCh38
rs768063026 Chr.2: 74519702-74519702 on Build GRCh38

View Full Product Details