Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172768759-172769263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP1
Entrez Gene ID
1843
Gene Name
dual specificity phosphatase 1
Gene Aliases
CL100, HVH1, MKP-1, MKP1, PTPN10
UniGene
Hs.171695
Gene Chromosome Location
Chr.5: 172768090-172771200 on Build GRCh38
Transcripts
NM_004417.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138529 c.1036C>T p.P346S Chr.5: 172768830-172768830 on Build GRCh38
1436049 c.991C>T p.R331* Chr.5: 172768875-172768875 on Build GRCh38
5643271 c.1035C>G p.I345M Chr.5: 172768831-172768831 on Build GRCh38
1316700 c.1065C>A p.S355R Chr.5: 172768801-172768801 on Build GRCh38
7271187 c.779C>A p.A260E Chr.5: 172769087-172769087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766201794 Chr.5: 172769171-172769171 on Build GRCh38
rs777804572 Chr.5: 172768797-172768797 on Build GRCh38
rs752632230 Chr.5: 172768786-172768786 on Build GRCh38
rs145742179 Chr.5: 172768921-172768921 on Build GRCh38
rs781436876 Chr.5: 172768909-172768909 on Build GRCh38
rs758258767 Chr.5: 172769023-172769023 on Build GRCh38
rs771383057 Chr.5: 172768922-172768922 on Build GRCh38
rs756976896 Chr.5: 172768889-172768889 on Build GRCh38
rs11553855 Chr.5: 172768980-172768980 on Build GRCh38
rs779028235 Chr.5: 172768864-172768866 on Build GRCh38
rs1039398081 Chr.5: 172769154-172769154 on Build GRCh38
rs768678783 Chr.5: 172768828-172768828 on Build GRCh38
rs776389762 Chr.5: 172768992-172768992 on Build GRCh38
rs748175042 Chr.5: 172768912-172768912 on Build GRCh38
rs756082359 Chr.5: 172769035-172769035 on Build GRCh38
rs138196129 Chr.5: 172768966-172768966 on Build GRCh38
rs375653201 Chr.5: 172769115-172769115 on Build GRCh38
rs762397077 Chr.5: 172768874-172768874 on Build GRCh38
rs776180782 Chr.5: 172769156-172769156 on Build GRCh38
rs140814607 Chr.5: 172768789-172768789 on Build GRCh38
rs778085088 Chr.5: 172768913-172768913 on Build GRCh38
rs144374272 Chr.5: 172769019-172769019 on Build GRCh38
rs200141460 Chr.5: 172768873-172768873 on Build GRCh38
rs763365656 Chr.5: 172769218-172769218 on Build GRCh38
rs138845473 Chr.5: 172768816-172768816 on Build GRCh38
rs373970586 Chr.5: 172769151-172769151 on Build GRCh38
rs760298379 Chr.5: 172768875-172768875 on Build GRCh38
rs150141714 Chr.5: 172768808-172768808 on Build GRCh38
rs752980356 Chr.5: 172769034-172769034 on Build GRCh38
rs763215828 Chr.5: 172768855-172768855 on Build GRCh38
rs373945254 Chr.5: 172768927-172768927 on Build GRCh38
rs370523646 Chr.5: 172769146-172769146 on Build GRCh38
rs762680658 Chr.5: 172769170-172769170 on Build GRCh38
rs1029358805 Chr.5: 172769213-172769213 on Build GRCh38
rs997787518 Chr.5: 172769191-172769192 on Build GRCh38
rs749709268 Chr.5: 172769071-172769071 on Build GRCh38
rs771845071 Chr.5: 172768928-172768928 on Build GRCh38
rs188553172 Chr.5: 172768807-172768807 on Build GRCh38
rs774936615 Chr.5: 172769127-172769127 on Build GRCh38
rs753825565 Chr.5: 172768879-172768879 on Build GRCh38
rs369007899 Chr.5: 172768962-172768962 on Build GRCh38
rs767622754 Chr.5: 172769026-172769026 on Build GRCh38
rs760974212 Chr.5: 172769181-172769181 on Build GRCh38
rs369562158 Chr.5: 172768864-172768864 on Build GRCh38
rs140606584 Chr.5: 172768817-172768817 on Build GRCh38
rs1017979716 Chr.5: 172768890-172768890 on Build GRCh38
rs759059090 Chr.5: 172768780-172768780 on Build GRCh38
rs149572166 Chr.5: 172769011-172769011 on Build GRCh38
rs202068138 Chr.5: 172768893-172768893 on Build GRCh38
rs750190666 Chr.5: 172769020-172769020 on Build GRCh38
rs376856024 Chr.5: 172768945-172768945 on Build GRCh38
rs538360202 Chr.5: 172768859-172768859 on Build GRCh38
rs1005296397 Chr.5: 172768869-172768869 on Build GRCh38
rs752822774 Chr.5: 172769173-172769173 on Build GRCh38
rs774834828 Chr.5: 172768923-172768923 on Build GRCh38
rs772577199 Chr.5: 172769121-172769121 on Build GRCh38
rs746702483 Chr.5: 172769150-172769150 on Build GRCh38
rs748034797 Chr.5: 172769137-172769137 on Build GRCh38
rs1002522435 Chr.5: 172769031-172769031 on Build GRCh38
rs781276559 Chr.5: 172768833-172768833 on Build GRCh38
rs372415052 Chr.5: 172769089-172769089 on Build GRCh38
rs773838496 Chr.5: 172768840-172768840 on Build GRCh38
rs3172775 Chr.5: 172769225-172769225 on Build GRCh38
rs1010275198 Chr.5: 172769157-172769157 on Build GRCh38
rs899610561 Chr.5: 172769162-172769162 on Build GRCh38
rs141498783 Chr.5: 172769050-172769050 on Build GRCh38
rs201099793 Chr.5: 172768781-172768781 on Build GRCh38
rs770172254 Chr.5: 172768839-172768839 on Build GRCh38
rs746219697 Chr.5: 172769104-172769104 on Build GRCh38
rs1026735479 Chr.5: 172769120-172769120 on Build GRCh38
rs773194541 Chr.5: 172769167-172769167 on Build GRCh38
rs748312900 Chr.5: 172768837-172768837 on Build GRCh38
rs1013256630 Chr.5: 172769092-172769092 on Build GRCh38
rs1047333158 Chr.5: 172769130-172769130 on Build GRCh38
rs750513633 Chr.5: 172769065-172769065 on Build GRCh38
rs893987874 Chr.5: 172768841-172768841 on Build GRCh38
rs149004705 Chr.5: 172769163-172769163 on Build GRCh38
rs553540303 Chr.5: 172768987-172768987 on Build GRCh38
rs751735848 Chr.5: 172768894-172768894 on Build GRCh38
rs965870542 Chr.5: 172769165-172769165 on Build GRCh38
rs779630094 Chr.5: 172769095-172769095 on Build GRCh38
rs755174678 Chr.5: 172768899-172768899 on Build GRCh38

View Full Product Details