Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206648854-206649364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK3
Entrez Gene ID
8444
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Gene Aliases
DYRK5, RED, REDK, hYAK3-2
UniGene
Hs.164267
Gene Chromosome Location
Chr.1: 206635536-206655158 on Build GRCh38
Transcripts
NM_001004023.1 NM_003582.2 XM_005273315.4 XM_011510061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463839 c.1679G>T p.G560V Chr.1: 206648877-206648877 on Build GRCh38
6991130 c.1624A>G p.I542V Chr.1: 206648927-206648927 on Build GRCh38
6341250 c.1763G>T p.S588I Chr.1: 206648961-206648961 on Build GRCh38
463841 c.1734C>T p.P578P Chr.1: 206648932-206648932 on Build GRCh38
1668425 c.1670C>T p.P557L Chr.1: 206648868-206648868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782111789 Chr.1: 206648998-206648998 on Build GRCh38
rs201145019 Chr.1: 206648991-206648991 on Build GRCh38
rs781787559 Chr.1: 206648887-206648887 on Build GRCh38
rs576111587 Chr.1: 206649195-206649195 on Build GRCh38
rs552152813 Chr.1: 206649290-206649290 on Build GRCh38
rs909776730 Chr.1: 206649169-206649169 on Build GRCh38
rs572029974 Chr.1: 206649278-206649278 on Build GRCh38
rs185204467 Chr.1: 206649211-206649211 on Build GRCh38
rs970049659 Chr.1: 206649333-206649333 on Build GRCh38
rs782601735 Chr.1: 206648891-206648891 on Build GRCh38
rs782780113 Chr.1: 206648999-206648999 on Build GRCh38
rs150539050 Chr.1: 206649182-206649182 on Build GRCh38
rs200134906 Chr.1: 206649005-206649005 on Build GRCh38
rs781820020 Chr.1: 206648976-206648976 on Build GRCh38
rs202226255 Chr.1: 206649038-206649038 on Build GRCh38
rs200436281 Chr.1: 206649039-206649039 on Build GRCh38
rs782033949 Chr.1: 206648983-206648983 on Build GRCh38
rs896989650 Chr.1: 206649198-206649198 on Build GRCh38
rs200225267 Chr.1: 206649027-206649027 on Build GRCh38
rs201612172 Chr.1: 206649034-206649034 on Build GRCh38
rs782671806 Chr.1: 206648907-206648907 on Build GRCh38
rs956083360 Chr.1: 206649046-206649046 on Build GRCh38
rs1018289265 Chr.1: 206649246-206649246 on Build GRCh38
rs199654996 Chr.1: 206649151-206649151 on Build GRCh38
rs192975758 Chr.1: 206649003-206649003 on Build GRCh38
rs782216378 Chr.1: 206648943-206648943 on Build GRCh38
rs200805518 Chr.1: 206649085-206649085 on Build GRCh38
rs141705007 Chr.1: 206649101-206649101 on Build GRCh38
rs199536987 Chr.1: 206649072-206649072 on Build GRCh38
rs782370025 Chr.1: 206648944-206648944 on Build GRCh38
rs201510016 Chr.1: 206649044-206649044 on Build GRCh38
rs1029374889 Chr.1: 206648997-206648997 on Build GRCh38
rs201787773 Chr.1: 206649026-206649026 on Build GRCh38
rs568923051 Chr.1: 206648905-206648905 on Build GRCh38
rs997969869 Chr.1: 206649207-206649207 on Build GRCh38
rs1000775981 Chr.1: 206649305-206649305 on Build GRCh38
rs889479257 Chr.1: 206649212-206649212 on Build GRCh38
rs576415405 Chr.1: 206649291-206649291 on Build GRCh38
rs201440619 Chr.1: 206649115-206649115 on Build GRCh38
rs201628362 Chr.1: 206649172-206649172 on Build GRCh38
rs969312127 Chr.1: 206649032-206649032 on Build GRCh38
rs782047095 Chr.1: 206648988-206648988 on Build GRCh38
rs1005365600 Chr.1: 206648959-206648959 on Build GRCh38
rs782292094 Chr.1: 206648913-206648913 on Build GRCh38
rs782598896 Chr.1: 206648942-206648942 on Build GRCh38
rs781860367 Chr.1: 206648893-206648893 on Build GRCh38
rs781966083 Chr.1: 206649055-206649055 on Build GRCh38
rs781821673 Chr.1: 206648993-206648993 on Build GRCh38
rs782138320 Chr.1: 206648877-206648877 on Build GRCh38
rs1036831718 Chr.1: 206649184-206649184 on Build GRCh38
rs1031877412 Chr.1: 206649308-206649308 on Build GRCh38
rs200892244 Chr.1: 206648886-206648886 on Build GRCh38
rs540849505 Chr.1: 206649321-206649321 on Build GRCh38
rs781981992 Chr.1: 206648965-206648965 on Build GRCh38
rs201018041 Chr.1: 206649215-206649215 on Build GRCh38
rs956198908 Chr.1: 206649314-206649314 on Build GRCh38
rs551854696 Chr.1: 206648976-206648976 on Build GRCh38
rs948707013 Chr.1: 206649076-206649076 on Build GRCh38
rs917280001 Chr.1: 206649073-206649073 on Build GRCh38
rs992900203 Chr.1: 206649320-206649320 on Build GRCh38
rs148139981 Chr.1: 206648978-206648978 on Build GRCh38
rs200756836 Chr.1: 206649191-206649191 on Build GRCh38
rs189793069 Chr.1: 206649228-206649228 on Build GRCh38
rs200754147 Chr.1: 206649165-206649165 on Build GRCh38
rs141266804 Chr.1: 206648919-206648919 on Build GRCh38
rs74147411 Chr.1: 206649013-206649013 on Build GRCh38
rs549234649 Chr.1: 206648890-206648890 on Build GRCh38
rs201457150 Chr.1: 206649197-206649197 on Build GRCh38
rs782074368 Chr.1: 206649120-206649120 on Build GRCh38

View Full Product Details