Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 62771322-62771809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
UniGene
Hs.271667
Gene Chromosome Location
Chr.2: 62673851-63046487 on Build GRCh38
Transcripts
NM_001142614.1 NM_001142615.2 NM_001142616.1 NM_015252.3 XM_005264225.2 XM_005264226.2 XM_005264227.1 XM_005264228.2 XM_006711971.2 XM_011532713.2 XM_011532714.2 XM_011532715.2 XM_011532716.2 XM_011532717.1 XM_011532718.2 XM_017003643.1 XM_017003644.1 XM_017003645.1 XM_017003646.1 XM_017003647.1 XM_017003648.1 XM_017003649.1 XM_017003650.1 XM_017003651.1 XM_017003652.1 XM_017003653.1 XM_017003654.1 XM_017003655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4917532 c.308A>C p.E103A Chr.2: 62771388-62771388 on Build GRCh38
359449 c.269C>T p.A90V Chr.2: 62771349-62771349 on Build GRCh38
4539683 c.271G>A p.E91K Chr.2: 62771351-62771351 on Build GRCh38
3391807 c.259-3C>T p.? Chr.2: 62771336-62771336 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935792174 Chr.2: 62771645-62771645 on Build GRCh38
rs570199848 Chr.2: 62771664-62771664 on Build GRCh38
rs761348668 Chr.2: 62771429-62771429 on Build GRCh38
rs537729801 Chr.2: 62771744-62771744 on Build GRCh38
rs183559027 Chr.2: 62771747-62771747 on Build GRCh38
rs760889210 Chr.2: 62771439-62771439 on Build GRCh38
rs992850872 Chr.2: 62771569-62771569 on Build GRCh38
rs1018278696 Chr.2: 62771720-62771720 on Build GRCh38
rs972199193 Chr.2: 62771734-62771734 on Build GRCh38
rs772628096 Chr.2: 62771436-62771437 on Build GRCh38
rs766769630 Chr.2: 62771450-62771450 on Build GRCh38
rs559859257 Chr.2: 62771616-62771616 on Build GRCh38
rs75835972 Chr.2: 62771350-62771350 on Build GRCh38
rs906504869 Chr.2: 62771615-62771615 on Build GRCh38
rs767149214 Chr.2: 62771437-62771437 on Build GRCh38
rs745656110 Chr.2: 62771398-62771398 on Build GRCh38
rs200927057 Chr.2: 62771349-62771349 on Build GRCh38
rs1016015572 Chr.2: 62771657-62771657 on Build GRCh38
rs548907699 Chr.2: 62771424-62771424 on Build GRCh38
rs766440108 Chr.2: 62771663-62771663 on Build GRCh38
rs1054312623 Chr.2: 62771656-62771656 on Build GRCh38
rs979779431 Chr.2: 62771781-62771781 on Build GRCh38
rs891121446 Chr.2: 62771662-62771662 on Build GRCh38
rs894434883 Chr.2: 62771639-62771639 on Build GRCh38
rs201855702 Chr.2: 62771400-62771400 on Build GRCh38
rs951646335 Chr.2: 62771564-62771564 on Build GRCh38
rs866700227 Chr.2: 62771376-62771376 on Build GRCh38
rs758671591 Chr.2: 62771384-62771384 on Build GRCh38
rs571283361 Chr.2: 62771630-62771630 on Build GRCh38
rs866434152 Chr.2: 62771738-62771738 on Build GRCh38
rs192016883 Chr.2: 62771660-62771660 on Build GRCh38
rs1006846738 Chr.2: 62771668-62771668 on Build GRCh38
rs918812340 Chr.2: 62771529-62771529 on Build GRCh38
rs748332929 Chr.2: 62771377-62771377 on Build GRCh38
rs779080782 Chr.2: 62771353-62771353 on Build GRCh38
rs910804599 Chr.2: 62771751-62771751 on Build GRCh38
rs974218124 Chr.2: 62771512-62771512 on Build GRCh38
rs1044129494 Chr.2: 62771627-62771627 on Build GRCh38
rs1046491796 Chr.2: 62771609-62771609 on Build GRCh38
rs768015523 Chr.2: 62771423-62771423 on Build GRCh38
rs910835837 Chr.2: 62771480-62771480 on Build GRCh38
rs898605652 Chr.2: 62771723-62771723 on Build GRCh38
rs985848704 Chr.2: 62771749-62771749 on Build GRCh38
rs373948377 Chr.2: 62771418-62771418 on Build GRCh38
rs891645855 Chr.2: 62771660-62771661 on Build GRCh38
rs113585035 Chr.2: 62771504-62771504 on Build GRCh38
rs916917048 Chr.2: 62771583-62771583 on Build GRCh38
rs560529331 Chr.2: 62771459-62771459 on Build GRCh38
rs933017912 Chr.2: 62771417-62771417 on Build GRCh38
rs892641105 Chr.2: 62771590-62771590 on Build GRCh38
rs936868729 Chr.2: 62771771-62771771 on Build GRCh38
rs772904659 Chr.2: 62771438-62771438 on Build GRCh38
rs936817208 Chr.2: 62771497-62771497 on Build GRCh38
rs922901399 Chr.2: 62771413-62771413 on Build GRCh38
rs1006094694 Chr.2: 62771642-62771642 on Build GRCh38
rs945411624 Chr.2: 62771599-62771599 on Build GRCh38
rs369784262 Chr.2: 62771411-62771411 on Build GRCh38
rs971019106 Chr.2: 62771776-62771776 on Build GRCh38
rs778182203 Chr.2: 62771392-62771392 on Build GRCh38
rs538364118 Chr.2: 62771656-62771656 on Build GRCh38
rs996022065 Chr.2: 62771728-62771728 on Build GRCh38
rs961729487 Chr.2: 62771370-62771370 on Build GRCh38

View Full Product Details