Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80137785-80138299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A3
Entrez Gene ID
9775
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
DDX48, MUK34, NMP265, NUK34, RCPS, eIF4AIII
UniGene
Hs.389649
Gene Chromosome Location
Chr.17: 80135214-80147183 on Build GRCh38
Transcripts
NM_014740.3 XM_011525522.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880825 c.781G>C p.E261Q Chr.17: 80138228-80138228 on Build GRCh38
6380683 c.784G>T p.E262* Chr.17: 80138225-80138225 on Build GRCh38
6418177 c.808G>T p.D270Y Chr.17: 80138201-80138201 on Build GRCh38
5485921 c.782A>G p.E261G Chr.17: 80138227-80138227 on Build GRCh38
6421718 c.826A>G p.T276A Chr.17: 80138183-80138183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006166838 Chr.17: 80137917-80137917 on Build GRCh38
rs955431184 Chr.17: 80138039-80138039 on Build GRCh38
rs149116773 Chr.17: 80137998-80137998 on Build GRCh38
rs35229151 Chr.17: 80138142-80138142 on Build GRCh38
rs587777204 Chr.17: 80138200-80138200 on Build GRCh38
rs938383862 Chr.17: 80138091-80138091 on Build GRCh38
rs767330769 Chr.17: 80137981-80137981 on Build GRCh38
rs1010316448 Chr.17: 80137844-80137844 on Build GRCh38
rs1002710212 Chr.17: 80137879-80137879 on Build GRCh38
rs572675637 Chr.17: 80137826-80137826 on Build GRCh38
rs79086383 Chr.17: 80137960-80137961 on Build GRCh38
rs1016592289 Chr.17: 80137936-80137936 on Build GRCh38
rs749139245 Chr.17: 80138095-80138095 on Build GRCh38
rs138448499 Chr.17: 80137979-80137979 on Build GRCh38
rs61756263 Chr.17: 80138244-80138244 on Build GRCh38
rs994198278 Chr.17: 80138154-80138154 on Build GRCh38
rs372548278 Chr.17: 80138133-80138133 on Build GRCh38
rs2361710 Chr.17: 80138078-80138078 on Build GRCh38
rs935375317 Chr.17: 80137964-80137964 on Build GRCh38
rs894300087 Chr.17: 80137911-80137911 on Build GRCh38
rs774111367 Chr.17: 80138106-80138106 on Build GRCh38
rs762283395 Chr.17: 80138125-80138125 on Build GRCh38
rs149634224 Chr.17: 80138118-80138118 on Build GRCh38
rs965687973 Chr.17: 80137863-80137863 on Build GRCh38
rs974990377 Chr.17: 80137864-80137864 on Build GRCh38
rs1025728698 Chr.17: 80138010-80138010 on Build GRCh38
rs868108374 Chr.17: 80138102-80138102 on Build GRCh38
rs368234190 Chr.17: 80138117-80138117 on Build GRCh38
rs752875401 Chr.17: 80138193-80138193 on Build GRCh38
rs180766063 Chr.17: 80138048-80138048 on Build GRCh38
rs924009651 Chr.17: 80137886-80137886 on Build GRCh38
rs146360583 Chr.17: 80137877-80137877 on Build GRCh38
rs796647524 Chr.17: 80137904-80137904 on Build GRCh38
rs990403363 Chr.17: 80138058-80138058 on Build GRCh38
rs756092935 Chr.17: 80138196-80138196 on Build GRCh38
rs144407953 Chr.17: 80138250-80138250 on Build GRCh38
rs777692344 Chr.17: 80138094-80138094 on Build GRCh38
rs751807674 Chr.17: 80138170-80138170 on Build GRCh38
rs542103045 Chr.17: 80138079-80138079 on Build GRCh38
rs1041739110 Chr.17: 80138085-80138085 on Build GRCh38
rs1054625616 Chr.17: 80137814-80137814 on Build GRCh38
rs568433391 Chr.17: 80138267-80138267 on Build GRCh38
rs777927648 Chr.17: 80138211-80138211 on Build GRCh38
rs763432971 Chr.17: 80138151-80138151 on Build GRCh38
rs906969937 Chr.17: 80138088-80138088 on Build GRCh38
rs185319349 Chr.17: 80138103-80138103 on Build GRCh38
rs139804240 Chr.17: 80138169-80138169 on Build GRCh38
rs1034161506 Chr.17: 80137903-80137903 on Build GRCh38
rs78585555 Chr.17: 80138110-80138110 on Build GRCh38
rs577031103 Chr.17: 80137972-80137972 on Build GRCh38
rs973108521 Chr.17: 80137990-80137990 on Build GRCh38
rs189912101 Chr.17: 80137873-80137873 on Build GRCh38
rs750167983 Chr.17: 80138040-80138040 on Build GRCh38
rs368423482 Chr.17: 80138235-80138235 on Build GRCh38
rs942856249 Chr.17: 80138049-80138049 on Build GRCh38
rs905796740 Chr.17: 80138025-80138025 on Build GRCh38
rs377221362 Chr.17: 80138229-80138229 on Build GRCh38
rs781352571 Chr.17: 80138190-80138190 on Build GRCh38
rs771670148 Chr.17: 80138260-80138260 on Build GRCh38

View Full Product Details