Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50201671-50202185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL4
Entrez Gene ID
1996
Gene Name
ELAV like neuron-specific RNA binding protein 4
Gene Aliases
HUD, PNEM
UniGene
Hs.213050
Gene Chromosome Location
Chr.1: 50048014-50203786 on Build GRCh38
Transcripts
NM_001144774.2 NM_001144775.2 NM_001144776.2 NM_001144777.2 NM_001294348.1 NM_001324208.1 NM_001324209.1 NM_001324212.1 NM_001324213.1 NM_001324214.1 NM_001324215.1 NM_021952.4 XM_006710411.3 XM_011540889.2 XM_011540890.2 XM_011540893.2 XM_011540894.2 XM_011540895.2 XM_017000538.1 XM_017000539.1

SNP Detail

SNP ID Chromosome Location
rs976939516 Chr.1: 50202066-50202066 on Build GRCh38
rs368949570 Chr.1: 50202151-50202151 on Build GRCh38
rs531277179 Chr.1: 50201812-50201812 on Build GRCh38
rs945757767 Chr.1: 50202152-50202152 on Build GRCh38
rs12085063 Chr.1: 50201729-50201729 on Build GRCh38
rs528185675 Chr.1: 50202009-50202009 on Build GRCh38
rs910931852 Chr.1: 50202008-50202008 on Build GRCh38
rs80237691 Chr.1: 50202114-50202114 on Build GRCh38
rs754018830 Chr.1: 50202067-50202067 on Build GRCh38
rs540590695 Chr.1: 50201931-50201931 on Build GRCh38
rs1014645744 Chr.1: 50201842-50201842 on Build GRCh38
rs939397135 Chr.1: 50202073-50202074 on Build GRCh38
rs993741791 Chr.1: 50201948-50201948 on Build GRCh38
rs778785144 Chr.1: 50201987-50201987 on Build GRCh38
rs952149288 Chr.1: 50202006-50202006 on Build GRCh38
rs939365755 Chr.1: 50201809-50201809 on Build GRCh38
rs912165553 Chr.1: 50201719-50201719 on Build GRCh38
rs986322314 Chr.1: 50202007-50202007 on Build GRCh38
rs986426832 Chr.1: 50201705-50201705 on Build GRCh38
rs939982407 Chr.1: 50202161-50202161 on Build GRCh38
rs761117144 Chr.1: 50201748-50201749 on Build GRCh38
rs1024292707 Chr.1: 50201902-50201902 on Build GRCh38
rs904362865 Chr.1: 50201721-50201721 on Build GRCh38
rs915425286 Chr.1: 50201782-50201782 on Build GRCh38
rs926688684 Chr.1: 50201806-50201806 on Build GRCh38
rs564724605 Chr.1: 50202080-50202080 on Build GRCh38
rs1041577738 Chr.1: 50202156-50202156 on Build GRCh38
rs181271753 Chr.1: 50202042-50202042 on Build GRCh38
rs887046759 Chr.1: 50201829-50201829 on Build GRCh38
rs956964900 Chr.1: 50201730-50201730 on Build GRCh38
rs1000036171 Chr.1: 50201742-50201742 on Build GRCh38
rs996596752 Chr.1: 50201904-50201905 on Build GRCh38
rs908554998 Chr.1: 50202145-50202145 on Build GRCh38
rs35504756 Chr.1: 50201847-50201848 on Build GRCh38
rs77398614 Chr.1: 50202115-50202115 on Build GRCh38
rs946833239 Chr.1: 50201783-50201783 on Build GRCh38
rs980855126 Chr.1: 50201804-50201804 on Build GRCh38
rs143923811 Chr.1: 50201928-50201928 on Build GRCh38
rs1041653245 Chr.1: 50201707-50201707 on Build GRCh38
rs939973108 Chr.1: 50201868-50201868 on Build GRCh38
rs894574179 Chr.1: 50201824-50201824 on Build GRCh38
rs560605605 Chr.1: 50202001-50202001 on Build GRCh38
rs968244112 Chr.1: 50202011-50202011 on Build GRCh38
rs922266707 Chr.1: 50202139-50202139 on Build GRCh38
rs1031318249 Chr.1: 50201988-50201988 on Build GRCh38

View Full Product Details