Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36154429-36154920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L1
Entrez Gene ID
2036
Gene Name
erythrocyte membrane protein band 4.1 like 1
Gene Aliases
4.1N, MRD11
UniGene
Hs.437422
Gene Chromosome Location
Chr.20: 36064891-36232799 on Build GRCh38
Transcripts
NM_001258329.1 NM_001258330.1 NM_001258331.1 NM_012156.2 NM_177996.2 XM_011528667.1 XM_011528671.1 XM_011528673.1 XM_011528677.1 XM_017027711.1 XM_017027712.1 XM_017027713.1 XM_017027714.1 XM_017027716.1 XM_017027717.1 XM_017027718.1 XM_017027719.1 XM_017027720.1

SNP Detail

SNP ID Chromosome Location
rs978569478 Chr.20: 36154821-36154821 on Build GRCh38
rs917977173 Chr.20: 36154495-36154495 on Build GRCh38
rs991238608 Chr.20: 36154481-36154481 on Build GRCh38
rs541774308 Chr.20: 36154581-36154581 on Build GRCh38
rs1054023053 Chr.20: 36154608-36154608 on Build GRCh38
rs959292121 Chr.20: 36154769-36154769 on Build GRCh38
rs1052056895 Chr.20: 36154885-36154885 on Build GRCh38
rs1049864061 Chr.20: 36154667-36154667 on Build GRCh38
rs1015371146 Chr.20: 36154737-36154737 on Build GRCh38
rs967450288 Chr.20: 36154802-36154802 on Build GRCh38
rs959340672 Chr.20: 36154468-36154468 on Build GRCh38
rs949505381 Chr.20: 36154533-36154533 on Build GRCh38
rs1042518111 Chr.20: 36154548-36154548 on Build GRCh38
rs1034164556 Chr.20: 36154636-36154636 on Build GRCh38
rs559956948 Chr.20: 36154693-36154693 on Build GRCh38
rs111230707 Chr.20: 36154884-36154884 on Build GRCh38
rs1008384492 Chr.20: 36154713-36154714 on Build GRCh38
rs896973102 Chr.20: 36154743-36154743 on Build GRCh38
rs936853360 Chr.20: 36154597-36154597 on Build GRCh38
rs1051892749 Chr.20: 36154872-36154872 on Build GRCh38
rs932300903 Chr.20: 36154653-36154653 on Build GRCh38
rs936799572 Chr.20: 36154852-36154852 on Build GRCh38
rs1025395356 Chr.20: 36154782-36154782 on Build GRCh38
rs924047075 Chr.20: 36154838-36154838 on Build GRCh38
rs1019996649 Chr.20: 36154793-36154793 on Build GRCh38
rs1026827703 Chr.20: 36154764-36154764 on Build GRCh38
rs890834810 Chr.20: 36154705-36154705 on Build GRCh38
rs957147785 Chr.20: 36154642-36154642 on Build GRCh38
rs6058419 Chr.20: 36154884-36154884 on Build GRCh38
rs989660376 Chr.20: 36154870-36154870 on Build GRCh38
rs900774249 Chr.20: 36154646-36154646 on Build GRCh38
rs577451138 Chr.20: 36154531-36154531 on Build GRCh38
rs995855505 Chr.20: 36154871-36154871 on Build GRCh38
rs995315885 Chr.20: 36154763-36154763 on Build GRCh38
rs975751359 Chr.20: 36154812-36154812 on Build GRCh38
rs1011323176 Chr.20: 36154766-36154766 on Build GRCh38

View Full Product Details