Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 65319761-65320261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA5
Entrez Gene ID
2044
Gene Name
EPH receptor A5
Gene Aliases
CEK7, EHK-1, EHK1, EK7, HEK7, TYRO4
UniGene
Hs.654492
Gene Chromosome Location
Chr.4: 65319563-65670838 on Build GRCh38
Transcripts
NM_001281765.2 NM_001281766.2 NM_001318761.1 NM_004439.7 NM_182472.4 XM_005265653.3 XM_011531735.2 XM_017007878.1 XM_017007880.1

SNP Detail

SNP ID Chromosome Location
rs976122445 Chr.4: 65319969-65319969 on Build GRCh38
rs146517923 Chr.4: 65319998-65319998 on Build GRCh38
rs80169835 Chr.4: 65320097-65320097 on Build GRCh38
rs986420508 Chr.4: 65319842-65319842 on Build GRCh38
rs1020626665 Chr.4: 65319977-65319977 on Build GRCh38
rs552153674 Chr.4: 65320015-65320015 on Build GRCh38
rs1009189335 Chr.4: 65319963-65319963 on Build GRCh38
rs747204361 Chr.4: 65320176-65320176 on Build GRCh38
rs1030554359 Chr.4: 65319817-65319817 on Build GRCh38
rs192510299 Chr.4: 65320170-65320170 on Build GRCh38
rs953775546 Chr.4: 65319821-65319821 on Build GRCh38
rs961059913 Chr.4: 65319870-65319870 on Build GRCh38
rs554735214 Chr.4: 65320116-65320116 on Build GRCh38
rs949168134 Chr.4: 65319911-65319911 on Build GRCh38
rs1053492415 Chr.4: 65320076-65320076 on Build GRCh38
rs926436805 Chr.4: 65320197-65320197 on Build GRCh38
rs1035462570 Chr.4: 65319843-65319843 on Build GRCh38
rs943562172 Chr.4: 65319951-65319951 on Build GRCh38
rs953598269 Chr.4: 65320045-65320045 on Build GRCh38
rs893456058 Chr.4: 65319950-65319950 on Build GRCh38
rs905086391 Chr.4: 65319922-65319922 on Build GRCh38
rs976228498 Chr.4: 65319812-65319812 on Build GRCh38
rs986779218 Chr.4: 65320083-65320083 on Build GRCh38
rs565605410 Chr.4: 65320046-65320046 on Build GRCh38
rs747646697 Chr.4: 65319930-65319932 on Build GRCh38
rs749201720 Chr.4: 65319906-65319906 on Build GRCh38
rs1002736354 Chr.4: 65320214-65320214 on Build GRCh38
rs182146200 Chr.4: 65320217-65320217 on Build GRCh38
rs772083184 Chr.4: 65319862-65319862 on Build GRCh38
rs1002621899 Chr.4: 65319839-65319839 on Build GRCh38
rs538343088 Chr.4: 65319916-65319916 on Build GRCh38
rs758532928 Chr.4: 65320019-65320019 on Build GRCh38
rs76455416 Chr.4: 65319997-65319997 on Build GRCh38
rs937893980 Chr.4: 65320233-65320233 on Build GRCh38
rs905730429 Chr.4: 65319819-65319819 on Build GRCh38
rs972138909 Chr.4: 65319866-65319866 on Build GRCh38
rs1053739474 Chr.4: 65319935-65319935 on Build GRCh38
rs187124666 Chr.4: 65320152-65320152 on Build GRCh38
rs568656382 Chr.4: 65320126-65320126 on Build GRCh38
rs149103918 Chr.4: 65320010-65320010 on Build GRCh38
rs540791026 Chr.4: 65319857-65319857 on Build GRCh38
rs180965980 Chr.4: 65319924-65319924 on Build GRCh38
rs982242205 Chr.4: 65320189-65320189 on Build GRCh38
rs919315780 Chr.4: 65320161-65320161 on Build GRCh38
rs972785866 Chr.4: 65319897-65319897 on Build GRCh38
rs866810479 Chr.4: 65319816-65319816 on Build GRCh38
rs560768779 Chr.4: 65319862-65319862 on Build GRCh38

View Full Product Details