Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48828836-48829377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAS
Entrez Gene ID
3266
Gene Name
ES cell expressed Ras
Gene Aliases
HRAS2, HRASP
UniGene
Hs.447330
Gene Chromosome Location
Chr.X: 48826513-48829869 on Build GRCh38
Transcripts
NM_181532.3

SNP Detail

SNP ID Chromosome Location
rs782212177 Chr.X: 48829298-48829298 on Build GRCh38
rs782373989 Chr.X: 48829277-48829277 on Build GRCh38
rs781936384 Chr.X: 48829189-48829189 on Build GRCh38
rs180877146 Chr.X: 48829049-48829049 on Build GRCh38
rs921680985 Chr.X: 48828914-48828914 on Build GRCh38
rs782547746 Chr.X: 48829166-48829166 on Build GRCh38
rs781963613 Chr.X: 48829291-48829291 on Build GRCh38
rs188037509 Chr.X: 48829034-48829034 on Build GRCh38
rs1048789775 Chr.X: 48828937-48828937 on Build GRCh38
rs782116749 Chr.X: 48829123-48829123 on Build GRCh38
rs782095963 Chr.X: 48829113-48829114 on Build GRCh38
rs782606636 Chr.X: 48829268-48829268 on Build GRCh38
rs369226473 Chr.X: 48829333-48829333 on Build GRCh38
rs202013189 Chr.X: 48828894-48828894 on Build GRCh38
rs782452157 Chr.X: 48829274-48829274 on Build GRCh38
rs782306726 Chr.X: 48829118-48829118 on Build GRCh38
rs782745358 Chr.X: 48829153-48829153 on Build GRCh38
rs1032411630 Chr.X: 48829237-48829237 on Build GRCh38
rs367881874 Chr.X: 48828899-48828899 on Build GRCh38
rs1037039011 Chr.X: 48829041-48829041 on Build GRCh38
rs781891384 Chr.X: 48829040-48829040 on Build GRCh38
rs185614831 Chr.X: 48828977-48828977 on Build GRCh38
rs111320229 Chr.X: 48829216-48829216 on Build GRCh38
rs146098580 Chr.X: 48829267-48829267 on Build GRCh38
rs782521876 Chr.X: 48829217-48829217 on Build GRCh38
rs781861918 Chr.X: 48829261-48829261 on Build GRCh38
rs375020221 Chr.X: 48829198-48829198 on Build GRCh38
rs782018130 Chr.X: 48829095-48829095 on Build GRCh38
rs896704897 Chr.X: 48829046-48829046 on Build GRCh38
rs782675881 Chr.X: 48829276-48829276 on Build GRCh38
rs782764481 Chr.X: 48829214-48829214 on Build GRCh38
rs781946945 Chr.X: 48829156-48829156 on Build GRCh38
rs782341405 Chr.X: 48828873-48828873 on Build GRCh38
rs1014176841 Chr.X: 48829168-48829168 on Build GRCh38
rs782195123 Chr.X: 48829271-48829271 on Build GRCh38
rs782685954 Chr.X: 48829203-48829203 on Build GRCh38
rs782770684 Chr.X: 48829218-48829218 on Build GRCh38
rs931771202 Chr.X: 48828924-48828924 on Build GRCh38

View Full Product Details