Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204151793-204152297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK2
Entrez Gene ID
55224
Gene Name
ethanolamine kinase 2
Gene Aliases
EKI2, HMFT1716
UniGene
Hs.497469
Gene Chromosome Location
Chr.1: 204131061-204152182 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4168823 c.25C>T p.Q9* Chr.1: 204151828-204151828 on Build GRCh38
4168824 c.25C>T p.Q9* Chr.1: 204151828-204151828 on Build GRCh38
5845195 c.42delT p.H15fs*2 Chr.1: 204151811-204151811 on Build GRCh38
5845194 c.42delT p.H15fs*2 Chr.1: 204151811-204151811 on Build GRCh38
1338051 c.29C>A p.P10Q Chr.1: 204151824-204151824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539709927 Chr.1: 204152078-204152078 on Build GRCh38
rs1018450607 Chr.1: 204152200-204152200 on Build GRCh38
rs994157608 Chr.1: 204152182-204152182 on Build GRCh38
rs535427587 Chr.1: 204152192-204152192 on Build GRCh38
rs187440692 Chr.1: 204152029-204152029 on Build GRCh38
rs770609376 Chr.1: 204151853-204151853 on Build GRCh38
rs922985251 Chr.1: 204151972-204151972 on Build GRCh38
rs952977555 Chr.1: 204152081-204152081 on Build GRCh38
rs191312273 Chr.1: 204152109-204152109 on Build GRCh38
rs984399380 Chr.1: 204152027-204152027 on Build GRCh38
rs955727639 Chr.1: 204152144-204152144 on Build GRCh38
rs776557336 Chr.1: 204151861-204151861 on Build GRCh38
rs934403564 Chr.1: 204151986-204151986 on Build GRCh38
rs3737655 Chr.1: 204151824-204151824 on Build GRCh38
rs985668913 Chr.1: 204152089-204152089 on Build GRCh38
rs1025857326 Chr.1: 204151866-204151866 on Build GRCh38
rs986106989 Chr.1: 204151938-204151938 on Build GRCh38
rs895168308 Chr.1: 204151854-204151854 on Build GRCh38
rs922931464 Chr.1: 204152140-204152140 on Build GRCh38
rs897153050 Chr.1: 204152177-204152177 on Build GRCh38
rs927170634 Chr.1: 204152169-204152169 on Build GRCh38
rs988496922 Chr.1: 204152251-204152251 on Build GRCh38
rs965972884 Chr.1: 204152230-204152230 on Build GRCh38
rs952920992 Chr.1: 204151921-204151921 on Build GRCh38
rs1027112623 Chr.1: 204151920-204151920 on Build GRCh38
rs947271142 Chr.1: 204152009-204152009 on Build GRCh38
rs892968292 Chr.1: 204151999-204151999 on Build GRCh38
rs553260192 Chr.1: 204152106-204152106 on Build GRCh38
rs571045838 Chr.1: 204152256-204152256 on Build GRCh38
rs955675657 Chr.1: 204152242-204152242 on Build GRCh38
rs1057015267 Chr.1: 204152173-204152173 on Build GRCh38
rs1052897428 Chr.1: 204151992-204151992 on Build GRCh38
rs530942837 Chr.1: 204151941-204151941 on Build GRCh38
rs938616447 Chr.1: 204152170-204152170 on Build GRCh38
rs548341006 Chr.1: 204151870-204151870 on Build GRCh38
rs914277927 Chr.1: 204152274-204152274 on Build GRCh38
rs1036161441 Chr.1: 204151900-204151901 on Build GRCh38
rs1031288308 Chr.1: 204152237-204152237 on Build GRCh38
rs1035662120 Chr.1: 204152042-204152042 on Build GRCh38
rs914350317 Chr.1: 204152159-204152159 on Build GRCh38
rs12738530 Chr.1: 204151886-204151886 on Build GRCh38
rs988547874 Chr.1: 204152150-204152150 on Build GRCh38
rs1044250218 Chr.1: 204152167-204152167 on Build GRCh38
rs772697570 Chr.1: 204151825-204151825 on Build GRCh38
rs961455043 Chr.1: 204151901-204151901 on Build GRCh38
rs998432123 Chr.1: 204152234-204152234 on Build GRCh38
rs897208035 Chr.1: 204152044-204152044 on Build GRCh38
rs574193872 Chr.1: 204152075-204152090 on Build GRCh38
rs905847595 Chr.1: 204151873-204151873 on Build GRCh38
rs746714627 Chr.1: 204151840-204151840 on Build GRCh38
rs555106946 Chr.1: 204152225-204152225 on Build GRCh38
rs951600443 Chr.1: 204151908-204151908 on Build GRCh38
rs972842141 Chr.1: 204151902-204151902 on Build GRCh38
rs994209424 Chr.1: 204152053-204152053 on Build GRCh38
rs12738682 Chr.1: 204151934-204151934 on Build GRCh38
rs550740913 Chr.1: 204151971-204151971 on Build GRCh38
rs755121524 Chr.1: 204151818-204151818 on Build GRCh38
rs879472136 Chr.1: 204151819-204151819 on Build GRCh38
rs568533842 Chr.1: 204151893-204151893 on Build GRCh38
rs567872941 Chr.1: 204152189-204152190 on Build GRCh38
rs924060063 Chr.1: 204151817-204151817 on Build GRCh38
rs866567199 Chr.1: 204151967-204151967 on Build GRCh38
rs1044688624 Chr.1: 204152010-204152010 on Build GRCh38
rs943009914 Chr.1: 204151943-204151943 on Build GRCh38
rs747596168 Chr.1: 204151822-204151822 on Build GRCh38
rs905799718 Chr.1: 204152013-204152013 on Build GRCh38
rs947186253 Chr.1: 204152161-204152161 on Build GRCh38

View Full Product Details