Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37779224-37779775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC3
Entrez Gene ID
51010
Gene Name
exosome component 3
Gene Aliases
CGI-102, PCH1B, RRP40, Rrp40p, bA3J10.7, hRrp-40, p10
UniGene
Hs.602571
Gene Chromosome Location
Chr.9: 37779714-37785092 on Build GRCh38
Transcripts
NM_001002269.2 NM_016042.3 XM_005251375.2 XM_005251379.2 XM_011517735.2 XM_011517736.2 XM_011517738.2 XM_011517739.2 XM_017014313.1

Gene Symbol
TRMT10B
Entrez Gene ID
158234
Gene Name
tRNA methyltransferase 10B
Gene Aliases
RG9MTD3, bA3J10.9
UniGene
Hs.655614
Gene Chromosome Location
Chr.9: 37753204-37780459 on Build GRCh38
Transcripts
NM_001002269.2 NM_016042.3 XM_005251375.2 XM_005251379.2 XM_011517735.2 XM_011517736.2 XM_011517738.2 XM_011517739.2 XM_017014313.1

SNP Detail

SNP ID Chromosome Location
rs571596234 Chr.9: 37779304-37779304 on Build GRCh38
rs386734577 Chr.9: 37779406-37779407 on Build GRCh38
rs573008856 Chr.9: 37779466-37779466 on Build GRCh38
rs1012554968 Chr.9: 37779583-37779583 on Build GRCh38
rs543767697 Chr.9: 37779552-37779552 on Build GRCh38
rs942018067 Chr.9: 37779551-37779551 on Build GRCh38
rs6476661 Chr.9: 37779626-37779626 on Build GRCh38
rs778004560 Chr.9: 37779706-37779706 on Build GRCh38
rs886646999 Chr.9: 37779713-37779713 on Build GRCh38
rs895488742 Chr.9: 37779581-37779581 on Build GRCh38
rs911927037 Chr.9: 37779463-37779463 on Build GRCh38
rs189990241 Chr.9: 37779512-37779512 on Build GRCh38
rs959684217 Chr.9: 37779398-37779398 on Build GRCh38
rs114826945 Chr.9: 37779441-37779441 on Build GRCh38
rs930936600 Chr.9: 37779692-37779692 on Build GRCh38
rs943385646 Chr.9: 37779678-37779678 on Build GRCh38
rs770179108 Chr.9: 37779520-37779520 on Build GRCh38
rs979069828 Chr.9: 37779610-37779610 on Build GRCh38
rs943333363 Chr.9: 37779494-37779494 on Build GRCh38
rs530895640 Chr.9: 37779725-37779725 on Build GRCh38
rs780315329 Chr.9: 37779486-37779486 on Build GRCh38
rs551825536 Chr.9: 37779697-37779697 on Build GRCh38
rs909334004 Chr.9: 37779536-37779536 on Build GRCh38
rs1027164376 Chr.9: 37779324-37779324 on Build GRCh38
rs1004349531 Chr.9: 37779354-37779354 on Build GRCh38
rs539351168 Chr.9: 37779523-37779523 on Build GRCh38
rs891947197 Chr.9: 37779568-37779568 on Build GRCh38
rs755086968 Chr.9: 37779521-37779521 on Build GRCh38
rs997430945 Chr.9: 37779716-37779716 on Build GRCh38
rs968642496 Chr.9: 37779602-37779602 on Build GRCh38
rs946259035 Chr.9: 37779608-37779608 on Build GRCh38
rs182482824 Chr.9: 37779406-37779406 on Build GRCh38
rs528594042 Chr.9: 37779666-37779666 on Build GRCh38
rs927288549 Chr.9: 37779556-37779556 on Build GRCh38
rs7039137 Chr.9: 37779384-37779384 on Build GRCh38
rs185885745 Chr.9: 37779705-37779705 on Build GRCh38
rs199954489 Chr.9: 37779533-37779533 on Build GRCh38
rs575140512 Chr.9: 37779544-37779544 on Build GRCh38
rs551521305 Chr.9: 37779277-37779277 on Build GRCh38
rs1031532545 Chr.9: 37779625-37779625 on Build GRCh38
rs938940334 Chr.9: 37779402-37779402 on Build GRCh38
rs987504617 Chr.9: 37779456-37779456 on Build GRCh38
rs974690507 Chr.9: 37779679-37779679 on Build GRCh38
rs1056911406 Chr.9: 37779738-37779738 on Build GRCh38
rs1047914905 Chr.9: 37779526-37779526 on Build GRCh38
rs753333476 Chr.9: 37779357-37779357 on Build GRCh38
rs1035433223 Chr.9: 37779566-37779566 on Build GRCh38
rs35738735 Chr.9: 37779508-37779509 on Build GRCh38
rs549207852 Chr.9: 37779635-37779635 on Build GRCh38
rs569527884 Chr.9: 37779522-37779522 on Build GRCh38
rs1022902946 Chr.9: 37779598-37779598 on Build GRCh38
rs764973143 Chr.9: 37779421-37779421 on Build GRCh38
rs559508508 Chr.9: 37779645-37779645 on Build GRCh38
rs950529927 Chr.9: 37779729-37779729 on Build GRCh38
rs978591111 Chr.9: 37779436-37779436 on Build GRCh38
rs182650669 Chr.9: 37779558-37779558 on Build GRCh38
rs554788498 Chr.9: 37779696-37779696 on Build GRCh38
rs994982849 Chr.9: 37779271-37779271 on Build GRCh38
rs951138526 Chr.9: 37779328-37779328 on Build GRCh38
rs139470852 Chr.9: 37779507-37779508 on Build GRCh38
rs1039268945 Chr.9: 37779502-37779502 on Build GRCh38
rs185234870 Chr.9: 37779407-37779407 on Build GRCh38
rs1045040256 Chr.9: 37779364-37779365 on Build GRCh38
rs12002972 Chr.9: 37779442-37779442 on Build GRCh38
rs920571697 Chr.9: 37779688-37779688 on Build GRCh38
rs11420298 Chr.9: 37779462-37779463 on Build GRCh38
rs533644778 Chr.9: 37779405-37779407 on Build GRCh38

View Full Product Details